Sigma-Aldrich
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > Second Messenger Systems
Cell Signaling & Neuroscience
Second Messenger Systems
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Second Messenger Systems Involved in Reward and Addiction
With repeated psychostimulant administration, these are changes in dopaminergic and glutamatergic transmission in the nucleus accumbens. The figure depicts the major second messenger systems in the nucleus accumbens activated by dopamine and glutamate that are influenced by acute and/or repeated psychostimulant injections.