Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > Structures of Three Chemokine Receptors
Cell Signaling & Neuroscience
Structures of Three Chemokine Receptors
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Structures of Three Chemokine Receptors
Distinctions are shown between the C-X-C chemokine receptors (CXC-Rs), the C-C chemokine receptors (CC-Rs), and the Duffy blood group antigen receptor (Duffy antigen). Generally, CXC-Rs will bind only selective C-X-C chemokines and CC-Rs will bind only selective C-X-C chemokines and CC-Rs will bind only selective C-C chemokines, but the Duffy antigen binds both C-X-C and C-C chemokines.
Back to Top