Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > Transcription Factor Regulation
Cell Signaling & Neuroscience
Transcription Factor Regulation
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Transcription Factor Regulation (CREB, Elk-1 and c-Fos)
The transcription factor CREB binds to the cAMP response element (CRE) and activates gene transcription in response to a wide variety of extracellular signals (including growth factors, hormones and neurotransmitters). Transcriptional activation of CREB is controlled through phosphorylation at Ser133 by p90Rsk and the p44/42 MAP kinase.
The transcriptional activity of the proto-oncogene c-Fos has been implicated in cell growth, differentiation, and development. Fos is induced by many stimuli, ranging from mitogens to pharmacological agents. c-Fos has been shown to be associated with another proto-oncogene, c-Jun, and together they bind to the AP-1 binding site to regulate gene transcription. Like CREB, c-Fos is regulated by p90Rsk.
Elk-1 is a transcription factor that is activated by the MAP kinase-mediated phosphorylation of a Ser/Thr cluster at the carboxyl terminus. Activated Elk-1 binds to the serum response element (SRE) to induce gene transcription in response to serum and growth factors. Recent studies have also demonstrated that Elk-1 is a target for the stress-activated kinase SAPK/JNK.
Back to Top
References:
Herdegen, T., and Leah, J.D., Inducible and constitutive transcription factors in the mammalian nervous system: control of gene expression by Jun, Fos and Krox, and CREB/ATF proteins. Brain Res. Brain Res. Rev., 28, 370-490 (1998).
Davis, R.J., Transcriptional regulation by MAP kinases. Mol. Reprod. Dev., 42, 459-467 (1995).