Sigma-Aldrich
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > Voltage-Gated Ion Channel
Cell Signaling & Neuroscience
Voltage-Gated Ion Channel
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Voltage-Gated Ion Channel: Activation States
In the closed resting state, the cell membrane is polarized, the extracellular portion of the pore is closed while the intracellular gating mechanism remains open. Upon activation, the membrane depolarizes, opening the extracellular portion of the pore and allowing the ion to enter the cell. The refractory period occurs while the membrane is still depolarized, the gating mechanism has closed the intracellular portion of the pore, rendering the channel inactive.