Obesity Research
Neurotransmission
|
Neuropeptides, growth hormones, and biogenic amines can contribute to obesity and its treatment in certain individuals. Neuropeptide Y (NPY), a neuronal signaling peptide that selectively binds to Y1 and Y5 neuropeptide Y receptors in the hypothalamus, induces orexigenic signals, thereby increasing food intake and decreasing energy expenditure. Ghrelin acts as an appetite-stimulating hormone and is implicated in both short- and long-term appetite and body weight regulation. Amphetamines such as ephedrine, phentermine, and mazindol were quickly shown to display food intake and appetite suppressing qualities. However many of these drugs were abandoned as treatment for obesity due to their strong stimulant properties. Other drugs such as fenfluramine and dexfenfluramine were later created. Both had a similar structure to the earlier amphetamines but demonstrated no stimulant activity. However, patients using either or a combination of both fenfluramine and dexfenfluramine developed valvular disease.
Other anti-obesity therapies have also proven unsuccessful. Serotonin reuptake inhibitors such as fluoxetine, fluvoxamine, and setraline produce weight loss but significant weight regain is shown despite continued use of the drugs. Norephedrine has been removed from weight loss drugs due to its risk of hemorrhagic stroke and ephedrine has been removed as an over-the-counter drug due to its undesirable side effects. The development of thermogenic drugs, which increase metabolic rates safely without increasing heart rate or blood pressure, has proven difficult. But the development of b3-adrenoceptor or 5-HT2C serotonin receptor agonists as anti-obesity treatments still holds promise.
Neuropeptide Y and Peptide YY Products
Vasoconstrictor; brain peptide that inhibits Ca2+-activated K+ channels in vascular smooth muscle; implicated in the control of blood pressure, sexual behavior and food intake; inhibits cholecystokinin- and secretin-stimulated pancreatic secretion.
| Neuropeptide Y and Peptide YY Products |
| Product # |
Product Name |
Notes |
Add to Cart |
| N5017 |
Neuropeptide Y human (hNPY; NPY) |
|
|
| N3266 |
Neuropeptide Y porcine (NPY) |
|
|
| N6269 |
Neuropeptide Y sheep (NPY) |
|
|
| N9398 |
N-Acetyl-[Leu28,Leu31]-Neuropeptide Y, Fragment 24-36 |
Selective Y2 neuropeptide Y receptor agonist. |
|
| N6146 |
[Leu31,Pro34]-Neuropeptide Y human |
Selective Y1 and Y5 neuropeptide Y receptor agonist. |
|
| N7768 |
[Leu31,Pro34]-Neuropeptide Y porcine |
Selective Y1 and Y5 neuropeptide Y receptor agonist. |
|
| N8154 |
[Ala31α-Aminoisobutyryl32]-Neuropeptide Y, porcine ([Ala31, Aib32]-NPY Porcine) |
Y5 Neuropeptide Y receptor agonist; exhibits high affinity and selectivity. |
|
| N8404 |
Neuropeptide Y Fragment 19-23-[Gly1, Ser3, Gln4, Thr6, Ala31, α-Aminoisobutyryl-α-methyl-Ala32, Gln34]-Pancreatic Polypeptide human |
Highly selective Y5 neuropeptide Y receptor agonist. |
|
| N9407 |
Neuropeptide Y Fragment 3-36 human, rat, (NPY 3-36) |
Endogenous neuropeptide; selectively binds to Y2 and Y5 neuropeptide Y receptors. |
|
| N6521 |
Neuropeptide Y Fragment 13-36 porcine, (NPY 13-36) |
Selective Y2 neuropeptide Y receptor agonist. |
|
| N9409 |
Neuropeptide Y Fragment 22-36 (Ser-Ala-Leu- Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-NH2) |
NPY fragment; acts as potent inhibitor of calmodulin-stimulated phosphodiesterase. |
|
| P9778 |
Pancreatic Polypeptide bovine |
Y4 Neuropeptide Y receptor agonist. |
|
| P4488 |
Pancreatic Polypeptide Fragment 1-17, [Ala31,α-Aminoisobutyryl32]-Neuropeptide Y Fragment 18-36 |
Neuropeptide Y receptor agonist. |
|
| P4363 |
Pancreatic Polypeptide Rana tempraria |
36 amino acid peptide regulating food intake, circadian rhythms, mood, blood pressure, intestinal secretion and gut mobility. |
|
| P1306 |
Peptide YY human |
Gut hormone; inhibits both secretin and cholecystokinin-stimulated pancreatic secretion; endogenous nonselective NPY receptor agonist. |
|
| P5801 |
Peptide YY porcine (PYY) |
Gut hormone; inhibits both secretin and cholecystokinin-stimulated pancreatic secretion; endogenous nonselective NPY receptor agonist. |
|
| P220 |
Peptide YY Fragment 3-36 human |
Y2/Y5 neuropeptide Y receptor agonist. |
|
| P3365 |
Peptide YY Fragment 13-36 human trifluoroacetate salt |
Analog that mimics peptide YY (PYY) effects. |
|
| P6613 |
Peptide YY Fragment 13-36 porcine, rat |
Supresses the noradrenaline release from sympathetic nerve endings and competes with NPY for binding sites in rat brain. |
|
| P4107 |
[Leu31,Pro34]-Peptide YY |
Y1,Y4, and Y5 neuropeptide Y receptor agonist. |
|
| P3982 |
[Pro34]-Peptide YY |
NPY agonist with a preference for the Y1 and Y5 subtypes. |
|
| N8648 |
Neuropeptide Y Antagonist (1229U91) |
High affinity and selective Y1 neuropeptide Y receptor antagonist. |
|
| B174 |
BIBP 3226 |
Selective non-peptide Y1 neuropeptide Y receptor antagonist. |
|
| G8165 |
Galanin (1-13)-Neuropeptide Y (25-36) amide, (m32) |
Chimeric peptide that is a high affinity galanin receptor antagonist; this peptide is also recognized by neuropeptide Y receptors and is a Y2 neuropeptide Y agonist in rat vas deferens. |
|
| Neuropeptide Y Antibody |
| Product # |
Product Name |
Species Reactivity |
Immunogen |
Application |
Add to Cart |
| N9528 |
Anti-Neuropeptide Y (NPY), Rabbit whole antiserum |
Rat, pig |
Synthetic porcine NPY |
Immunochemistry, Radioimmunoassay, Dot Blot |
|
back to top
Galanins
Galanin is co-localized with acetylcholine and inhibits its release; inhibits the release of glutamic acid, decreases excitability of spinal neurons; blocks voltage-activated Ca2+ channels; linked to behavioral and cognitive deficits in Alzheimer's disease.
| Product # |
Product Name |
Notes |
Add to Cart |
| G0278 |
Galanin human |
|
|
| G5773 |
Galanin porcine |
|
|
| G8272 |
Galanin rat |
|
|
| G112 |
Galanin Fragment 1-16, porcine, rat |
Neuropeptide that inhibits the release of acetylcholine, dopamine, norepinephrine and other neurotransmitters. |
|
| G8916 |
[2-Aminobenzoyl-Gly1]-Galanin fragment, (1-10)-Lys(retro-m-Nitro-Tyr) amide, human, trifluoroacetate salt |
Substrate for a galanin degrading zinc-metallopeptidase from bovine spinal cord. |
|
| G9166 |
[D-Trp2]-Galanin fragment 1-29 human |
Exhibits high affinity for the GALR2 galanin receptor. |
|
| G9041 |
Galanin Fragment (1-13)-Bradykinin Fragment, (2-9) Amide trifluoroacetate salt |
Antagonist for galanin having a high specificity and affinity to the binding sites |
|
| G8041 |
Galanin-Like Peptide Fragment 1-60 porcine, trifluoroacetate salt |
Endogenous ligand that preferentially binds to the GALR2 galanin receptor. |
|
| G5024 |
Big Gastrin I human (Gregory Structure) |
Storage form of gastrin that also has biological activity. |
|
| G9020 |
Gastrin I human (Gastrin I, heptadecapeptide) |
Produced in the stomach; stimulates gastric acid secretion. |
|
| G1276 |
Gastrin I rat |
Produced in the stomach; stimulates gastric acid secretion. |
|
| G9145 |
[Leu15]-Gastrin I human ([Leucine15]-Gastrin I, heptadecapeptide; [Leucine15]-Little gastrin; [Leu15]-HG-17; LHG-17 |
Gastrin I analog with full biological activity and improved stability in aqueous solutions. Leu15 replaces Met15, which is readily oxidized resulting in loss of biological activity. |
|
| G0267 |
Mini Gastrin I human (HG-13) |
CCKB/gastrin receptor agonist. Corresponds to fragment 5-17 of human gastrin I. This form of gastrin has been isolated from tumor tissue. |
|
back to top
Growth Hormone Releasing Factors
Hypothalamic peptide that stimulates release of somatotropin from anterior pituitary.
| Product # |
Product Name |
Notes |
Add to Cart |
| G0644 |
Growth Hormone Releasing Factor bovine (GRF) |
|
|
| G8895 |
Growth Hormone Releasing Factor human (GRF) |
|
|
| G0769 |
Growth Hormone Releasing Factor porcine (GRF) |
|
|
| G6646 |
Growth Hormone Releasing Factor rat (GRF) |
|
|
| G6771 |
Growth Hormone Releasing Factor Fragment, 1-29 amide human |
Amidated GRF fragment that is equipotent to GRF in releasing somatotropin from anterior pituitary. |
|
| G4535 |
[D-Lys3]-GHRP-6 ([D-Lys3]-Growth Hormone Releasing Peptide 6 |
GHRP-6 antagonist. |
|
| G8903 |
Ghrelin rat |
Endogenous ligand to the orphan growth-hormone secretagogue receptor that stimulates the release of growth hormone from the pituitary. |
|
| G3902 |
Ghrelin human |
Endogenous ligand to the orphan growth-hormone secretagogue receptor that stimulates the release of growth hormone from the pituitary. |
|
back to top
Tachykinins
| Product # |
Product Name |
Notes |
Add to Cart |
| S6883 |
Substance P acetate salt hydrate |
NK1 tachykinin receptor agonist; potent vasodilator and hypotensive agent; induces salivation; increases capillary permeability; induces mast cell degranulation; putative neurotransmitter in sensory (pain) afferents. |
|
| S3144 |
[D-Arg1,D-Phe5,D-Trp7,9,Leu11]-Substance P (Antagonist D) |
NK1 tachykinin receptor antagonist |
|
| S0274 |
[D-Arg1,D-Trp7,9,Leu11]-Substance P acetate salt (Spantide I) |
Potent NK1 tachykinin receptor antagonist. |
|
| S6636 |
[Sar9]-Substance P |
NK1 tachykinin receptor agonist. |
|
| S3672 |
[Sar9,Met(O2)11]-Substance P |
Highly selective NK1 tachykinin receptor agonist. |
|
| S2275 |
Acetyl-[Arg6,Sar9,Met(O2)11]-Substance P, Fragment 6-11 |
Selective NK1 tachykinin receptor agonist. |
|
| S2314 |
δ-Aminovaleryl-[Pro9,Leu(N-Me)10]-Substance P, Fragment 7-11 |
Selective NK1 tachykinin receptor agonist. |
|
back to top
Adrenergics
| Product # |
Product Name |
Notes |
Add to Cart |
| B169 |
BRL 37344 sodium salt (BRL 37344A) |
Selective β3-adrenoceptor agonist. |
|
| C125 |
(±)-CGP-12177 hydrochloride |
Full agonist at β1/2-adrenoceptors and partial agonist at β3-adrenoceptors receptors. |
|
| C5976 |
CL 316,243 |
β3-Adrenoceptor agonist; anti-obesity agent. Sold with the exclusive permission of Wyeth-Ayerst Pharmaceticals. |
|
| A7257 |
(–)-Norepinephrine |
Adrenergic neurotransmitter. |
|
| S8688 |
SR 59230A oxalate salt |
β3-Adrenoceptor antagonist. |
|
back to top
Biogenic Amine Transport Inhibitors
Effects of Biogenic Amines on Food Intake and Obesity
| Product # |
Product Name |
Notes |
Add to Cart |
| A164 |
Alaprocate hydrochloride |
Potent and selective serotonin uptake inhibitor. |
|
| A8404 |
Amitriptyline hydrochloride |
Tricyclic antidepressant; inhibits the norepinephrine and serotonin transporters; high in vitro affinity for α1-adrenoceptors, serotonin and muscarinic acetylcholine receptors. |
|
| A129 |
Amoxapine |
Tricyclic antidepressant; inhibits uptake of norepinephrine; inhibits 5-HT2 serotonin receptors. |
|
| B102 |
Bupropion |
Inhibits the dopamine and norepinephrine transporters; does not inhibit the serotonin transporter; antidepressant. |
|
| C7861 |
Citalopram hydrobromide |
Potent and selective serotonin reuptake inhibitor; antidepressant. |
|
| C7291 |
Clomipramine hydrochloride |
Tricyclic antidepressant; inhibits serotonin and norepinephrine transporters. |
|
| D3900 |
Desipramine hydrochloride |
Tricyclic antidepressant that is a more potent inhibitor of norepinephrine reuptake than of serotonin reuptake. |
|
| D4526 |
Doxepin hydrochloride |
Tricyclic antidepressant that is a more potent inhibitor of norepinephrine uptake than of serotonin reuptake; antagonist at H1 histamine, muscarinic cholingergic and α-adrenergic receptors. |
|
| F132 |
Fluoxetine hydrochloride |
Selective serotonin reuptake inhibitor; antidepressant. |
|
| F1553 |
S(+)-Fluoxetine hydrochloride |
Selective serotonin reuptake inhibitor. |
|
| F1678 |
R(–)-Fluoxetine hydrochloride |
Selective serotonin reuptake inhibitor. |
|
| F133 |
Norfluoxetine hydrochloride |
Fluoxetine metabolite; more potent inhibitor of serotonin reuptake than fluoxetine with half-life in vivo of 5-7 days. |
|
| F2802 |
Fluvoxamine maleate |
Selective serotonin reuptake inhibitor; antidepressant. |
|
| G9659 |
GBR 12935 dihydrochloride |
Inhibits the dopamine and norepinephrine transporters; does not inhibit the serotonin transporter; binds to a nondopaminergic piperazine site in blood platelets and brain that has been identified as cytochrome P450. |
|
| I7379 |
Imipramine hydrochloride |
Tricyclic antidepressant; inhibits the serotonin and norepinephrine transporters; has little effect on the dopamine transporter. |
|
| I119 |
Indatraline hydrochloride (Lu 19-005) |
Potent dopamine, norepinephrine and serotonin reuptake inhibitor. |
|
| M9651 |
Maprotiline hydrochloride |
Selective norepinephrine reuptake inhibitor. |
|
| M2017 |
Mazindol |
Anorectic agent; dopamine, norepinephrine and serotonin reuptake inhibitor. |
|
| M0443 |
Mirtazapine |
Noradrenergic and specific serotonergic antidepressant; antagonizes selective adrenergic and serotonergic receptors so that both NE release and 5-HT1A mediated serotonergic signaling are increased. |
|
| N5536 |
Nefazodone hydrochloride |
Novel antidepressant; mixed 5-HT2A serotonin receptor antagonist/serotonin reuptake inhibitor. |
|
| N151 |
Nisoxetine hydrochloride |
Potent and selective norepinephrine reuptake inhibitor. |
|
| Q109 |
6-Nitroquipazine maleate |
Potent and selective serotonin reuptake inhibitor. |
|
| N7261 |
Nortriptyline hydrochloride |
Tricyclic antidepressant that is a more potent inhibitor of norepinephrine reuptake than of serotonin reuptake; 5-HT2 serotonin receptor antagonist. |
|
| P8813 |
Protriptyline hydrochloride |
Norepinephrine reuptake inhibitor. |
|
| R6527 |
Reboxetine mesylate |
Selective norepinephrine reuptake inhibitor. |
|
| T1692 |
Tianeptine sodium-potassium salt |
Tricyclic antidepressant; enhances serotonin uptake. |
|
| T7947 |
(R)-Tomoxetine hydrochloride |
Norepinephrine reuptake inhibitor. |
|
| T6154 |
Trazodone hydrochloride |
Antidepressant that potentiates the activity of serotonin reuptake inhibitors and has full 5-HT2C serotonin receptor agonist activity; metabolized to the 5-HT1 serotonin agonist 1-(3-Chlorophenyl) piperazine. |
|
| T3146 |
Trimipramine maleate |
Antidepressant; serotonin and norepinephrine reuptake inhibitor. |
|
| Z101 |
Zimelidine dihydrochloride |
Serotonin reuptake inhibitor. |
|
back to top
Serotonergics
| Product # |
Product Name |
Notes |
Add to Cart |
| C5554 |
1-(3-Chlorophenyl)piperazine hydrochloride (m-CPP) |
5-HT2c serotonin receptor agonist; metabolite of trazodone. |
|
| C238 |
S(–)-Cyanopindolol hemifumarate salt |
5-HT1A serotonin receptor antagonist; β3-adrenoceptor antagonist. |
|
| D7412 |
(±)-2,5-Dimethoxy-4-bromoamphetamine hydrobromide ((±)-DOB hydrobromide) |
Hallucinogenic; 5-HT2 serotonin receptor agonist. |
|
| D101 |
(±)-DOI hydrochloride ((±)-1-(2,5-Dimethoxy-4-iodophenyl)- 2-aminopropane hydrochloride) |
Potent and selective 5-HT2 serotonin receptor agonist that crosses the blood-brain barrier. |
|
| D153 |
R(–)-DOI hydrochloride (–)-1-(2,5-Dimethoxy-4-iodophenyl)-2-aminopropane hydrochloride |
Potent and selective 5-HT2 serotonin receptor agonist that crosses the blood-brain barrier; more potent enantiomer of (±)-DOI hydrochloride (D101). |
|
| S006 |
Ketanserin tartrate salt (R 41468 tartrate salt) |
Selective 5-HT2 serotonin receptor antagonist. |
|
| L107 |
LY-53,857 maleate salt |
Potent and selective 5-HT2 serotonin receptor antagonist. Manufactured and sold with the permission of Eli Lilly and Company. |
|
| M110 |
α-Methylserotonin maleate salt (α-Methyl-5-hydroxytryptamine maleate salt) |
5-HT2 serotonin receptor agonist. |
|
| S003 |
1-(1-Naphthyl)piperazine hydrochloride |
5-HT1 serotonin receptor agonist; 5-HT2 serotonin receptor antagonist. |
|
| P126 |
Pirenperone (R-47,465) |
5-HT2 serotonin receptor antagonist. |
|
| S180 |
SB 206553 hydrochloride |
Potent 5-HT2B/5-HT2C serotonin receptor antagonist. Manufactured and sold under exclusive license from SmithKline Beecham Pharmaceuticals. |
|
| S8061 |
SB 242084 dihydrochloride |
Selective 5-HT2c serotonin receptor antagonist; crosses the blood-brain barrier. Manufactured and sold with the permission of SmithKline Beecham Pharmaceuticals. |
|
| V1889 |
VER-3323 hemifumarate salt |
5-HT2C/5-HT2B serotonin receptor agonist. |
|
back to top
GABAergic
| Product # |
Product Name |
Notes |
Add to Cart |
| A2129 |
γ-Aminobutyric acid |
Major inhibitory neurotransmitter in brain; GABAA and GABAB receptor agonist; increases Cl- conductance. |
|
back to top
Stimulants
| Product # |
Product Name |
Notes |
Add to Cart |
| A5880 |
D-Amphetamine sulfate salt ((+)-α-Methyl-phenethylamine sulfate salt; Dextroamphetamine sulfate salt) |
Induces release of catecholamines and serotonin by displacing the monoamines from their vesicular storage sites; blocks catecholamine reuptake; more active isomer of amphetamine. |
|
| A1263 |
DL-Amphetamine sulfate salt ((±)-α-Methyl-phenethylamine sulfate salt) |
Induces release of catecholamines and serotonin by displacing the monoamines from their vesicular storage sites; blocks catecholamine reuptake. DEA Schedule II. |
|
| C222 |
Cathine hydrochloride (Pseudonorephedrine hydrochloride) from Khat shrub (Arabia, Ethiopia) |
Anorexic; with pharmacological effects similar to those of the amphetamines. DEA Schedule IV. |
|
| C3196 |
S(–)-Cathinone hydrochloride (α-Aminopropiophenone hydrochloride) |
Alkaloid that is the major central nervous system stimulant of the Khat shrub. Like amphetamine, it is a potent releaser of norepinephrine and dopamine from their intracellular stores. DEA Schedule I. |
|
| C8912 |
Cocaine free base (Ecgonine methyl ester benzoate) |
Inhibits the dopamine, norepinephrine, and serotonin transporters with KIs of approx. 300 nM, 900 nM, and 400 nM, respectively. Unlike amphetamines, it has no effect on catecholamine release. DEA Schedule II. |
|
| C5776 |
Cocaine hydrochloride Ecgonine methyl ester benzoate hydrochloride) |
Inhibits the dopamine, norepinephrine, and serotonin transporters with KIs of approx. 300 nM, 900 nM, and 400 nM, respectively. Unlike amphetamines, it has no effect on catecholamine release. DEA Schedule II. |
|
| E2750 |
(+)-Pseudoephedrine hydrochloride ((+)-ψ-Ephedrine hydrochloride; d-Isoephedrine hydrochloride) |
Non-selective adrenergic agonist; decongestant. |
|
| F8507 |
(±)-Fenfluramine hydrochloride |
Neurotoxic for prolonged administration or at high dosage; releases serotonin from axon terminals by a nonexocytotic mechanism; anorectic. DEA Schedule IV. |
|
| F112 |
(+)-Fenfluramine hydrochloride |
Neurotoxic on prolonged administration or at high dosage; releases serotonin from axon terminals by a nonexocytotic mechanism; anorectic. DEA Schedule IV. |
|
| H1392 |
Harmaline hydrochloride dihydrate |
CNS stimulant; may act through NMDA receptors. |
|
| H8646 |
Harmine |
CNS stimulant. |
|
| I7003 |
Ibogaine hydrochloride |
Alkaloid isolated from the African shrub, Tabernanthe iboga. CNS system stimulant; competitive inhibitor of serotonin and dopamine transport. DEA Schedule I. |
|
| M8750 |
(+)-Methamphetamine hydrochloride (d-N,α-Dimethylphenethylamine hydrochloride; d-Desoxyephedrine hydrochloride; Methylamphetamine hydrochloride) |
Sympathomimetic with more potent central effects than amphetamine. It is transported into terminals via the monoamine transporters and induces release of dopamine, norepinephrine, epinephrine, and serotonin. Serotonergic neurotoxin. DEA Schedule II. |
|
| M3404 |
(±)-p-Methoxyamphetamine hydrochloride (PMA hydrochloride) |
CNS stimulant and hallucinogen; more potent transport inhibitor and releaser of serotonin than of dopamine. DEA Schedule I. |
|
| M5037 |
2-(Methylamino)propiophenone hydrochloride (Methcathinone hydrochloride) |
Potent amphetamine-like stimulant; dimethylpropion metabolite. DEA Schedule I |
|
| M200 |
(±)-N-Methyl-1-(3,4-methylenedioxyphenyl)-2-butanamine hydrochloride (MBDB hydrochloride) |
Non-hallucinogenic; CNS stimulant; analog of MDMA. DEA Schedule I. |
|
| M2892 |
Methylphenidate hydrochloride (Ritalin hydrochloride) |
CNS stimulant; releases catecholamines; potent inhibitor of dopamine reuptake. DEA Schedule II. |
|
| M6810 |
D-threo-Methylphenidate hydrochloride |
CNS stimulant; more potent enantiomer of Methylphenidate. DEA Schedule II. |
|
| M6935 |
L-threo-Methylphenidate hydrochloride |
CNS stimulant; less potent enantiomer of Methylphenidate. DEA Schedule II. |
|
back to top back to Obesity Research