|
The type of "impurities" in a sample depends on whether or not the sample was purified by reverse phase HPLC. The following summarizes the difference between the two cases.
Impurities found in Non-Purified Peptides:
- Deletion sequences
- Truncation sequences
- Incompletely deprotected sequences
- Sequences modified during cleavage (reattachment of protecting groups at other locations on the peptide)
- DTT (dithiothreitol)
- TFA (trifluoroacetic acid)
- Acetic acid
- Peptides that have undergone side reactions such as proline isomerization or isoaspartimide formation, etc.
Impurities found in Purified Peptides:
- Deletion sequences
- Truncation sequences
- Incompletely deprotected sequences
- Sequences modified during cleavage (reattachment of protecting groups at other locations on the peptide)
- TFA (trifluoroacetic acid)
- Peptides that have undergone side reactions such as proline isomerization or aspartimide formation etc.
Peptides and Proteins homepage
|