Life Science > Cell Biology > Arrays & Interaction Profiling > Panorama® Antibody Arrays
Arrays & Interaction Profiling
Panorama® Antibody Arrays
Capture!
Panorama Antibody Array Technology
Protein expression profiling... broad and fast
  • Rapid, multiplex analysis of protein expression
  • Antibodies react with human, mouse, and rat proteins
  • Compatible with Cy™3/Cy5 detection
  • Use serum, cell or tissue samples
The development of antibody-based technologies has been driven by the often-poor correlation between mRNA and protein expression. Sigma’s Panorama Antibody Array technology enables researchers to screen expression changes in large numbers of proteins in significantly less time than it would take using conventional protein detection methods due to the antibody detection sensitivity.
Panorama Antibody Microarray kits are designed for studying protein expression in cell or tissue extracts. The Panorama Antibody Microarray kits encompass the areas of cell signaling proteins, kinases and phosphatases of the MAPK and PKC pathways, chromatin remodeling and gene expression, including transcription factors, histone modifying enzymes, and various P53 pathways. With market-leading content, the Panorama Antibody Array - XPRESS Profiler725 provides a high-throughput tool for multiplex expression profiling of the most important proteome-related activities.
XPRESS Profiler725 | Cell Signaling | Gene Regulation | MAPK & PKC Pathways | p53 Pathways
Frequently Asked Questions | Data | Literature | Webinar | Ordering Information
XPRESS Profiler725 (Product No. XP725)
Panorama Antibody Array XPRESS - Profiler725 kit is a high-throughput tool for multiplex protein expression profiling. The kit contains 725 highly specific antibodies that detect human, mouse, and rat proteins associated with a variety of key cell signaling and gene regulation pathways.
Cell Signaling (Product No. CSAA1)
Kit contains 224 antibodies to key cellular proteins with a special emphasis on cell signaling proteins. These antibodies represent biological pathways including apoptosis, cell cycle, neurobiology, cytoskeleton, signal transduction, and nuclear proteins.
Gene Regulation (Product No. GRAA2)
Kit contains 112 different antibodies, representing families of proteins found in the nucleus including transcription factors, histone modifying enzymes, and antibodies that specifically recognize histone modification.
MAPK & PKC Pathways (Product No. MPAA3)
Kit contains 84 different antibodies specific for kinases and phosphatases of the MAPK and PKC pathways.
p53 Pathways (Product No. PPAA4)
Kit contains 112 different antibodies involved in various p53 pathways.
back to top
Data
Protein Expression in F9 Mouse Stem Cells Induced by Retinoic Acid
Protein Expression in F9 Mouse Stem Cells Induced by Retinoic Acid Image
Figure 1. Expression of proteins in F9 mouse stem cells induced to differentiate by retinoic acid utilizing the Panorama Antibody Array-Cell Signaling. The total cell extract of F9 cells non-treated or treated for four days with retinoic acid were prepared and labeled with Cy3 and Cy5, respectively. Equal amounts of Cy3 or Cy5 labeled protein were incubated on the slide. The experiment was repeated at least three times. Proteins marked in red were up-regulated while those in green were down-regulated.
Differential Expression between Different Tissue Types
Differential Expression between Different Tissue Types
Figure 2. Differential protein expression in mouse tissues utilizing the Panorama Antibody Array-Cell Signaling. Equal concentration of mouse tissue extract (5 µg/mL) from liver, lung, and cerebellum were incubated on different slides.
Ordering Information
Product No. Description Add to Cart
XP725 Panorama Antibody Microarray - XPRESS Profiler725 Kit
CSAA1 Panorama Antibody Microarray - Cell Signaling Kit
GRAA2 Panorama Antibody Microarray - Gene Regulation Kit
MPAA3 Panorama Antibody Microarray - MAPK & PKC Pathways Kit
PPAA4 Panorama Antibody Microarray - p53 Pathways Kit
Visit the Arrays & Interaction Profiling homepage
back to top