Sigma-Aldrich
Life Science > Cell Biology > Arrays & Interaction Profiling > Panorama® Antibody Arrays > Panorama® Antibody Array – XPRESS Profiler725
Arrays & Interaction Profiling
Panorama® Antibody Array – XPRESS Profiler725
The Panorama Antibody Array - XPRESS Profiler725 is designed for studying protein expression in cell or tissue extracts. The antibody array contains 725 different antibodies each spotted in duplicate on nitrocellulose-coated glass slides. These antibodies represent families of proteins known to be involved in a variety of important biological pathways, including cell signaling, gene regulation and MAPK/PKC.
  • Accurate & robust protein expression profiling of your sample in less than half a day.
  • Antibodies are spotted in high density to ensure strong signals.
  • Proprietary treatment of the slide coating minimizes background staining.
Lot Specific Information
071M4826
Antibody List (1.11 Mb PDF), Excel File (763 Kb)
Slide Analysis Software (12 Kb zipped GAL file)
Antibody Worksheet (1.34 Mb PDF)
Antibody Number (108 Kb PDF)
019K4771
Antibody List (1.3 Mb PDF), Excel File (809 Kb)
Slide Analysis Software (10 Kb zipped GAL file)
Antibody Worksheet (1.44 Mb PDF)
Antibody Number (135 Kb PDF)
059K4766
Antibody List (1.24 Mb PDF), Excel File (644 Kb)
Slide Analysis Software (10 Kb zipped GAL file)
Antibody Worksheet (1.44 Mb PDF)
Antibody Number (146 Kb PDF)
019K4771
Antibody List (1.3 Mb PDF), Excel File (809 Kb)
Slide Analysis Software (10 Kb zipped GAL file)
Antibody Worksheet (1.44 Mb PDF)
Antibody Number (135 Kb PDF)
108K4858
Antibody List (1.3 Mb PDF), Excel File (614 Kb)
Slide Analysis Software (11 Kb zipped GAL file)
Antibody Worksheet (1.39 Mb PDF)
Antibody Number (126 Kb PDF)
089K4791
Antibody List (527 Kb PDF), Excel File (294 Kb)
129K4830
Antibody List (527 Kb PDF), Excel File (294 Kb)
129K4831
Antibody List (527 Kb PDF), Excel File (294 Kb)
Frequently Asked Questions
View Recent Literature
Ordering Information
Product No. Product Description Technical Bulletin
XP725
Panorama Antibody Microarray - XPRESS Profiler725 Kit Technical_Bulletin_icon
Please note that featured antibody content is lot specific and is subject to change.
Antibody Microarray - p53 Pathways
Kit Contents
  • Panorama Antibody Slides - EXPRESS Profiler725 (2 Each)
  • QuadriPERM® Cell Culture Vessel (2 Each)
  • Extraction/Labeling Buffer
  • Protease Inhibitor Cocktail
  • Phosphatase Inhibitor Cocktail II
  • Benzonase®, Ultrapure
  • Array Incubation Buffer
  • Phosphate Buffered Saline, pH 7.4, with Tween® 20 (Washing Buffer)
  • SigmaSpin™ Post-Reaction Clean-Up Columns
  • Collection Tubes, Polypropylene, 2 ml
  • Panorama Antibody List - XPRESS Profiler725
Panorama is a registered trademark of Sigma-Aldrich Co. LLC
SigmaSpin, CelLytic and NuCLEAR are trademarks of Sigma-Aldrich Co. LLC
ScanArray Express is a registered trademark of PerkinElmer.
GenePix Pro is a registered trademark of Molecular Devices.
Benzonase® Nuclease is manufactured by Merck KGaA and its affiliates. 
Benzonase® is a registered trademark of Merck KGaA, Darmstadt, Germany.
Cy is a trademark of Amersham Pharmacia Biotech, UK Ltd.
QuadriPERM is a registered trademark of WC Heraeus Instrument GmbH.
TWEEN is a registered trademark of Unigema, a business unit of ICI Americas Inc.
back to Antibody Arrays
back to Arrays & Interaction Profiling
back toProteomics