|
||||||
|
Panorama® Functional Protein Arrays utilize a proprietary tagging technology based on the presence and biotinylation of a BCCP (biotin-carboxy carrier protein) tag. Panorama Functional Protein Arrays can be used to identify cellular response to external signals (cell signaling), researching protein involvement in cancer development and identifying protein-protein interaction and phosphorylation events. |
|||||
|
|
||||||