Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Arrays & Interaction Profiling > Panorama® Antibody Arrays > Panorama® Antibody Array – Gene Regulation
Arrays & Interaction Profiling
Panorama® Antibody Array – Gene Regulation
Panorama Antibody Array – Gene Regulation I is designed for studying protein expression in cell or tissue extracts. The antibody array contains 112 different antibodies each spotted in duplicate on a nitrocellulose-coated slide. These antibodies represent families of proteins found in the nucleus that are involved in chromatin remodeling and regulation of gene expression and include transcription factors, histone modifying enzymes, and antibodies that specifically recognize histone modification.
  • Accurate and robust protein expression profiling of your sample in less than half a day
  • Establish protein expression profile comparisons of two samples (test vs. reference samples), simply label Cy3 or Cy5 dyes to sample and apply simultaneously to array
  • Proprietary slide treatment minimizes background staining
  • Suitable for use with Cy3/Cy5 –based detection chemistries
Array Map (91 Kb PDF)
Slide Analysis Software (3 Kb GAL file)
Antibody List (95 Kb PDF)
Frequently Asked Questions
View Recent Literature
Order Information
Product No. Description Technical Bulletin
GRAA-2 Panorama Antibody Microarray - Gene Regulation I Kit Technical_Bulletin_icon
Please note that featured antibody content is lot specific and is subject to change.
Panorama-image
back to Antibody Arrays
back to Proteomics
back to Enzyme Explorer
back to top