|
|||||||||||||
|
Panorama Antibody Array – MAPK & PKC Pathways is designed for studying protein expression in cell or tissue extracts. The antibody array contains 84 different antibodies each spotted in duplicate on a nitrocellulose-coated slide. The antibodies are specific for kinases and phosphatases of the MAPK and PKC pathways.
Array Map (87 Kb PDF) Slide Analysis Software (3 Kb GAL file) Antibody List (94 Kb PDF) Worksheet for Data Analysis and Calculations (17 Kb PDF)
back to top |
||||||||||||
|
|
|||||||||||||