Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Arrays & Interaction Profiling > Panorama® Antibody Arrays > Panorama® Antibody Array – p53 Pathways
Arrays & Interaction Profiling
Panorama® Antibody Array – p53 Pathways
Panorama Antibody Array – p53 Pathways is designed for studying protein expression in cell or tissue extracts. The array contains 112 different antibodies each spotted in duplicate on a nitrocellulose-coated slide. The immobilized antibodies represent various p53 pathways and are also available independently.
  • Accurate and robust protein expression profiling of your sample in less than half a day
  • Qualitative characterization of protein expression against 112 different antibodies
  • Establish comparative protein expression profiles between two samples.
  • Proprietary slide treatment minimizes background staining
  • Suitable for use with Cy™3/Cy5 –based detection chemistries
Antibody List (64.5 kb PDF)
Slide Analysis Software (3 Kb GAL file)
Array Map (7.75 kb PDF)
Frequently Asked Questions
View Recent Literature
Order Information
Product No. Description Technical Bulletin
PPAA4 Panorama Antibody Microarray – p53 Pathways Technical_Bulletin_icon
Please note that featured antibody content is lot specific and is subject to change.
Antibody Microarray - p53 PathwaysKit Contents
  • Panorama Antibody Slides – p53 Pathways (2 Each)
  • QuadriPERM™ Cell Culture Vessel (2 Each)
  • Extraction/Labeling Buffer
  • Protease Inhibitor Cocktail
  • Phosphatase Inhibitor Cocktail II
  • Benzonase®, Ultrapure
    Array Incubation Buffer
  • Phosphate Buffered Saline, pH 7.4, with TWEEN® 20 (Washing Buffer)
  • SigmaSpin™ Post-Reaction Clean-Up Columns
    Collection Tubes, Polypropylene, 2 ml
  • Panorama Antibody List – p53 Pathways
QuadriPERM is a trademark of WC Heraeus GmbH, Ltd.
TWEEN is a registered trademark of the ICI Group.
Benzonase is a trademark of Merck KGaA.
Cy3 is a trademark of GE Healthcare.
Panorama is a registered trademark of Sigma-Aldrich.
back to Antibody Arrays
back to Proteomics
back to Cell Signaling
back to Antibody Explorer