|
Panorama Antibody Array – p53 Pathways is designed for studying protein expression in cell or tissue extracts. The array contains 112 different antibodies each spotted in duplicate on a nitrocellulose-coated slide. The immobilized antibodies represent various p53 pathways and are also available independently.
- Accurate and robust protein expression profiling of your sample in less than half a day
- Qualitative characterization of protein expression against 112 different antibodies
- Establish comparative protein expression profiles between two samples.
- Proprietary slide treatment minimizes background staining
- Suitable for use with Cy™3/Cy5 –based detection chemistries
Antibody List (64.5 kb PDF)
Slide Analysis Software (3 Kb GAL file)
Array Map (7.75 kb PDF)
Frequently Asked Questions
View Recent Literature
| Order Information |
| Product No. |
Description |
Technical Bulletin |
| PPAA4 |
Panorama Antibody Microarray – p53 Pathways |
|
| Please note that featured antibody content is lot specific and is subject to change. |
Kit Contents
- Panorama Antibody Slides – p53 Pathways (2 Each)
- QuadriPERM™ Cell Culture Vessel (2 Each)
- Extraction/Labeling Buffer
- Protease Inhibitor Cocktail
- Phosphatase Inhibitor Cocktail II
- Benzonase®, Ultrapure
Array Incubation Buffer
- Phosphate Buffered Saline, pH 7.4, with TWEEN® 20 (Washing Buffer)
- SigmaSpin™ Post-Reaction Clean-Up Columns
Collection Tubes, Polypropylene, 2 ml
- Panorama Antibody List – p53 Pathways
QuadriPERM is a trademark of WC Heraeus GmbH, Ltd. TWEEN is a registered trademark of the ICI Group. Benzonase is a trademark of Merck KGaA. Cy3 is a trademark of GE Healthcare. Panorama is a registered trademark of Sigma-Aldrich.
back to Antibody Arrays back to Proteomics back to Cell Signaling back to Antibody Explorer
|