Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Arrays & Interaction Profiling > Panorama® Antibody Arrays > Panorama® Antibody Array – Cell Signaling
Arrays & Interaction Profiling
Panorama® Antibody Array – Cell Signaling
Panorama Antibody Array – Cell Signaling provides a comprehensive system to establish protein expression profiles in biological samples. Based on a state-of-the-art manufacturing process, the antibody microarray contains 224 human antibodies to key cellular proteins with a special emphasis on cell signaling proteins. The antibodies are spotted in duplicate on a nitrocellulose-coated glass slide that has been blocked with a proprietary solution to eliminate background. The Panorama Antibody Microarray reacts with samples from human, mouse, and rat. Additionally, the antibodies are also available separately for more extensive analysis of your sample.
  • Accurate & robust protein expression profiling of your sample in less than half a day
  • Establish relative abundance of hundreds of proteins in less than 5 hours
  • Suitable for use with Cy™3/Cy5 – based detection chemistries
  • Compatible with most commercially available scanners
Lot Specific Information
058K4796
Antibody List (155 Kb PDF), Excel file (281 Kb)
Slide Analysis Software (5 Kb zipped GAL file)
Antibody Worksheet (118 Kb PDF)
Antibody Number (63 Kb PDF)
Application Article (480 Kb PDF)
ScanArray® Express Slide Analysis Software (11 Kb TXT file)
GenePix® Pro Slide Analysis Software (18 Kb TXT file)
Technical Poster (222 Kb PDF)
Brochure (298 Kb PDF)
Frequently Asked Questions
View Recent Literature
Order Information
Product No. Description Technical Bulletin
CSAA1
Panorama® Antibody Microarray – Cell Signaling Kit Technical_Bulletin_icon
Please note that featured antibody content is lot specific and is subject to change.
Panorama® Antibody Microarray -Cell SignalingKit Contents
  • Panorama Antibody Slides – Cell Signaling (2 Each)
  • QuadriPERM Cell Culture Vessel (2 Each)
  • Extraction/Labeling Buffer
  • Protease Inhibitor Cocktail
  • Phosphatase Inhibitor Cocktail II
  • Benzonase®, Ultrapure
  • Array Incubation Buffer
  • Phosphate Buffered Saline, pH 7.4, with Tween 20 (Washing Buffer)
  • SigmaSpin™ Post-Reaction Clean-Up Columns
  • Collection Tubes, Polypropylene, 2 ml
  • Panorama Antibody List – Cell Signaling
ScanArray Express is a registered trademark of PerkinElmer.
GenePix Pro is a registered trademark of Molecular Devices.
QuadiPERM is a trademark of WC Heraeus GmbH, Ltd.
TWEEN is a registered trademark of the ICI Group.
Benzonase is a trademark of Merck KGaA.
Cy3 is a trademark of GE Healthcare.
Panorama is a registered trademark of Sigma-Aldrich.
back to Antibody Arrays
back to Arrays
back to Proteomics
back to Antibody Explorer