Sigma-Aldrich
Life Science > Cell Biology > Arrays & Interaction Profiling > Panorama® Antibody Array – Cancer Protein
Arrays & Interaction Profiling
Panorama® Antibody Array – Cancer Protein
Accurately identify your target's interaction against 130 fully functional cancer proteins with the Panorama Cancer Protein Array. Based on a proprietary tagging technology that ensures only functional proteins are immobilized, the cancer protein array is an essential research tool for unveiling details of protein involvement in cancer development.
  • Accurate representation of 130 cancer proteins.
  • Proteins are expressed in insect cells and analyzed by Western blot to verify generation of full-length proteins.
  • Compatible with probing by DNA, proteins, kinases & small molecules and with Cy™3/Cy5 –based detection chemistries.
Functional Array Technology
Cancer Protein Content (192 Kb Excel file)
Slide Analysis Software (6.71 Kb GAL file)
Slide Analysis Template (2.01 Mb Excel file)
Order Information
Product No. Product Description Technical Bulletin
HPFM2 Panorama® Human Protein Function Array – Cancer v1 Array Technical_Bulletin_icon
Cancer-array-kit-picture_v2Kit Contents
  • Panorama Human Cancer v1 Protein Array - 2 ea
  • Calmodulin - Cy5
  • Monoclonal Anti-c-Myc-Cy3 Conjugate
  • Cover slips
  • quadiPERM Vessel
  • 50 mL Conicol - 2 ea
  • Pap Jar - 4 ea
  • Assay Buffer
  • Calmodulin Buffer
  • BSA
  • 1M DTT
  • Cancer v1 Array Diskette (GAL file and analysis spreadsheet)
This product is sold under an exclusive worldwide license and a joint collaborative
agreement between Sigma-Aldrich Corp. and Procognia Limited.
back to Proteomics
back to Cancer Research
back to Enzyme Explorer