Life Science > Cell Biology > Arrays & Interaction Profiling > Panorama® Antibody Array - Kinase Protein
Arrays & Interaction Profiling
Panorama® Antibody Array - Kinase Protein
The Panorama Human Kinase v1 provides a reliable platform for identifying protein-protein interactions and phosphorylation events. Confidently explore kinase interactions with accurate detection of activity, inhibition, or substrate properties!
  • Perform multiplex assays against 152 commonly studied kinases
  • Profile interactions with other proteins, antibodies, peptides, kinases and drug candidates
  • Unique fusion tag serves to tether & uniformly orientate proteins on the streptavidin coated slide
Functional Array Technology
Slide Analysis Software (6.89 Kb GAL file)
Slide Analysis Template (1.55 Mb Excel file)
Kinase Array Content
Order Information
Product Code Product Description Technical
Bulletin
HPFM3 Panorama® Human Kinase v1 Protein Array- Technical_Bulletin_icon
Kit-image
  Kit Contents:
  • Panorama Human Kinase v1 Protein Array - 2 ea
  • Anti-phosphotyrosine - Cy5
  • Monoclonal Anti-c-Myc-Cy3 Conjugate
  • Assay Buffer
  • Functional Assay Buffer
  • BSA
  • 1M DTT
  • 100 mM ATP
  • quadiPERM® Vessel
  • Cover slips
  • 50 mL Conicol - 2 ea
  • Pap Jar - 4 ea
  • Kinase v1 Array Disc (GAL file and analysis spreadsheet)
Kinase Array Content:
Symbol Gene Name Entrez Gene ID
AAK1
AP2 associated kinase 1
22848
ACVR1
activin A receptor, type I
90
ACVR1C
activin A receptor, type IC
130399
ADCK4
aarF domain containing kinase 4
79934
ADRBK2
adrenergic, beta, receptor kinase 2
157
AKT1
v-akt murine thymoma viral oncogene homolog 1
207
ALS2CR2
amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 2
55437
APEG1
aortic preferentially expressed protein 1
10290
ARAF1
v-raf murine sarcoma 3611 viral oncogene homolog 1
369
AURKB
serine/threonine kinase 12
9212
BLK
B lymphoid tyrosine kinase
640
BMX
BMX non-receptor tyrosine kinase
660
BRD3
bromodomain containing 3
8019
BUB1
BUB1 budding uninhibited by benzimidazoles 1 homolog (yeast)
699
BUB1B
BUB1 budding uninhibited by benzimidazoles 1 homolog beta (yeast)
701
C20orf64 / PRPK
chromosome 20 open reading frame 64
112858
C20orf97
chromosome 20 open reading frame 97
57761
CAMK1G
calcium/calmodulin-dependent protein kinase IG
57172
CAMK2B
calcium/calmodulin-dependent protein kinase (CaM kinase) II beta
816
CAMK2D
calcium/calmodulin-dependent protein kinase (CaM kinase) II delta
817
CAMK2G
calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma
818
CAMK4
calcium/calmodulin-dependent protein kinase IV
814
CAMKK1
calcium/calmodulin-dependent protein kinase kinase 1, alpha
84254
CAMKK2
calcium/calmodulin-dependent protein kinase kinase 2, beta
10645
CCRK
cell cycle related kinase
23552
CDC2
cell division cycle 2, G1 to S and G2 to M
983
CDK2
cyclin-dependent kinase 2
1017
CDK4
cyclin-dependent kinase 4
1019
CDK5
cyclin-dependent kinase 5
1020
CDK7
cyclin-dependent kinase 7
1022
CDK9
cyclin-dependent kinase 9
1025
CDKN2A
cyclin-dependent kinase inhibitor 2A
1029
CHEK1
CHK1 checkpoint homolog (S. pombe)
1111
CHEK2
CHK2 checkpoint homolog (S. pombe)
11200
CLK1
CDC-like kinase 1
1195
CLK2
CDC-like kinase 2
1196
CLK3
CDC-like kinase 3
1198
COL4A3BP
Similar to collagen, type IV, alpha 3 (Goodpasture antigen) binding protein
10087
CSNK1D
casein kinase 1, delta, transcript variant 2
1453
CSNK1G1
casein kinase 1, gamma 1
53944
CSNK1G2
casein kinase 1, gamma 2
1455
CSNK2A1
casein kinase 2, alpha 1 polypeptide, transcript variant 2
1457
CSNK2A2
casein kinase 2, alpha prime polypeptide
1459
DYRK2
dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2
8445
DYRK4
dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 4
8798
FASTK
FAST kinase
10922
FES
feline sarcoma oncogene
2242
FGFR2
fibroblast growth factor receptor 2
2263
FLJ14813
hypothetical protein FLJ14813
84930
FLJ20574
hypothetical protein FLJ20574
54986
FRK
fyn-related kinase
2444
FYN
FYN oncogene related to SRC, FGR, YES
2534
GPRK6
G protein-coupled receptor kinase 6
2870
GSK3B
glycogen synthase kinase 3 beta
2932
H11
protein kinase H11
26353
HK1
hexokinase 1
3098
IKBKB
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta
3551
ILK
integrin-linked kinase
3611
JIK
STE20-like kinase
51347
KIS
kinase interacting with leukemia-associated gene (stathmin)
127933
LCK
lymphocyte-specific protein tyrosine kinase
3932
LIMK2
LIM domain kinase 2
3985
MAK
male germ cell-associated kinase
4117
MAP2K3
mitogen-activated protein kinase kinase 3
5606
MAP2K5
mitogen-activated protein kinase kinase 5
5607
MAP2K6
mitogen-activated protein kinase kinase 6
5608
MAP2K7
mitogen-activated protein kinase kinase 7
5609
MAP3K14
mitogen-activated protein kinase kinase kinase 14
9020
MAP3K6
mitogen-activated protein kinase kinase kinase 6
9064
MAP3K7
mitogen-activated protein kinase kinase kinase 7
6885
MAP4K5
mitogen-activated protein kinase kinase kinase kinase 5
11183
MAPK1
mitogen-activated protein kinase 1
5594
MAPK13
mitogen-activated protein kinase 13
5603
MAPK14
mitogen-activated protein kinase 14
1432
MAPK3
mitogen-activated protein kinase 3
5595
MAPK6
mitogen-activated protein kinase 6
5597
MAPK7
mitogen-activated protein kinase 7
5598
MAPK9
mitogen-activated protein kinase 9
5601
MAPKAPK3
mitogen-activated protein kinase-activated protein kinase 3
7867
MARK3
MAP/microtubule affinity-regulating kinase 3
4140
MATK
megakaryocyte-associated tyrosine kinase
4145
MGC16169
hypothetical protein MGC16169
93627
MGC42105
hypothetical protein MGC42105
167359
MGC45428
hypothetical protein MGC45428
166614
MKNK1
MAP kinase-interacting serine/threonine kinase 1
8569
NEK11
NIMA (never in mitosis gene a)-related kinase 11
79858
NEK3
NIMA (never in mitosis gene a)-related kinase 3
4752
PACE-1
ezrin-binding partner PACE-1
57147
PAK4
p21 (CDKN1A)-activated kinase 4
10298
PCK2
phosphoenolpyruvate carboxykinase 2 (mitochondrial)
5106
PCTK1
PCTAIRE protein kinase 1
5127
PCTK2
PCTAIRE protein kinase 2
5128
PDK1
pyruvate dehydrogenase kinase, isoenzyme 1
5163
PDK2
pyruvate dehydrogenase kinase, isoenzyme 2
5164
PDK3
pyruvate dehydrogenase kinase, isoenzyme 3
5165
PDK4
pyruvate dehydrogenase kinase, isoenzyme 4
5166
PHKG2
phosphorylase kinase, gamma 2 (testis)
5261
PIK3C3
phosphoinositide-3-kinase, class 3
5289
PIK3R1
phosphoinositide-3-kinase, regulatory subunit, polypeptide 1 (p85 alpha)
5295
PIM1
pim-1 oncogene
5292
PIM2
pim-2 oncogene
11040
PKE
PKE protein kinase
282974
PKLR
pyruvate kinase, liver and RBC
5313
PKM2
pyruvate kinase, muscle
5315
PLK
polo (Drosophila)-like kinase
5347
PRKACB
protein kinase, cAMP-dependent, catalytic, beta
5567
PRKACG
protein kinase, cAMP-dependent, catalytic, gamma
5568
PRKCB1
protein kinase C, beta 1
5579
PRKCH
protein kinase C, eta
5583
PRKCI
protein kinase C, iota
5584
PRKCN
protein kinase C, nu
23683
PRKCZ
protein kinase C, zeta
5590
PRKD2
protein kinase D2
25865
PTK2
PTK2 protein tyrosine kinase 2
5747
PTK9
PTK9 protein tyrosine kinase 9
5756
PXK
PX domain containing serine/threonine kinase
54899
RAF1
v-raf-1 murine leukemia viral oncogene homolog 1
5894
RIOK2
RIO kinase 2 (yeast)
55781
RIOK3
RIO kinase 3 (yeast)
8780
RIPK2
receptor-interacting serine-threonine kinase 2
8767
RPS6KA1
ribosomal protein S6 kinase, 90kDa, polypeptide 1
6195
RPS6KA2
ribosomal protein S6 kinase, 90kDa, polypeptide 2
6196
RPS6KA5
ribosomal protein S6 kinase, 90kDa, polypeptide 5
9252
RPS6KL1
ribosomal protein S6 kinase-like 1
83694
SGK
serum/glucocorticoid regulated kinase
6446
SGKL
serum/glucocorticoid regulated kinase-like, transcript variant 1
23678
SNARK
likely ortholog of rat SNF1/AMP-activated protein kinase
81788
SNK
serum-inducible kinase
10769
SRC
v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian)
6714
SRPK1
SFRS protein kinase 1
6732
STK11
serine-threonine kinase 11 (Peutz-Jeghers syndrome)
6794
STK16
serine-threonine kinase 16
8576
STK17B
serine-threonine kinase 17b (apoptosis-inducing)
9262
STK22C
serine-threonine kinase 22C (spermiogenesis-associated)
81629
STK22D
serine-threonine kinase 22D (spermiogenesis-associated)
83942
STK24
serine-threonine kinase 24 (STE20 homolog, yeast)
8428
STK25
serine-threonine kinase 25 (STE20 homolog, yeast)
10494
STK3
serine-threonine kinase 3 (STE20 homolog, yeast)
6788
STK33
serine-threonine kinase 33
65975
STK38
serine-threonine kinase 38
11329
STK38L
serine-threonine kinase 38 like
23012
STK6
serine-threonine kinase 6
6790
SYK
spleen tyrosine kinase
6850
TBK1
TANK-binding kinase 1
29110
TEK
TEK tyrosine kinase, endothelial
7010
TLK1
tousled-like kinase 1
9874
TTK
TTK protein kinase
7272
VRK3
vaccinia related kinase 3
51231
ZAK
sterile alpha motif and leucine zipper containing kinase AZK
51776
ZAP70
zeta-chain (TCR) associated protein kinase 70 kDa
7535
This product is sold under an exclusive worldwide license and a joint collaborative agreement between Sigma-Aldrich Corp. and Procognia Limited.
back to top
back to Proteomics
back to Cancer Research
back to Enzyme Explorer