Sigma-Aldrich
Life Science > Cell Biology > Arrays & Interaction Profiling > Panorama® Antibody Array – Kinase Protein
Arrays & Interaction Profiling
Panorama® Antibody Array – Kinase Protein
The Panorama Human Kinase v1 provides a reliable platform for identifying protein-protein interactions and phosphorylation events. Confidently explore kinase interactions with accurate detection of activity, inhibition, or substrate properties.
  • Perform multiplex assays against 152 commonly studied kinases
  • Profile interactions with other proteins, antibodies, peptides, kinases and drug candidates
  • Unique fusion tag serves to tether & uniformly orientate proteins on the streptavidin coated slide
Functional Array Technology
Slide Analysis Software (6.89 Kb GAL file)
Slide Analysis Template (1.55 Mb Excel file)
Kinase Array Content
Order Information
Product No. Description Technical Bulletin
HPFM3 Panorama® Human Protein Function Array - Kinase v1 Technical_Bulletin_icon
Kit-imageKit Contents
  • Panorama Human Kinase v1 Protein Array - 2 ea
  • Anti-phosphotyrosine - Cy5
  • Monoclonal Anti-c-Myc-Cy3 Conjugate
  • Assay Buffer
  • Functional Assay Buffer
  • BSA
  • 1M DTT
  • 100 mM ATP
  • quadiPERM® Vessel
  • Cover slips
  • 50 mL Conicol - 2 ea
  • Pap Jar - 4 ea
  • Kinase v1 Array Disc (GAL file and analysis spreadsheet)
Kinase Array Content
Symbol Gene Name Entrez Gene ID
AAK1 AP2 associated kinase 1 22848
ACVR1 activin A receptor, type I 90
ACVR1C activin A receptor, type IC 130399
ADCK4 aarF domain containing kinase 4 79934
ADRBK2 adrenergic, beta, receptor kinase 2 157
AKT1 v-akt murine thymoma viral oncogene homolog 1 207
ALS2CR2 amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 2 55437
APEG1 aortic preferentially expressed protein 1 10290
ARAF1 v-raf murine sarcoma 3611 viral oncogene homolog 1 369
AURKB serine/threonine kinase 12 9212
BLK B lymphoid tyrosine kinase 640
BMX BMX non-receptor tyrosine kinase 660
BRD3 bromodomain containing 3 8019
BUB1 BUB1 budding uninhibited by benzimidazoles 1 homolog (yeast) 699
BUB1B BUB1 budding uninhibited by benzimidazoles 1 homolog beta (yeast) 701
C20orf64 / PRPK chromosome 20 open reading frame 64 112858
C20orf97 chromosome 20 open reading frame 97 57761
CAMK1G calcium/calmodulin-dependent protein kinase IG 57172
CAMK2B calcium/calmodulin-dependent protein kinase (CaM kinase) II beta 816
CAMK2D calcium/calmodulin-dependent protein kinase (CaM kinase) II delta 817
CAMK2G calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma 818
CAMK4 calcium/calmodulin-dependent protein kinase IV 814
CAMKK1 calcium/calmodulin-dependent protein kinase kinase 1, alpha 84254
CAMKK2 calcium/calmodulin-dependent protein kinase kinase 2, beta 10645
CCRK cell cycle related kinase 23552
CDC2 cell division cycle 2, G1 to S and G2 to M 983
CDK2 cyclin-dependent kinase 2 1017
CDK4 cyclin-dependent kinase 4 1019
CDK5 cyclin-dependent kinase 5 1020
CDK7 cyclin-dependent kinase 7 1022
CDK9 cyclin-dependent kinase 9 1025
CDKN2A cyclin-dependent kinase inhibitor 2A 1029
CHEK1 CHK1 checkpoint homolog (S. pombe) 1111
CHEK2 CHK2 checkpoint homolog (S. pombe) 11200
CLK1 CDC-like kinase 1 1195
CLK2 CDC-like kinase 2 1196
CLK3 CDC-like kinase 3 1198
COL4A3BP Similar to collagen, type IV, alpha 3 (Goodpasture antigen) binding protein 10087
CSNK1D casein kinase 1, delta, transcript variant 2 1453
CSNK1G1 casein kinase 1, gamma 1 53944
CSNK1G2 casein kinase 1, gamma 2 1455
CSNK2A1 casein kinase 2, alpha 1 polypeptide, transcript variant 2 1457
CSNK2A2 casein kinase 2, alpha prime polypeptide 1459
DYRK2 dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2 8445
DYRK4 dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 4 8798
FASTK FAST kinase 10922
FES feline sarcoma oncogene 2242
FGFR2 fibroblast growth factor receptor 2 2263
FLJ14813 hypothetical protein FLJ14813 84930
FLJ20574 hypothetical protein FLJ20574 54986
FRK fyn-related kinase 2444
FYN FYN oncogene related to SRC, FGR, YES 2534
GPRK6 G protein-coupled receptor kinase 6 2870
GSK3B glycogen synthase kinase 3 beta 2932
H11 protein kinase H11 26353
HK1 hexokinase 1 3098
IKBKB inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta 3551
ILK integrin-linked kinase 3611
JIK STE20-like kinase 51347
KIS kinase interacting with leukemia-associated gene (stathmin) 127933
LCK lymphocyte-specific protein tyrosine kinase 3932
LIMK2 LIM domain kinase 2 3985
MAK male germ cell-associated kinase 4117
MAP2K3 mitogen-activated protein kinase kinase 3 5606
MAP2K5 mitogen-activated protein kinase kinase 5 5607
MAP2K6 mitogen-activated protein kinase kinase 6 5608
MAP2K7 mitogen-activated protein kinase kinase 7 5609
MAP3K14 mitogen-activated protein kinase kinase kinase 14 9020
MAP3K6 mitogen-activated protein kinase kinase kinase 6 9064
MAP3K7 mitogen-activated protein kinase kinase kinase 7 6885
MAP4K5 mitogen-activated protein kinase kinase kinase kinase 5 11183
MAPK1 mitogen-activated protein kinase 1 5594
MAPK13 mitogen-activated protein kinase 13 5603
MAPK14 mitogen-activated protein kinase 14 1432
MAPK3 mitogen-activated protein kinase 3 5595
MAPK6 mitogen-activated protein kinase 6 5597
MAPK7 mitogen-activated protein kinase 7 5598
MAPK9 mitogen-activated protein kinase 9 5601
MAPKAPK3 mitogen-activated protein kinase-activated protein kinase 3 7867
MARK3 MAP/microtubule affinity-regulating kinase 3 4140
MATK megakaryocyte-associated tyrosine kinase 4145
MGC16169 hypothetical protein MGC16169 93627
MGC42105 hypothetical protein MGC42105 167359
MGC45428 hypothetical protein MGC45428 166614
MKNK1 MAP kinase-interacting serine/threonine kinase 1 8569
NEK11 NIMA (never in mitosis gene a)-related kinase 11 79858
NEK3 NIMA (never in mitosis gene a)-related kinase 3 4752
PACE-1 ezrin-binding partner PACE-1 57147
PAK4 p21 (CDKN1A)-activated kinase 4 10298
PCK2 phosphoenolpyruvate carboxykinase 2 (mitochondrial) 5106
PCTK1 PCTAIRE protein kinase 1 5127
PCTK2 PCTAIRE protein kinase 2 5128
PDK1 pyruvate dehydrogenase kinase, isoenzyme 1 5163
PDK2 pyruvate dehydrogenase kinase, isoenzyme 2 5164
PDK3 pyruvate dehydrogenase kinase, isoenzyme 3 5165
PDK4 pyruvate dehydrogenase kinase, isoenzyme 4 5166
PHKG2 phosphorylase kinase, gamma 2 (testis) 5261
PIK3C3 phosphoinositide-3-kinase, class 3 5289
PIK3R1 phosphoinositide-3-kinase, regulatory subunit, polypeptide 1 (p85 alpha) 5295
PIM1 pim-1 oncogene 5292
PIM2 pim-2 oncogene 11040
PKE PKE protein kinase 282974
PKLR pyruvate kinase, liver and RBC 5313
PKM2 pyruvate kinase, muscle 5315
PLK polo (Drosophila)-like kinase 5347
PRKACB protein kinase, cAMP-dependent, catalytic, beta 5567
PRKACG protein kinase, cAMP-dependent, catalytic, gamma 5568
PRKCB1 protein kinase C, beta 1 5579
PRKCH protein kinase C, eta 5583
PRKCI protein kinase C, iota 5584
PRKCN protein kinase C, nu 23683
PRKCZ protein kinase C, zeta 5590
PRKD2 protein kinase D2 25865
PTK2 PTK2 protein tyrosine kinase 2 5747
PTK9 PTK9 protein tyrosine kinase 9 5756
PXK PX domain containing serine/threonine kinase 54899
RAF1 v-raf-1 murine leukemia viral oncogene homolog 1 5894
RIOK2 RIO kinase 2 (yeast) 55781
RIOK3 RIO kinase 3 (yeast) 8780
RIPK2 receptor-interacting serine-threonine kinase 2 8767
RPS6KA1 ribosomal protein S6 kinase, 90kDa, polypeptide 1 6195
RPS6KA2 ribosomal protein S6 kinase, 90kDa, polypeptide 2 6196
RPS6KA5 ribosomal protein S6 kinase, 90kDa, polypeptide 5 9252
RPS6KL1 ribosomal protein S6 kinase-like 1 83694
SGK serum/glucocorticoid regulated kinase 6446
SGKL serum/glucocorticoid regulated kinase-like, transcript variant 1 23678
SNARK likely ortholog of rat SNF1/AMP-activated protein kinase 81788
SNK serum-inducible kinase 10769
SRC v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian) 6714
SRPK1 SFRS protein kinase 1 6732
STK11 serine-threonine kinase 11 (Peutz-Jeghers syndrome) 6794
STK16 serine-threonine kinase 16 8576
STK17B serine-threonine kinase 17b (apoptosis-inducing) 9262
STK22C serine-threonine kinase 22C (spermiogenesis-associated) 81629
STK22D serine-threonine kinase 22D (spermiogenesis-associated) 83942
STK24 serine-threonine kinase 24 (STE20 homolog, yeast) 8428
STK25 serine-threonine kinase 25 (STE20 homolog, yeast) 10494
STK3 serine-threonine kinase 3 (STE20 homolog, yeast) 6788
STK33 serine-threonine kinase 33 65975
STK38 serine-threonine kinase 38 11329
STK38L serine-threonine kinase 38 like 23012
STK6 serine-threonine kinase 6 6790
SYK spleen tyrosine kinase 6850
TBK1 TANK-binding kinase 1 29110
TEK TEK tyrosine kinase, endothelial 7010
TLK1 tousled-like kinase 1 9874
TTK TTK protein kinase 7272
VRK3 vaccinia related kinase 3 51231
ZAK sterile alpha motif and leucine zipper containing kinase AZK 51776
ZAP70 zeta-chain (TCR) associated protein kinase 70 kDa 7535
This product is sold under an exclusive worldwide license and a joint collaborative agreement between Sigma-Aldrich Corp. and Procognia Limited.
back to Proteomics
back to Cancer Research
back to Enzyme Explorer