Sigma-Aldrich
Life Science > Cell Biology > Arrays & Interaction Profiling > Panorama® Protein Array - Tissue Extract
Arrays & Interaction Profiling
Panorama® Protein Array - Tissue Extract
Our Panorama® Mouse/Rat Tissue Extract Protein Array kit provides a reliable platform for the rapid evaluation of serum, hybridoma supernatant, ascites fluid, purified antibody, or fractionated sera. The mouse and rat tissue proteins spotted are extracts of total tissue as well as nuclear and cytoplasmic cell fractions. Each tissue extract and its fractions are spotted in three concentrations.
  • Efficient and accurate profiling of biological sample against 62 extracts in ~5 hours
  • High density spotting of extracts from mouse and rat tissues ensures strong signals
  • Proprietary treatment of slides eliminates background commonly associated with nitrocellulose-coated slides and extends shelf life
Tissue Extract Format
Tissue Extract List
Technical Article
Order Information
Product Code Product Description Technical
Bulletin
MRPA1 Panorama® Mouse/Rat Tissue Extract Protein Array Kit Technical_Bulletin_icon