Life Science > Cell Biology > Special Offers > Antibody Photo Contest Winners November 2008
Cell Signaling & Neuroscience
Antibody Photo Contest Winners November 2008
CONGRATULATIONS to our Antibody Photo Contest Winners, November 2008!
Image created by M. K. Kandasamy   M. K. Kandasamy, Genetics Department, University of Georgia, Athens, GA
Immunofluorescence
Arabidoposis suspension cells with the Monoclonal Anti-Actin, clone 10-B3 (MabGPa) (A0480) at a 1:100 dilution. The actin filaments were revealed with Anti-Mouse IgG-FITC (F6257) (green) and the nucleus is stained with DAPI (red).
Image created by Shyam Khatau
  Shyam Khatau, Department of Chemical and Biomolecular Engineering, Johns Hopkins University, Baltimore, MD
Immunofluorescence
Imna(-/-) mouse embryonic fibroblasts stained with Monoclonal Anti-Vimentin, clone LN-6 (V2258) at a 1:40 dilution (red), anti-actin (blue) and DAPI (yellow).
Image created by Christopher M. Hale
  Christopher M. Hale, Department of Chemical & Biomolecular Engineering Johns Hopkins University, Baltimore, MD
Immunofluorescence
Mouse adult fibroblast stained with Monoclonal Anti-Vinculin, clone VIN-11-5 (V4505) at 1:40 dilution (aqua). Actin (purple) and nuclear DNA (blue) were stained using phalloidin-Alexa Fluor 568 and DAPI, respectively.
Image created by Raleigh Kladney   Raleigh Kladney, Surgery Department, Washington University School of Medicine, St. Louis, MO
Immunofluorescence
Mouse anterior prostate stained using Monoclonal Anti-phospho-Histone H3 (pSer28), clone HTA28 (H9908) (red) at a 1:2,000 dilution, anti-phospho-S6 (pSer235/236) (green) and DAPI (blue). The figure shows a mitotic prostate epithelial cell with nuclear pH3 staining as well as cytoplasmic pS6 staining.
Image created by Kenneth Reese
  Kenneth Reese, BrainBits, Springfield, IL
Immunofluorescence
Primary neurons isolated from the hippocampus of a day 18 embryonic Sprague Daley rat stained with Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP), clone G-A-5 (G3893) at a 1:600 dilution (green), rabbit anti-neurofilament (N4142) at a 1:600 dilution (red) and bisBenzimide (blue).
Image created by Lynn Burgess
  Lynn Burgess, Natural Sciences Department, Dickinson State University, Dickinson, ND
Immunofluorescence
Equine exuberant granulation (proud flesh) tissue stained using Anti-von Willebrand Factor (F3520) at a 1:100 dilution and Anti-Rabbit IgG-FITC (F0382) (green) and DAPI (blue).
Image created by Katy Milne
  Katy Milne, Nelson Lab, Deeley Research Centre, BC Cancer Agency, Victoria, BC
Immunohistochemistry
Human ovarian tumor stained with Monoclonal Anti-pan Cytokeratin, clone PCK-26 (C1801) at a 1:300 dilution using DAB as a substrate and hematoxylin as a counterstain, 20x magnification.
Image created by Pam Wirth
  Pamela Wirth, Plastic Surgery Department, Vanderbilt University, Nashville, TN
Immunofluorescence
Human Tonsil stained with Anti-CD3, T Cell (C7930) at a 1:150 dilution (red) and Monoclonal Anti-Proliferating Cell Nuclear Antigen (P8825) at a 1:500 dilution (green). Counterstained with DAPI.
back to Antibody Explorer