|
CONGRATULATIONS to our Antibody Photo Contest Winners, November 2008!
 |
|
M. K. Kandasamy, Genetics Department, University of Georgia, Athens, GA
Immunofluorescence Arabidoposis suspension cells with the Monoclonal Anti-Actin, clone 10-B3 (MabGPa) (A0480) at a 1:100 dilution. The actin filaments were revealed with Anti-Mouse IgG-FITC (F6257) (green) and the nucleus is stained with DAPI (red). |
|
|
|
Shyam Khatau, Department of Chemical and Biomolecular Engineering, Johns Hopkins University, Baltimore, MD
Immunofluorescence Imna(-/-) mouse embryonic fibroblasts stained with Monoclonal Anti-Vimentin, clone LN-6 (V2258) at a 1:40 dilution (red), anti-actin (blue) and DAPI (yellow). |
|
|
|
Christopher M. Hale, Department of Chemical & Biomolecular Engineering Johns Hopkins University, Baltimore, MD
Immunofluorescence Mouse adult fibroblast stained with Monoclonal Anti-Vinculin, clone VIN-11-5 (V4505) at 1:40 dilution (aqua). Actin (purple) and nuclear DNA (blue) were stained using phalloidin-Alexa Fluor 568 and DAPI, respectively. |
|
 |
|
Raleigh Kladney, Surgery Department, Washington University School of Medicine, St. Louis, MO
Immunofluorescence Mouse anterior prostate stained using Monoclonal Anti-phospho-Histone H3 (pSer28), clone HTA28 (H9908) (red) at a 1:2,000 dilution, anti-phospho-S6 (pSer235/236) (green) and DAPI (blue). The figure shows a mitotic prostate epithelial cell with nuclear pH3 staining as well as cytoplasmic pS6 staining. |
|
|
|
Kenneth Reese, BrainBits, Springfield, IL
Immunofluorescence Primary neurons isolated from the hippocampus of a day 18 embryonic Sprague Daley rat stained with Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP), clone G-A-5 (G3893) at a 1:600 dilution (green), rabbit anti-neurofilament (N4142) at a 1:600 dilution (red) and bisBenzimide (blue). |
|
|
|
Lynn Burgess, Natural Sciences Department, Dickinson State University, Dickinson, ND
Immunofluorescence Equine exuberant granulation (proud flesh) tissue stained using Anti-von Willebrand Factor (F3520) at a 1:100 dilution and Anti-Rabbit IgG-FITC (F0382) (green) and DAPI (blue). |
|
|
|
Katy Milne, Nelson Lab, Deeley Research Centre, BC Cancer Agency, Victoria, BC
Immunohistochemistry Human ovarian tumor stained with Monoclonal Anti-pan Cytokeratin, clone PCK-26 (C1801) at a 1:300 dilution using DAB as a substrate and hematoxylin as a counterstain, 20x magnification. |
|
|
|
Pamela Wirth, Plastic Surgery Department, Vanderbilt University, Nashville, TN
Immunofluorescence Human Tonsil stained with Anti-CD3, T Cell (C7930) at a 1:150 dilution (red) and Monoclonal Anti-Proliferating Cell Nuclear Antigen (P8825) at a 1:500 dilution (green). Counterstained with DAPI. |
back to Antibody Explorer
|