|
Animal Component-Free Classical Media - Strict raw material control and stringent raw material specifications allow Sigma to offer these traditional classical media formulations that are animal component-free (ACF).
| Product # |
Product Name |
Add to Cart |
| B1522 |
Basal Medium Eagle liquid, cell culture tested, With Earle's salts and NaHCO3, without L-glutamine. |
|
| D5546 |
Dulbecco’s Modified Eagle’s Medium - low glucose With 1000 mg/L glucose, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, cell culture tested |
|
| D5671 |
Dulbecco’s Modified Eagle’s Medium - high glucose With 4500 mg/L glucose and sodium bicarbonate, without L-glutamine and sodium pyruvate, liquid, sterile-filtered, cell culture tested, hybridoma tested |
|
| D5796 |
Dulbecco’s Modified Eagle’s Medium - high glucose With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate., liquid, sterile-filtered, cell culture tested |
|
| D5921 |
Dulbecco’s Modified Eagle’s Medium - low glucose With 1000 mg/L glucose, and sodium bicarbonate, without L-glutamine, sodium pyruvate, and phenol red, liquid, sterile-filtered, cell culture tested |
|
| D6046 |
Dulbecco’s Modified Eagle’s Medium - low glucose With 1000 mg/L glucose, L-glutamine, and sodium bicarbonate, liquid, sterile-filtered, cell culture tested |
|
| D6171 |
Dulbecco’s Modified Eagle’s Medium - high glucose HEPES modification, With 4500 mg/L glucose, 25 mM HEPES, and sodium bicarbonate, without L-glutamine and sodium pyruvate, liquid, sterile-filtered, cell culture tested |
|
| D6421 |
Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham With 15 mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, cell culture tested |
|
| D6429 |
Dulbecco’s Modified Eagle’s Medium - high glucose With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, cell culture tested |
|
| D6434 |
Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham With 15 mM HEPES and sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, cell culture tested |
|
| D6546 |
Dulbecco’s Modified Eagle’s Medium - high glucose With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, cell culture tested |
|
| D8062 |
Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham With L-glutamine and sodium bicarbonate, without HEPES, liquid, sterile-filtered, cell culture tested |
|
| G5154 |
Glasgow Minimum Essential Medium With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, cell culture tested |
|
| I3390 |
Iscove’s Modified Dulbecco’s Medium With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, cell culture tested, hybridoma tested |
|
| M2279 |
Minimum Essential Medium Eagle With Earle's salts and sodium bicarbonate, without L-glutamine., liquid, sterile-filtered, cell culture tested |
|
| M2414 |
Minimum Essential Medium Eagle liquid, Modified, with Earle's salts and reduced NaHCO3. without L-glutamine., cell culture tested |
|
| M4526 |
Minimum Essential Medium Eagle Alpha Modification, with sodium bicarbonate, without L-glutamine, ribonucleosides and deoxyribonucleosides, liquid, sterile-filtered, cell culture tested |
|
| M4655 |
Minimum Essential Medium Eagle With Earle's salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, cell culture tested |
|
| M5650 |
Minimum Essential Medium Eagle With Earle's salts, non-essential amino acids and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, cell culture tested |
|
| M5775 |
Minimum Essential Medium Eagle With Hanks' salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, cell culture tested |
|
| M7278 |
Minimum Essential Medium Eagle HEPES Modification, with Earle's salts, 25mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, cell culture tested |
|
| M8167 |
Minimum Essential Medium Eagle Spinner Modification, with Earle's salts and sodium bicarbonate, without calcium chloride and L-glutamine, liquid, sterile-filtered, cell culture tested |
|
| N2147 |
Nutrient Mixture F-10 Ham With 20 mM HEPES, without sodium bicarbonate and L-glutamine, liquid, sterile-filtered, cell culture tested |
|
| N4888 |
Nutrient Mixture F-12 Ham With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, cell culture tested |
|
| N6013 |
Nutrient Mixture F-10 Ham With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, cell culture tested |
|
| N8641 |
Nutrient Mixture F-12 Ham HEPES Modification, with 25 mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, cell culture tested |
|
| R0883 |
RPMI-1640 Medium With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, cell culture tested |
|
| R5886 |
RPMI-1640 Medium HEPES Modification, With 25mM HEPES, without L-glutamine., liquid, sterile-filtered, cell culture tested |
|
| R7509 |
RPMI-1640 Medium Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, cell culture tested |
|
| R7513 |
RPMI-1640 Medium Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, cell culture tested |
|
| R7638 |
RPMI-1640 Medium Dutch Modification, with sodium bicarbonate and 20mM HEPES, without L-glutamine, sterile-filtered, cell culture tested |
|
| R8758 |
RPMI-1640 Medium With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, cell culture tested |
|
| W1878 |
Williams’ Medium E With sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, cell culture tested |
|
| W4128 |
Williams’ Medium E With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, cell culture tested |
|
| Back to top |
|