|
Classic Media and Salts
Basal Salt Mixtures
|
|
|
The use of balanced salt solution (BSS) in cell and tissue culture is generally attributed to early workers in the field. In 1885, Sydney Ringer developed a solution of inorganic salts designed to maintain contractility of mammalian heart tissue. A less specific salt solution was designed by Tyrode for use in work with primary mammalian cells. Tyrode's salt solution became the accepted fluid for diluting protein components of media of natural origin. Since that time, many other balanced salt solutions have been developed for use in cell and tissue culture. The current role of balanced salt solution in cell culture is multi-faceted and can be divided into four principle functions.
back to top
| Dulbecco's Phosphate Buffered Saline (D-PBS) |
| Product # |
Product Name |
Description |
Application |
| D 8662 |
Dulbecco's Phosphate Buffered Saline |
- Liquid
- With calcium chloride and magnesium chloride
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Known as D-PBS, this is a general use, isotonic saline solution for washing cells and tissues.
|
| D 1283 |
Dulbecco's Phosphate Buffered Saline (10x) |
- Liquid
- With calcium chloride and magnesium chloride
- pH 4.2-4.6 at 10x
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Concentrated form of D 8662.
|
| D 8537 |
Dulbecco's Phosphate Buffered Saline Modified |
- Liquid
- Without calcium chloride and magnesium chloride
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Calcium and magnesium promote cell adhesion. Use this formulation of D-PBS when tissue or cell dissociation or release is the objective.
|
| D 1408 |
Dulbecco's Phosphate Buffered Saline (10x) |
- Liquid
- Sterile-filtered
- pH 6.6-7.0 at 10x
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Concentrated form of D 8537.
|
| D 5652 |
Dulbecco's Phosphate Buffered Saline Modified |
- Powder
- Without calcium chloride and magnesium chloride
- Formulated at 9.6 grams of powder per liter of medium
- Related products: Dulbecco's Phosphate Buffered Saline Hybri-Max (D 7030)
- Cell culture tested
|
Powder form of D 8537.
Calcium and magnesium promote cell adhesion.
Use this formulation of D-PBS when tissue or cell dissociation or release is the objective.
|
| D 7030 |
Dulbecco's Phosphate Buffered Saline Modified Hybri-Max. |
- Powder
- Without calcium chloride and magnesium chloride
- Formulated at 9.6 grams of powder per liter of medium
- Endotoxin tested
- Hybridoma tested
|
Same as D 5652 except it is tested for use with hybridoma cells.
|
| D 5773 |
Dulbecco's Phosphate Buffered Saline |
- Powder
- Without calcium chloride
- Formulated at 9.7 grams of powder per liter of medium
- Cell culture tested
|
Calcium promotes cell adhesion.
Use this formulation of D-PBS when tissue or cell dissociation or release is the objective.
Also, suitable for studies that involve calcium metabolism.
|
| D 4031 |
Dulbecco's Phosphate Buffered Saline Modified |
- Liquid
- With 36 mg/L sodium pyruvate, 50 mg/L streptomycin sulfate, 100 mg/L kanamycin monosulfate, 1000 mg/L glucose, calcium chloride, and magnesium chloride.
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Fully supplemented isotonic solution.
Contains energy sources and antibiotics.
Frequently used with primary cells where risk of contamination may be high.
|
| D 6650 |
Dulbecco's Phosphate Buffered Saline Modified |
- Powder
- With 36 mg/L sodium pyruvate, 50 mg/L streptomycin sulfate and 1.000 mg/L glucose.
- w/o calcium chloride and penicillin G
- Penicillin G supplied separately with each package for addition at a final concentration of 1 million units/L. Calcium chloride di-hydrate is supplied separately with each package for final concentration of 133 mg/L.
- Cell culture tested
|
Isotonic saline solution that contains energy sources and antibiotics.
Provides the option to formulate with or without penicillin G or calcium.
Frequently used with primary cells where risk of contamination may be high and where tissue dissociation is an objective.
|
| |
back to top
|
| Earle's Balanced Salts (EBSS) |
|
| Product # |
Product |
Description |
Application |
| E 2888 |
Earle's Balanced Salt Solution |
- Liquid
- With sodium bicarbonate
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Complete and ready to use formulation of EBSS.
|
| E 7510 |
Earle's Balanced Salt Solution (10x) |
- Liquid
- Without sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate at 1x
- pH 4.0-4.4 at 10x
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Concentrated version of E 2888. Must be diluted to 1x, supplemented with sodium bicarbonate and pH adjusted before use.
|
| E 6132 |
Earle's Balanced Salts |
- Powder
- Without sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate
- Formulated at 8.7 grams of powder per liter of medium
- Cell culture tested
|
Powder version of the complete EBSS formulation (E 2888).
Sodium bicarbonate is added after the EBSS is dissolved in tissue culture grade water. The pH is adjusted after addition of sodium bicarbonate.
|
| E 3024 |
Earle's Balanced Salt Solution |
- Liquid
- With sodium bicarbonate
- Without phenol red
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same as E 2888, except without phenol red.
Phenol red has been shown to interfere with the growth of some cells at cloning densities.
Use this medium when working with stem cells or when growing cells at low densities.
|
| E 3261 |
Earle's Balanced Salt Solution Modified |
- Powder
- Without phenol red and sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate
- Formulated at 8.7 grams of powder per liter of medium
- Cell culture tested
|
Powder version of E 3024.
Sodium bicarbonate is added after the EBSS is dissolved in tissue culture grade water. The pH is adjusted after addition of sodium bicarbonate.
Phenol red has been shown to interfere with the growth of some cells at cloning densities.
Use this medium when working with stem cells or when growing cells at low densities.
|
| E 6267 |
Earle's Balanced Salt Solution |
- Liquid
- With sodium bicarbonate
- Without calcium chloride, magnesium sulfate
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same as E 2888, except that calcium and magnesium have been removed.
Calcium and magnesium support cell adhesion.
Recommended for suspension cultures or whenever cell clumping is a problem.
|
| E 2899 |
Earle's Balanced Salt Solution |
- Liquid
- With sodium bicarbonate
- Without calcium chloride, magnesium sulfate, and phenol red
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same as E 6267, except that phenol red has also been removed.
Phenol red has been shown to interfere with the growth of some cells at cloning densities.
Use this medium when working with stem cells or when growing cells at low densities.
|
|
back to top
|
| Hanks' Balanced Salts (HBSS) |
|
| Product # |
Product |
Description |
Application |
| H 9269 |
Hanks' Balanced Salt Solution |
- Liquid
- With sodium bicarbonate
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Complete and ready to use formulation of HBSS.
|
| H 1641 |
Hanks' Balanced Salt Solution (10x) |
- Liquid
- Without sodium bicarbonate; supplement with 0.35 gm/L sodium bicarbonate at 1x
- pH 5.3-6.3 at 10x
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Concentrated version of H 9269. Must be diluted to 1x, supplemented with sodium bicarbonate and pH adjusted before use.
|
| H 6136 |
Hanks' Balanced Salts |
- Powder
- Without sodium bicarbonate; supplement with 0.35 gm/L sodium bicarbonate
- Formulated at 9.8 grams of powder per liter of medium
- Cell culture tested
|
Powder version of the complete HBSS formulation (H 9269).
Sodium bicarbonate is added after the HBSS is dissolved in tissue culture grade water. The pH is adjusted after addition of sodium bicarbonate.
Same formulation as Hanks' Balanced Salts, Hybri-Max. (H 6393).
|
| H 6393 |
Hanks' Balanced Salts, Hybri-Max. |
- Powder
- Without sodium bicarbonate; supplement with 0.35 gm/L sodium bicarbonate
- Formulated at 9.8 grams of powder per liter of medium
- Endotoxin tested
- Hybridoma tested
|
Related to H 6136. Specifically, tested for use with hybridoma cells.
Labeled for in vitro diagnostic use.
|
| H 8264 |
Hanks' Balanced Salt Solution Modified |
- Liquid
- With sodium bicarbonate
- Without phenol red
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same as H 9269, except without phenol red.
Phenol red has been shown to interfere with the growth of some cells at cloning densities.
Use this medium when working with stem cells or when growing cells at low densities.
|
| H 1387 |
Hanks' Balanced Salts Modified |
- Powder
- Without phenol red and sodium bicarbonate; supplement with 0.35 gm/L sodium bicarbonate
- Formulated at 9.8 grams of powder per liter of medium
- Cell culture tested
|
Powder version of H 8264.
Sodium bicarbonate is added after the HBSS is dissolved in tissue culture grade water. The pH is adjusted after addition of sodium bicarbonate.
Phenol red has been shown to interfere with the growth of some cells at cloning densities.
Use this medium when working with stem cells or when growing cells at low densities.
|
| H 9394 |
Hanks' Balanced Salt Solution Modified |
- Liquid
- With sodium bicarbonate
- Without calcium chloride, magnesium sulfate,
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same as H 6649, except that calcium and magnesium have been removed.
Calcium and magnesium support cell adhesion.
Recommended for suspension cultures or whenever cell clumping is a problem.
|
| H 4641 |
Hanks' Balanced Salt Solution (10x) |
- Liquid
- Without calcium chloride, magnesium sulfate, and sodium bicarbonate; supplement with 0.35 gm/L sodium bicarbonate
- pH 5.4-6.4 at 10x
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Concentrated version of H 9394. Must be diluted to 1x, supplemented with sodium bicarbonate and pH adjusted before use.
Calcium and magnesium support cell adhesion.
Recommended for suspension cultures or whenever cell clumping is a problem.
|
| H 2387 |
Hanks' Balanced Salts Modified |
- Powder
- Without calcium chloride, magnesium sulfate, and sodium bicarbonate; supplement with 0.35 gm/L sodium bicarbonate
- Formulated at 9.5 grams of powder per liter of medium
- Cell culture tested
|
Powder version of H 9394.
Sodium bicarbonate is added after the HBSS is dissolved in tissue culture grade water. The pH is adjusted after addition of sodium bicarbonate.
Calcium and magnesium support cell adhesion.
Recommended for suspension cultures or whenever cell clumping is a problem.
|
| H 6648 |
Hanks' Balanced Salt Solution Modified |
- Liquid
- With sodium bicarbonate
- Without calcium chloride, magnesium sulfate, and phenol red
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same as H 9394, except that phenol red has been removed.
Calcium and magnesium support cell adhesion. Phenol red has been shown to interfere with the growth of some cells at cloning densities.
Use this medium when working with stem cells or when growing cells at low densities. Use to minimize cell attachment or aggregation.
|
| H 4891 |
Hanks' Balanced Salts Modified |
- Powder
- Without calcium chloride, magnesium sulfate, phenol red and sodium bicarbonate; supplement with 0.35 gm/L sodium bicarbonate
- Formulated at 9.5 grams of powder per liter of medium
- Cell culture tested
|
Powder version of H 6648.
Sodium bicarbonate is added after the HBSS is dissolved in tissue culture grade water. The pH is adjusted after addition of sodium bicarbonate.
Calcium and magnesium support cell adhesion.
Phenol red has been shown to interfere with the growth of some cells at cloning densities.
Use this medium when working with stem cells or when growing cells at low densities. Use to minimize cell attachment or aggregation.
|
|
back to top
|
|
|
| Product # |
Product |
Description |
Application |
| T 2397 |
Tyrode's Salt Solution |
- Liquid
- With sodium bicarbonate
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Complete and ready to use formulation of Tyrode's salt solution.
For cell washing.
|
| T 2145 |
Tyrode's Salts |
- Powder
- Without sodium bicarbonate; supplement with 1.0 gm/L of sodium bicarbonate
- Formulated at 9.7 grams of powder per liter of medium
- Endotoxin tested
- Sterile-filtered
- Cell culture tested
|
Powder version of the complete Tyrode's salts formulation (T 2397).
Sodium bicarbonate is added after the salts are dissolved in tissue culture grade water. The pH is adjusted after addition of sodium bicarbonate.
For cell washing.
|
| T 1788 |
Tyrode's Solution, Acidic |
- Liquid
- Contains 4.0 gm/L polyvinylpyrrolidone (average molecular weight 40,000)
- pH: 2.5 +/- 0.3
- Shipped in dry ice
- Sterile-filtered
- Endotoxin tested
- Embryo tested
|
Special formulation of Tyrode's salts (see formulation).
Recommended for removal of the zona pellucida from mouse embryos.
|
|
back to top
|
| Other Salt Mixtures
The following salts solutions are available for use in specific cell culture or related applications. Generally, the use of one of these formulations for an application is specified in publications or technical protocols.
|
|
| Product # |
Product |
Description |
Application |
| A 3551 |
Alsever's Solution |
- Liquid
- Sterile-filtered
- Endotoxin Tested
- Cell culture tested
|
Essentially a solution containing citrate, sodium chloride and glucose. Provided without bicarbonate, which may be added.
Hypotonic solution for lysing cells. Especially, red blood cells.
|
| G 9779 |
Gey's Balanced Salt Solution |
- Liquid
- Sterile-filtered
- Endotoxin Tested
- Cell culture tested
|
A complete balanced salt solution with a unique combination of "salt forms". Contains the same five basic ions as EBSS, HBSS and D-PBS.
|
| K 3753 |
Kreb's-Henseleit Buffer Modified |
- Powder
- With 2000 mg/L glucose
- Without calcium chloride and sodium bicarbonate; supplement with 2.1 gm/L of sodium bicarbonate;
- Formulated at 9.6 grams of powder per liter of medium
- Cell culture tested
|
Contains the same components as K 4002. However, these ion components are at different concentrations and in different salt forms. (see the formulation for details).
|
| K 4002 |
Kreb's-Ringer Bicarbonate Buffer |
- Powder
- With 1800 mg/L glucose
- Without calcium chloride and sodium bicarbonate; supplement with 1.26 gm/L sodium bicarbonate
- Formulated at 9.5 grams of powder per liter of medium
- Cell culture tested
|
Bicarbonate buffered isotonic saline containing magnesium, potassium, sodium and phosphate ions.
|
| P 2917 |
Puck's Saline |
- Liquid
- With sodium bicarbonate
- Without calcium chloride, magnesium sulfate, potassium phosphate and sodium phosphate
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
A very simple isotonic salt solution. Contains sodium and potassium chloride, glucose and low concentration of bicarbonate buffer (0.35 gm/L)
For in vitro diagnostic use
|
back to top
-
as an irrigating, transporting and diluting fluid while maintaining intra- and extra-cellular osmotic balance;
-
provides cells with water and certain bulk inorganic ions essential for normal cell metabolism;
-
combined with a carbohydrate, such as glucose, provides the principle energy source for cell metabolism; and
-
provides buffering system to maintain the medium within the physiological pH range (7.2-7.6)
Families of basal salt solutions, generally named after the inventor, have been developed around the early discovery that cells have a basic requirement for five ions: calcium, magnesium, potassium, sodium, and phosphate. Each balanced salts family of products differs in the concentration and "salt form" of these ions.
Please note that products listed as "Other Salt Solutions" may have uses other than the culturing of cells and that they may not contain all ions listed above.
Probably the three most important basal salt solutions for cell culture are Dulbecco's phosphate buffered saline (D-PBS); Earle's balanced salts (EBSS) and Hanks' balanced salts (HBSS). Dulbecco's formulation contains no bicarbonate and hence is less susceptible to pH shift during filtration. Earle's formulation contains 2.2 gm/L of sodium bicarbonate, and Hanks' formulation contains 0.35 gm/L of sodium bicarbonate. Both of these formulations may require pH adjustment after filtration.
D-PBS is offered with and without glucose as an energy source. The complete formulae for EBSS and HBSS both contain 1.0 mg/L of glucose.