| Product # |
Product Name |
Description |
Application |
| D 8662 |
Dulbecco's Phosphate Buffered Saline |
- Liquid
- With calcium chloride and magnesium chloride
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Known as D-PBS, this is a general use, isotonic saline solution for washing cells and tissues.
|
| D 1283 |
Dulbecco's Phosphate Buffered Saline (10x) |
- Liquid
- With calcium chloride and magnesium chloride
- pH 4.2-4.6 at 10x
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Concentrated form of D 8662.
|
| D 8537 |
Dulbecco's Phosphate Buffered Saline Modified |
- Liquid
- Without calcium chloride and magnesium chloride
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Calcium and magnesium promote cell adhesion. Use this formulation of D-PBS when tissue or cell dissociation or release is the objective.
|
| D 1408 |
Dulbecco's Phosphate Buffered Saline (10x) |
- Liquid
- Sterile-filtered
- pH 6.6-7.0 at 10x
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Concentrated form of D 8537.
|
| D 5652 |
Dulbecco's Phosphate Buffered Saline Modified |
- Powder
- Without calcium chloride and magnesium chloride
- Formulated at 9.6 grams of powder per liter of medium
- Related products: Dulbecco's Phosphate Buffered Saline Hybri-Max (D 7030)
- Cell culture tested
|
Powder form of D 8537.
Calcium and magnesium promote cell adhesion.
Use this formulation of D-PBS when tissue or cell dissociation or release is the objective.
|
| D 7030 |
Dulbecco's Phosphate Buffered Saline Modified Hybri-Max. |
- Powder
- Without calcium chloride and magnesium chloride
- Formulated at 9.6 grams of powder per liter of medium
- Endotoxin tested
- Hybridoma tested
|
Same as D 5652 except it is tested for use with hybridoma cells.
|
| D 5773 |
Dulbecco's Phosphate Buffered Saline |
- Powder
- Without calcium chloride
- Formulated at 9.7 grams of powder per liter of medium
- Cell culture tested
|
Calcium promotes cell adhesion.
Use this formulation of D-PBS when tissue or cell dissociation or release is the objective.
Also, suitable for studies that involve calcium metabolism.
|
| D 4031 |
Dulbecco's Phosphate Buffered Saline Modified |
- Liquid
- With 36 mg/L sodium pyruvate, 50 mg/L streptomycin sulfate, 100 mg/L kanamycin monosulfate, 1000 mg/L glucose, calcium chloride, and magnesium chloride.
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Fully supplemented isotonic solution.
Contains energy sources and antibiotics.
Frequently used with primary cells where risk of contamination may be high.
|
| D 6650 |
Dulbecco's Phosphate Buffered Saline Modified |
- Powder
- With 36 mg/L sodium pyruvate, 50 mg/L streptomycin sulfate and 1.000 mg/L glucose.
- w/o calcium chloride and penicillin G
- Penicillin G supplied separately with each package for addition at a final concentration of 1 million units/L. Calcium chloride di-hydrate is supplied separately with each package for final concentration of 133 mg/L.
- Cell culture tested
|
Isotonic saline solution that contains energy sources and antibiotics.
Provides the option to formulate with or without penicillin G or calcium.
Frequently used with primary cells where risk of contamination may be high and where tissue dissociation is an objective.
|
| |
back to top
|
| Earle's Balanced Salts (EBSS) |
|
| Product # |
Product |
Description |
Application |
| E 2888 |
Earle's Balanced Salt Solution |
- Liquid
- With sodium bicarbonate
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Complete and ready to use formulation of EBSS.
|
| E 7510 |
Earle's Balanced Salt Solution (10x) |
- Liquid
- Without sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate at 1x
- pH 4.0-4.4 at 10x
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Concentrated version of E 2888. Must be diluted to 1x, supplemented with sodium bicarbonate and pH adjusted before use.
|
| E 6132 |
Earle's Balanced Salts |
- Powder
- Without sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate
- Formulated at 8.7 grams of powder per liter of medium
- Cell culture tested
|
Powder version of the complete EBSS formulation (E 2888).
Sodium bicarbonate is added after the EBSS is dissolved in tissue culture grade water. The pH is adjusted after addition of sodium bicarbonate.
|
| E 3024 |
Earle's Balanced Salt Solution |
- Liquid
- With sodium bicarbonate
- Without phenol red
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same as E 2888, except without phenol red.
Phenol red has been shown to interfere with the growth of some cells at cloning densities.
Use this medium when working with stem cells or when growing cells at low densities.
|
| E 3261 |
Earle's Balanced Salt Solution Modified |
- Powder
- Without phenol red and sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate
- Formulated at 8.7 grams of powder per liter of medium
- Cell culture tested
|
Powder version of E 3024.
Sodium bicarbonate is added after the EBSS is dissolved in tissue culture grade water. The pH is adjusted after addition of sodium bicarbonate.
Phenol red has been shown to interfere with the growth of some cells at cloning densities.
Use this medium when working with stem cells or when growing cells at low densities.
|
| E 6267 |
Earle's Balanced Salt Solution |
- Liquid
- With sodium bicarbonate
- Without calcium chloride, magnesium sulfate
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same as E 2888, except that calcium and magnesium have been removed.
Calcium and magnesium support cell adhesion.
Recommended for suspension cultures or whenever cell clumping is a problem.
|
| E 2899 |
Earle's Balanced Salt Solution |
- Liquid
- With sodium bicarbonate
- Without calcium chloride, magnesium sulfate, and phenol red
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same as E 6267, except that phenol red has also been removed.
Phenol red has been shown to interfere with the growth of some cells at cloning densities.
Use this medium when working with stem cells or when growing cells at low densities.
|