Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Culture > Classical Media & Salts > Minimum Essential Medium Eagle
Cell Culture
Minimum Essential Medium Eagle
Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.  Early attempts to cultivate normal mammalian fibroblasts and certain subtypes of HeLa cells revealed that they had specific nutritional requirements that could not be met by Eagle's Basal Medium (BME).  Subsequent studies using these and other cells in culture indicated that additions to BME could be made to aid growth of a wider variety of fastidious cells.  MEM, which incorporates these modifications, includes higher concentrations of amino acids so that the medium more closely approximates the protein composition of mammalian cells. MEM has been used for cultivation of a wide variety of cells grown in mono-layers.  Optional supplementation of non-essential amino acids to the formulations that incorporate either Hanks' or Eagles' salts has broadened the usefulness of this medium.  The formulation has been further modified by optional elimination of calcium to permit the growth of cells in suspension.
The following table contains a large selection of MEM formulations.  Selection of the most appropriate formulation for your application requires an understanding of the way these formulations are constructed and offered.
For more information on medium selections click here 
Product # Product Name Description Application
M 2279 Minimum Essential Medium Eagle
  • Liquid
  • With Earle's salts and sodium bicarbonate
  • Without L-glutamine, supplement with 0.292 gm/L of L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Essentially the same as the original formula.
Generally used to grow attached cell lines, such as fibroblasts, in the presence of FBS, calf or horse sera.
M 4655 Minimum Essential Medium Eagle
  • Liquid
  • With Earle's salts, L-glutamine and sodium bicarbonate
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Essentially the same as the original formula.
Generally used to grow attached cell lines, such as fibroblasts, in the presence of FBS, calf or horse sera.
M 0275 Minimum Essential Medium Eagle (10x)
  • Liquid
  • With Earle's salts
  • Without L-glutamine and sodium bicarbonate
  • Supplement with 0.292 gm/L of L-glutamine and 2.2 gm/L of sodium bicarbonate at 1x
  • pH 5.1-5.5 at 10x
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Essentially the same as the original formula.
Generally used to grow attached cell lines, such as fibroblasts, in the presence of FBS, calf or horse sera.
The L-glutamine and sodium bicarbonate are added to the medium after dilution to 1x and pH adjustment.
M 0268 Minimum Essential Medium Eagle
  • Powder
  • With Earle's salts and L-glutamine
  • Without sodium bicarbonate, supplement with 2.2 gm/L sodium bicarbonate
  • Formulated at 9.6 grams of powder per liter of medium
  • Cell culture tested
Essentially the same as the original formula, except it is provided as a powder.
Generally used to grow attached cell lines, such as fibroblasts, in the presence of FBS, calf or horse sera.
M 5650 Minimum Essential Medium Eagle
  • Liquid
  • With Earle's salts, non-essential amino acids and sodium bicarbonate
  • Without L-glutamine, supplement with 0.292 gm/L of L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Same as M 2279, with one major difference.  It contains 19 amino acids.  The essential amino acids and the non-essential amino acids: L-ala; L-asn; L-asp; L-glu; L-gly; L-pro and L-ser.
May be preferred for growth of adherent cells in the presence of reduced serum or as a base for development of a serum free medium.
M 0643 Minimum Essential Medium Eagle
  • Powder
  • With Earle's salts, L-glutamine and non-essential amino acids
  • Without sodium bicarbonate, supplement with 2.2 gm/L sodium bicarbonate
  • Formulated at 9.7 grams of powder per liter of medium
  • Cell culture tested
Offered as powder form of M 5650
It contains 19 amino acids.  The essential amino acids and the non-essential amino acids: L-ala; L-asn; L-asp; L-glu; L-gly; L-pro and L-ser.
May be preferred for growth of adherent cells in the presence of reduced serum or as a base for development of a serum free medium.
M 3024 Minimum Essential Medium Eagle Modified
  • Powder
  • With Earle's salts and non-essential amino acids
  • Without L-glutamine, phenol red, and sodium bicarbonate.
  • Supplement with 0.292 gm/L L-glutamine; with 2.2 gm/L of sodium bicarbonate
  • Formulated at 9.4 grams of powder per liter of medium
  • Cell culture tested
It contains 19 amino acids.  The essential amino acids and the non-essential amino acids: L-ala; L-asn; L-asp; L-glu; L-gly; L-pro and L-ser.
May be preferred for growth of adherent cells in the presence of reduced serum or as a base for development of a serum free medium.
Same as M 0643, except that L-glutamine may be added at time of use.
M 4144 Minimum Essential Medium Eagle Modified
  • Powder
  • With Earle's salts
  • Without phenol red, L-glutamine and sodium bicarbonate.
  • Supplement with 0.292 gm/L L-glutamine; with 2.2 gm/L of sodium bicarbonate
  • Formulated at 9.3 grams of powder per liter of medium
  • Cell culture tested
Essentially the same as the original formula, except it is provided as a powder and without phenol red.
Generally used to grow attached cell lines, such as fibroblasts, in the presence of FBS, calf or horse sera. 
Media without phenol red is often preferred for work with stem cells and in cell cloning.
M 7278 Minimum Essential Medium Eagle HEPES Modification
  • Liquid
  • With Earle's salts, 25 mM HEPES and sodium bicarbonate
  • Without L-glutamine; supplement with 0.292 gm/L of L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Same as M 2279, except that it is supplemented with 25 mM HEPES.
HEPES increases the buffer strength of the medium and raises the maximum buffering range.
Recommended for cells that grow better at a pH above 7.2.
M 2645 Minimum Essential Medium Eagle HEPES Modification
  • Powder
  • With Earle's salts, L-glutamine and HEPES
  • Without sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate
  • Formulated at 14.2 grams of powder per liter of medium
  • Cell culture tested
Similar to M 7278, except that it is a powder without sodium bicarbonate.
HEPES increases the buffer strength of the medium and raises the maximum buffer range.  Leaving the bicarbonate out gives the investigator greater control over the buffer system.
Recommended for cells that grow better at a pH above 7.2.
M 8042 Minimum Essential Medium Eagle Alpha Modification
  • Liquid
  • With ribonucleosides and deoxyribonucleosides and sodium bicarbonate
  • Without L-glutamine, supplement with 0.292 gm/L of L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
This is the most enriched variation of the MEM formulation offered.  It contains all 21 normal amino acids, some at increased concentrations.  In addition, it contains 5 additional vitamins.  See formulation for details.
Often used in cytogenetic analysis when supplemented with FBS.
M 4526 Minimum Essential Medium Eagle Alpha Modification
  • Liquid
  • With sodium bicarbonate 
  • Without L-glutamine, supplement with 0.292 gm/L of L-glutamine
  • Without ribonucleosides and deoxyribonucleosides
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Same as M 8042, except that it lacks the ribonucleosides and deoxyribonucleosides.
This is the most enriched variation of the MEM formulation offered.  It contains all 21 normal amino acids, some at increased concentrations. In addition, it contains 5 additional vitamins.
M 0644 Minimum Essential Medium Eagle Alpha Modification
  • Powder
  • With L-glutamine, ribonucleosides and deoxyribonucleosides.
  • Without sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate
  • Formulated at 10.2 grams of powder per liter of medium
  • Cell culture tested
This is the most enriched variation of the MEM formulation offered.  It contains all 21 normal amino acids, some at increased concentrations. In addition, it contains 5 additional vitamins.
Often used in cytogenetic analysis when supplemented with FBS.
Offered as a powder form of M 8042.
M 0894 Minimum Essential Medium Eagle Alpha Modification
  • Powder
  • With L-glutamine
  • Without ribonucleosides, deoxyribonucleosides and sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate
  • Formulated at 10.1 grams of powder per liter of medium
  • Cell culture tested
This is the most enriched variation of the MEM formulation offered.  It contains all 21 normal amino acids, some at increased concentrations. In addition, it contains 5 additional vitamins.
Offered as a powder form of M 4526.
M 2414 Minimum Essential Medium Eagle Modified
  • Liquid
  • With Earle's salts and reduced sodium bicarbonate
  • Without L-glutamine; supplement with 0.292 gm/L of L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Same as M 2279, except that the sodium bicarbonate concentration is reduced to 0.85 gm/L. 
Recommended for use in a carbon dioxide-free culture system.  May also be supplemented with HEPES to raise the buffering range.
M 3786 Minimum Essential Medium Eagle
  • Liquid
  • With Earle's salts, sodium bicarbonate and L-glutamine
  • Without sodium phosphate; supplement with 0.122 gm/L of sodium phosphate monobasic (anhydrous)
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Same as M 2279, except that sodium phosphate has been left out of the formulation.
Recommended for investigators who  want to use a formulation that is totally buffered by sodium bicarbonate.
M 8028 Minimum Essential Medium Eagle Joklik Modification
  • Liquid
  • With sodium bicarbonate 
  • Without calcium chloride and L-glutamine, supplement with 0.292 gm/L of L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Calcium and magnesium support cell adhesion.  Calcium has been removed from this formulation.  However, it still contains magnesium chloride.
Recommended for suspension cultures.
M 0518 Minimum Essential Medium Eagle Joklik Modification
  • Powder
  • With  L-glutamine
  • Without calcium chloride and sodium bicarbonate, supplement with 2.0 gm/L sodium bicarbonate
  • Formulated at 11.2 grams of powder per liter of medium
  • Cell culture tested
Calcium and magnesium support cell adhesion.  Calcium has been removed from this formulation.  However, it still contains magnesium chloride.
Recommended for suspension cultures.
Offered as the powder form of M 8028
M 8167 Minimum Essential Medium Eagle Spinner Modification (S-MEM)
  • Liquid
  • With Earle's salts and sodium bicarbonate
  • Without calcium chloride and L-glutamine; supplement with 0.292 gm/L of L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Calcium and magnesium support cell adhesion.  Calcium has been removed from this formulation.  However, it still contains magnesium sulfate.
Recommended for suspension cultures.
M 4767 Minimum Essential Medium Eagle Spinner Modification (S-MEM)
  • Powder
  • With L-glutamine
  • Without calcium chloride and sodium bicarbonate
  • Formulated at 10.4 grams of powder per liter of medium
  • Cell culture tested
Calcium and magnesium support cell adhesion.  Calcium has been removed from this formulation.  However, it still contains magnesium sulfate.
Recommended for suspension cultures
Offered as the powder form of M 8167
M 7395 Minimum Essential Medium Eagle D-Valine Modification
  • Powder
  • With Earle's salts, L-glutamine and D-valine substituted for L-valine.
  • Without sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate.
  • Formulated at 9.6 grams of powder per liter of medium
  • Cell culture tested
Same formulation as M 2279 except that D-valine has been substituted for L-valine.
Recommended for the preferential selection and growth of cells possessing D-amino acid oxidase, such as epithelial and Schwann cells.
M 7270 Minimum Essential Medium Eagle
  • Powder
  • With Earle's salts and L-glutamine.
  • Without L-leucine, L-lysine, L-methionine and sodium bicarbonate
  • Supplement with 0.052 gm/L of L-leucine; 0.0725 gm/L of L-lysine; 0.015 gm/L of  L-methionine and 2.2 gm/L of sodium bicarbonate.
  • Formulated at 9.4 grams of powder per liter of medium
  • Cell culture tested
Same as M 0268, except that 3 essential amino acids are not included in the powder.
This base formulation is particularly suitable for studies of cell protein metabolism and label-tagged amino acid studies.
Offered as the powder form of M 2289
M 3911 Minimum Essential Medium Eagle
  • Liquid
  • With Earle's salts, sodium bicarbonate and L-glutamine
  • Without L-methionine; supplement with 0.015 gm/L L-methionine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Same as M 4655, except that L-methionine is not included.
Useful for studies of methionine metabolism.
M 2289 Minimum Essential Medium Eagle Modified
  • Liquid
  • With Earle's salts and sodium bicarbonate
  • Without L-methionine, L-cystine, and L-glutamine
  • Supplement with 0.313 gm/L of L-cystine dihydrochloride; 0.292 gm/L of L-glutamine and 0.015 gm/L of  L-methionine.
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Same as M 2279, except that 3 essential amino acids are not included.
This base formulation is particularly suitable for studies of cell protein metabolism and label-tagged amino acid studies.
M 0769 Minimum Essential Medium Eagle Auto-ModTM
  • Powder
  • Modified for autoclaving
  • With Earle's salts
  • Without L-glutamine and sodium bicarbonate.
  • Supplement with 0.292 gm/L L-glutamine; with 2.2 gm/L of sodium bicarbonate
  • Formulated at 9.4 grams of powder per liter of medium
  • Cell culture tested
Modification of the original MEM formulation. Contains a succinate buffering system.  See formulations table for specific information.
Medium may be autoclaved. 
M 5775 Minimum Essential Medium Eagle
  • Liquid
  • With Hanks' salts and sodium bicarbonate
  • Without L-glutamine, supplement with 0.292 gm/L of L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Modification of the original MEM formulation.  Earle's salts have been replaced with Hanks' salts.
Since the bicarbonate concentration is substantially lower, this medium has a substantially reduced buffering capacity.
Same as M 4780, except that the L-glutamine may be added at time of use.
M 4780 Minimum Essential Medium Eagle
  • Liquid
  • With Hanks' salts, L-glutamine and sodium bicarbonate
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Modification of the original MEM formulation.  Earle's salts have been replaced with Hanks' salts.
Since the bicarbonate concentration is substantially lower, this medium has a substantially reduced buffering capacity.
M 9288 Minimum Essential Medium Eagle (10x)
  • Liquid
  • With Hanks' salts
  • Without L-glutamine and sodium bicarbonate
  • Supplement with 0.292 gm/L of L-glutamine and 0.35 gm/L of sodium bicarbonate at 1x
  • pH 5.2-5.4 at 10x
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
This is the 10x concentrated version of M 5775 and M 4780.
L-glutamine and sodium bicarbonate are added after the medium has been diluted to 1x and the pH has been adjusted.
M 4642 Minimum Essential Medium Eagle
  • Powder
  • With Hanks' salts and  L-glutamine.
  • Without sodium bicarbonate; supplement with 0.35 gm/L sodium bicarbonate
  • Formulated at 10.7 grams of powder per liter of medium
  • Cell culture tested
Modification of the original MEM formulation.  Earle's salts have been replaced with Hanks' salts.
Since the bicarbonate concentration is substantially lower, this medium has a substantially reduced buffering capacity.
M 3149 Minimum Essential Medium Eagle Modified
  • Powder
  • With Hanks' salts and non-essential amino acids
  • Without L-glutamine, phenol red and sodium bicarbonate
  • Supplement with 0.292 gm/L of L-glutamine
  • Formulated at 10.5 grams of powder per liter of medium
  • Cell culture tested
Modification of the original MEM formulation.  Earle's salts have been replaced with Hanks' salts.
Since the bicarbonate concentration is substantially lower, this medium has a substantially reduced buffering capacity.
It contains 19 amino acids.  The essential amino acids and the non-essential amino acids: L-ala; L-asn; L-asp; L-glu; L-gly; L-pro and L-ser.
May be preferred for growth of adherent cells in the presence of reduced serum or as a base for development of a serum free medium.
M 1018 Minimum Essential Medium Eagle
  • Powder
  • With Hanks' salts,  L-glutamine and non-essential amino acids
  • Without sodium bicarbonate; supplement with 0.35 gm/L sodium bicarbonate
  • Formulated at 10.8 grams of powder per liter of medium
  • Cell culture tested
Modification of the original MEM formulation.  Earle's salts have been replaced with Hanks' salts.
Since the bicarbonate concentration is substantially lower, this medium has a substantially reduced buffering capacity.
It contains 19 amino acids.  The essential amino acids and the non-essential amino acids: L-ala; L-asn; L-asp; L-glu; L-gly; L-pro and L-ser.
May be preferred for growth of adherent cells in the presence of reduced serum or as a base for development of a serum free medium.
Medium Selection Information
Most formulations are offered in both a powder and a liquid form.  In addition, most formulations are offered as a complete medium or in deficient variations that allow the user to supplement with common components such as bicarbonate, glutamine or calcium.  These variations provide more flexibility and control over the ultimate use and suitability of the medium.
Most of the formulations offered contain the original Earle's salts.  However, a number of formulations contain Hanks' salts.  Both of these phosphate and bicarbonate buffered saline solutions contain the same ions: calcium; magnesium; potassium; sodium,and phosphate.  They differ in the concentrations of calcium, sodium and in the buffer component.  Hanks' buffered salts solution contains a substantially lower concentration of sodium bicarbonate than does Earle's buffered salts.
The original MEM formulation contained Earle's salts and a group of amino acids generally referred to as essential amino acids. They were considered essential because the cells for which the medium was developed would not grow in their absence.  The 12 essential amino acids are: L-arginine; L-cystine; L-glutamine; L-histidine; L-isoleucine; L-leucine; L-methionine; L-phenylalanine; L-threonine; L-tryptophan; L-tyrosine; and L-valine.
back to top