Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Culture > Classical Media & Salts > Other Classic Media
Cell Culture
Other Classic Media
   Ames' Media
BGJb Medium (Fitton-Jackson Modification)
Click's Medium
CMRL-1066 Medium
Fischer's Medium
Glascow Minimum Essential Medium (GMEM)
Iscove's Modified Dulbecco's Medium (IMDM)
L-15 Medium (Leibovitz)
McCoy's 5A Modified Medium
NCTC Medium
Swim's S-77 Medium
Waymouth Medium
William's Medium E
Ames' Medium
The retina is an important in vitro model for the central nervous system. It is more readily accessible than most nervous tissue and is strong enough to remain intact during manipulations.  The retina of the rabbit commonly contains no penetrating blood vessels, but is nurtured by diffusion from capillary networks on either side.  For this reason, the tissue will survive and functions without circulating blood, as long as it is bathed in a medium that closely resembles in vivo conditions.
Ames' medium was formulated to support retinal tissue in relatively short-term culture.  Rabbit retina has been incubated in Ames' medium for over 2 days with its metabolism and electrical responses to light stimuli well maintained.  This mixture is a medium of choice for maintaining central nervous system tissue in vitro.
Product # Product Name Description Application
A 1420 Ames' Medium
  • Powder
  • With L-glutamine
  • Without sodium bicarbonate; supplement with 1.9 gm/L sodium bicarbonate
  • Formulated at 8.9 grams of powder per liter of medium;
  • Cell culture tested
Use to maintain retinal tissues for in vitro studies on central nervous system.
back to top
BGJb Medium (Fitton-Jackson Modification)
Medium BGJ was originally developed by Biggers, Gwatkin and Judah in the early 1960's at the Wistar Institute.  Subsequent studies resulted in a modification designated BGJb which has been used for supporting cultures of cartilaginous embryonic bone.  An additional modification, developed by Sylvia Fitton-Jackson at Strangeways Laboratory in England, is further enriched over the prior formulae.  Additional amino acids and vitamins, and increased buffering capacity conferred by the phosphates in the Fitton-Jackson modification, create conditions that permit calcification, as well as, growth of cartilaginous embryonic bone.
Product # Product Name Description Application
B 6644 BGJb Medium (Fitton-Jackson Modification)
  • Powder
  • With L-glutamine
  • Without sodium bicarbonate; supplement with 3.5 gm/L sodium bicarbonate
  • Formulated at 21.3 grams of powder per liter of medium;
  • Cell culture tested
Culturing of cartilaginous embryonic bone cells.
back to top
Click's Medium
Product # Product Name Description Application
C 5572 Click's Medium (Eagle's Ham's amino acids, EHAA)
  • Liquid
  • With sodium bicarbonate
  • Without mercaptoethanol and L-glutamine; supplement with 0.0.584 gm/L L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
back to top
CMRL-1066 Medium
CMRL-1066 is a chemically defined medium developed in the late 1960's at the Connaugh Medical Research Laboratories.  A less complex and extensively modified version of Medium 199, CMRL-1066 was designed initially for use with mouse L-cells in non-supplemented culture.  Although developed for use in serum-free culture, CMRL-1066 can be supplemented with serum and used to support the growth of many cell types.
Product # Product Name Description Application
C 0422 CMRL-1066 Medium
  • Powder
  • With L-glutamine
  • Without sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate
  • Cell culture tested
Suitable for growing a wide variety of cell types, especially fibroblast-like cells.
back to top
Fischer's Medium
Fischer's Medium was originally formulated to support serial propagation of cells from leukemic mice.  In initial studies, cells in culture were examined for resistance to chemotherapeutic agents.  While these studies were typically carried out in whole animals, Fischer's medium made it possible to conduct parallel studies in vitro.  Fischer's medium supports clonal reproduction of cells, particularly lymphoblasts from primary explants or from cells in culture.
Product # Product Name Description Application
F 5008 Fischer's Medium
  • Powder
  • With L-glutamine
  • Without sodium bicarbonate; supplement with 1.125 gm/L sodium bicarbonate;
  • Formulated at 10.6 grams of powder per liter of medium;
  • Cell culture tested
Useful for cloning and low density culture of hematopoeitic cells, especially lymphocytes.
back to top
Glascow Minimum Essential Medium (GMEM)
Glascow Minimum Essential Medium was originally developed by Ian MacPherson and Michael Stoker as a modification of Eagle's medium (BME). The modifications included adding 10% tryptose phosphate and twice the normal concentration of amino acids and vitamins.  This medium was used to study the genetic factors affecting cell competence.  Polyoma virus was used to transform four fibroblast clones from a culture of baby hamster kidney cells.
Product # Product Name Description Application
G 5154 Glasgow Minimum Essential Medium
  • Powder
  • With L-glutamine
  • Without tryptose phosphate broth and sodium bicarbonate; supplement with 2.75 gm/L sodium bicarbonate;
  • Formulated at 12.5 grams of powder per liter of medium;
  • Cell culture tested
For use with adherent kidney cell lines such as baby hamster kidney cells (BHK).
Complete GMEM formulation by supplementing  with tryptose phosphate broth (T 8159) at 100 mL per liter of medium.
G 6148 Glasgow Minimum Essential Medium
  • Liquid
  • With sodium bicarbonate
  • Without L-glutamine; supplement with 0.292 gm/L L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Liquid version of G 5154.
Complete GMEM formulation by supplementing  with tryptose phosphate broth (T 8159) at 100 mL per liter of medium.
back to top
Iscove's Modified Dulbecco's Medium (IMDM)
Guilber and Iscove demonstrated that precursor cells of erythrocytes and macrophages could be cultured in a totally defined serum-free medium supplemented with albumin, transferrin, lecithin, and selenium.  This medium is a modification of Dulbecco's Modified Eagle's medium (DME) and contains selenium, additional amino acids and vitamins, sodium pyruvate, HEPES buffer and potassium nitrate instead of ferric nitrate.
Studies have demonstrated that Iscove's medium supports murine B lymphocytes, hematopoeitic tissue from bone marrow, B cells stimulated with lipopolysaccharide, T lymphocytes, and a variety of hybrid cells.
Product # Product Name Description Application
I 2510 Iscove's Modified Dulbecco's Medium Hybri-Max.
  • Powder
  • With L-glutamine and 25 mM HEPES
  • Without sodium bicarbonate; supplement with 3.024 gm/L sodium bicarbonate;
  • Formulated at 17.7 grams of powder per liter of medium;
  • Endotoxin tested
  • Hybridoma tested
Hybridoma tested version of I 7633.
Use this product when working with hybridoma cells.
I 3390 Iscove's Modified Dulbecco's Medium Hybri-Max.
  • Liquid
  • With sodium bicarbonate
  • Without L-glutamine; supplement with 0.584 gm/L L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Hybridoma tested
Liquid and hybridoma tested version of I 7633
Use this product when working with hybridoma cells.
I 7633 Iscove's Modified Dulbecco's Medium
  • Powder
  • With L-glutamine and 25 mM HEPES
  • Without sodium bicarbonate; supplement with 3.024 gm/L sodium bicarbonate;
  • Formulated at 17.7 grams of powder per liter of medium;
  • Cell culture tested
IMDM is frequently used for serum-free growth of hematopoeitic cells.  This includes fusion cell lines such as hybridomas.
Recommended as a base formulation for the development of unique serum-free formulations.
HEPES increases the buffer strength of the medium and raises the maximum buffering range.
Recommended for cells that grow better at a pH above 7.2.
back to top
L-15 Medium (Leibovitz)
L-15 Medium (Leibovitz) was originally formulated for use in carbon dioxide free systems requiring sodium bicarbonate supplementation.  L-15 is buffered by its complement of salts, free base amino acids and galactose (in place of glucose).  When properly supplemented, L-15 medium supports established cell lines, such as HEp-2 and LLC-MK2, and primary explants of embryonic and adult human tissue.  Many viruses have been successfully cultivated in this medium.
Product # Product Name Description Application
L 4386 L-15 Medium (Leibovitz)
  • Powder
  • With L-glutamine
  • Formulated at 14.8 grams of powder per liter of medium;
  • Cell culture tested
Primarily used for the culture of adherent primary explants of embryonic and adult human tissue; especially, when these explanted cells; are used for production of virus.
L 5520 L-15 Medium (Leibovitz)
  • Liquid
  • Without L-glutamine; supplement with 0.3 gm/L L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Liquid formulation of L 4386.
Primarily used for the culture of adherent primary explants of embryonic and adult human tissue; especially, when these explanted cells are used for production of virus.
back to top
McCoy's 5A Modified Medium
In 1959, McCoy and his coworkers reported the amino acid requirements for in vitro cultivation of Novikoff Hepatoma cells.  These studies were performed using Basal Medium 5A, which was subsequently modified to create a new medium known as McCoy's 5A medium.  Hsu and Kellogg employed this medium to support the growth of primary cultures derived from normal bone marrow, skin, gingiva, testes, mouse kidney, omentum, adrenal glands, lung, spleen, rat embryos, and other tissues.
Product # Product Name Description Application
M 4892 McCoy's 5A Modified
  • Powder
  • With L-glutamine
  • Without sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate;
  • Formulated at 12.0 grams of powder per liter of medium;
  • Cell culture tested
Used for the primary culture of cells from a wide range of tissues.
Recommend  use for production of virus in primary cell cultures.
M 8403 McCoy's 5A Modified
  • Liquid
  • With sodium bicarbonate
  • Without L-glutamine; supplement with 0.22 gm/L L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Liquid form of M 4892.
Used for the primary culture of cells from a wide range of tissues.
Recommend  use for production of virus in primary cell cultures.
M 9270 McCoy's 5A Modified
  • Powder
  • With L-glutamine
  • Without phenol red and sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate;
  • Formulated at 11.9 grams of powder per liter of medium;
  • Cell culture tested
Same as M 4892, except that phenol red is not included.
Phenol red has been shown to interfere with the growth of some cells at cloning densities.
Use this version of McCoy's 5A when working with stem cells or cells at low densities..
back to top
NCTC Medium
NCTC-135 was developed by the Tissue Culture Section, Laboratory of Biology, National Cancer Institute (NCI), Bethesda, MD. NCTC-135 is a modification of NCTC-109 medium, also developed by the NCI.  NCTC-135 differs from NCTC-109 in that the L-cysteine-HCl, which is toxic to certain cell types, was replaced with L-cystine.  NCTC-109 and the 135 modification were formulated to establish and maintain a strain of mouse cells (L929) in a chemically defined and serum free medium.  Successful establishment of L 929 cells led to further nutritional and  metabolic studies and the establishment of at least ten other cell lines adapted to this medium.
Product # Product Name Description Application
N 1140
NCTC 109 Medium
  • Liquid
  • With sodium bicarbonate
  • Without L-glutamine; supplement with 0.136 gm/L L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested>
NCTC-109 and the 135 modification were formulated to establish and maintain a strain of mouse cells (L929) in a chemically defined and serum free medium.
back to top
Swim's S-77 Medium
Swim's S-77 medium (designated S-77) is a modification of MEM Eagle's medium that lacks biotin and has increased levels of amino acids and inositol.  This medium was designed to support suspension cultures of cells derived from the Novikoff hepatoma.  It is an incomplete basal medium, requiring additions of L-glutamine, L-cystine, calcium chloride and other components.  When properly supplemented, Swim's S-77 medium has been used to establish, in culture, a number of stable cell lines (N1-31, N1-S1/FUdr) derived from the Novikoff hepatoma.
Product # Product Name Description Application
S 2513 Swim's S-77 Medium
  • Powder
  • With 12.5 mg/L streptomycin sulfate.
  • Without penicillin G sodium, L-glutamine, L-cystine, calcium chloride, phenol red, and sodium bicarbonate;
  • Supplement with 0.265 gm/L calcium chloride; 0.012 gm/L L-cystine; 0.292 gm/L  L-glutamine; 2.2 gm/L sodium bicarbonate
  • Formulated at 9.0 grams of powder per liter of medium;
  • Cell culture tested.
Designed to support suspension cultures of cells derived from the Novikoff hepatoma.
back to top
Waymouth Medium
Waymouth Medium MB 752/1 was developed as a totally defined synthetic medium for the cultivation of mouse L929 cells in a serum-free environment.  Clones of L929 cells, conditioned to grow in serum-supplemented or serum free medium were cultivated for more than thirty passages in serum-free Waymouth medium and demonstrated a doubling time of approximately 24 hours.  The applicability of Waymouth Medium MF 752/1 has been extended to include whole organ culture, establishment of carcinoma cell lines from pleural effusions, and the growth of potentially tumorigenic cells prior to their assessment in vitro.
Product # Product Name Description Application
W 1625 Waymouth MB 752/1 Medium
  • Liquid
  • With sodium bicarbonate
  • Without L-glutamine; supplement with 0.3 gm/L L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
For the cultivation of mouse L929 cells in a serum-free environment.  Also used in whole organ culture, establishment of carcinoma cell lines from pleural effusions, and the growth of potentially tumorigenic cells prior to their assessment in vitro.
back to top
William's Medium E
In 1971, Williams et. al. described a procedure for enriching isolated hepatocyte cultures for poylgonal epithelial cells and reducing the number of contaminating fibroblasts.  This method was a sequential plating technique based on the observation that fibroblast-like cells adhere to a substrate more rapidly than epithelial cells.  The isolated epithelial cells resulting from this treatment could then be cultured in a rich medium designate William's Medium D.  Newborn animals were the source of cells used in these studies, as they were in most studies of liver cell culture.  Since newborn liver in not functionally mature, further studies were conducted by Williams and Gunn to explore the possibility of culturing adult liver cells on a long term basis.  The medium developed during the course of these studies was designated William's Medium E.  It has been shown to support the growth in long-term culture of adult liver epithelial cells.
Product # Product Name Description Application
W 4125 Williams' Medium E
  • Powder
  • With L-glutamine
  • Without sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate;
  • Formulated at 10.8 grams of powder per liter of medium;
  • Cell culture tested
For long-term culture of adult liver epithelial cells.
W 4128 Williams' Medium E
  • Liquid
  • With sodium bicarbonate
  • Without L-glutamine; supplement with 0.292 gm/L L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Liquid version of W 4125.
Add glutamine at time of formulation.
W 1878 Williams' Medium E
  • Liquid
  • With sodium bicarbonate
  • Without L-glutamine and phenol red; supplement with 0.292 gm/L L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Same as W 4128, except that phenol red has been removed.
Phenol red has been shown to interfere with the growth of some cells at low or cloning densities.
Use this version of William's E when working with stem cells or cells at low densities.
back to top