Life Science > Cell Culture > Learning Center > ECACC Handbook > Techniques Table 4
Cell Culture
Techniques Table 4
Technology
Suspension
Attached
Max Vol
(ml)
Max S/A
(cm2)
Max cells
(susp)
Max cells
(att)
Advantages
Disadvantages
T Flask
Yes
Yes
150
225
1.5x108
~107
Cheap, disposable, no cleaning / sterilization required
Small scale. Multiples required for larger batches.
Triple Flask
(Yes)
Yes
150
525
1.5x108
3x107
Cheap, disposable, no cleaning / sterilization required
Difficult to harvest attached cells. Multiples required for larger batches.
Cell Factory
N/A
Yes
8000
25280
N/A
1.5x1010
Disposable – Single batch manufacture
Require additional equipment (vessels etc which may require cleaning). Difficult to harvest cells.
Roller Bottles
Yes
Yes
1000
1700
1x109
1x108
Cheap, disposable, no cleaning / sterilization required. Versatile. Automated systems available.
Require "decks" to turn. Multiples required for larger batches. Automation very costly.
Expanded Roller Bottles
N/A
Yes
(1000)
3400
N/A
2x108
As above
As above (no advantage for suspension cells).
Shake Flasks
Yes
N/A
1000
N/A
1 x 109
N/A
Some disposables available.
Suspension Only*. Glass vessels to be cleaned & sterilized. Requires Shaker incubator.
Spinner Flasks
Yes
N/A
1000
N/A
1 x 109
N/A
Some semi-disposables available
Suspension Only* Glass vessels to be cleaned & sterilized. Requires Stirrer-base + incubator.
Back to Alternative Cell Culture Systems
Back to Cell Culture Home