Sigma-Aldrich
Life Science > Cell Culture > Learning Center > ECACC Handbook > Techniques 10
Cell Culture
Techniques 10
Fundamental Techniques in
Cell Culture
. . . a Laboratory Handbook
fundamental_tech_1_cropped
1.0   The ECACC and its Collections
2.0   Design and Equipment for the Cell Culture Laboratory
3.0   Safety Aspects of Cell Culture
4.0   Sourcing of Cell Lines
5.0   Main Types of Cell Culture
6.0   The Cell Environment
7.0   Cryopreservation and Storage of Cell Lines
8.0   Good Cell Banking Practices
9.0   Quality Control Considerations
10.0   Authentication of Cell Lines
    10.1   Authentication Techniques
    10.2   Iso-Enzyme Analysis
    10.3   DNA Fingerprinting
    10.4   Multi Locus DNA Fingerprinting
    10.5   Multiplex - PCR (STR) DNA profiling
11.0   Alternative Cell Culture Systems
12.0   Cell Culture Protocols
Glossary of Terms
Who to Contact
Visit our Cell Culture Home Page
10.0   Authentication of Cell Lines
10.1   Authentication Techniques
Whatever the scope of work to be carried out it is important to know that the work is being conducted using the correct reagents. This is no less important for cell cultures, since if cell cultures are not what they are reported to be then work can be invalidated and resources wasted. There is now considerable evidence of gross cross-contamination of cell lines, in particular with HeLa (Prod. No. 93021013-1v1) where up to 16 lines was offered to ECACC with DNA profiles identical to HeLa. These include Hep 2,WISH, INT 407, Chang liver and Giradi heart. To minimize the risk of working with contaminated cell lines it is advisable to obtain cells from a recognized source such as a culture collection that will have confirmed the identity of the cells as part of the banking process. Tests used to authenticate cell cultures include iso-enzyme analysis, karyotyping/ cytogenetic analysis and more recently molecular techniques of DNA profiling. Whilst most of the techniques above are generalized tests and are applicable to all cell lines additional specific tests may also be required to confirm the presence of a product or antigen of interest.
Back to Top
10.2   Iso-Enzyme Analysis
Iso-enzymes are a series of enzymes present in different species that have similar catalytic properties but differ in their structure. By studying the iso-enzymes present in cell lines it is possible to identify the species from which the cell line was derived. The technique is also used as a means of excluding the possibility of gross cross-contamination of the cell line with another culture of a different species.
The principles upon which iso-enzyme analysis is based are:
  • Each iso-enzyme has multiple gene loci coding for different polypeptides with identical enzyme activity (e.g. lactate dehydrogenase, LD)
  • Electrophoretic migration rates change dependent on sub-unit composition e.g. LD has five possible iso-forms (LD 1-5)
  • Different species have different combinations of these iso-forms
  • Using a typical panel of 4 iso-enzymes a composite picture is built up enabling the species of origin to be determined by the use of reference tables
Back to Top
10.3   DNA Fingerprinting
DNA fingerprinting enables the following:
  • Identification of individual cell lines from the same species
  • Confirmation of the identity of cell banks compared to reference master stocks
  • Detection of cross-contamination
Multi locus DNA fingerprinting and multiplex - PCR DNA profiling are the methods used routinely as part of ECACC's routine cell banking procedures.
Back to Top
10.4   Multi Locus DNA Fingerprinting
  • Uses multi locus Jeffrey’s probes 33.15 or 33.6, along with Southern blotting technology producing a complex banding pattern.
  • Probes cross-hybridize with most common species
  • Has the disadvantage that the profiles require visual interpretation and comparison with other samples can be subjective
Click here for Figure 4. Multi Locus DNA Fingerprinting
Back to Top
10.5   Multiplex - PCR (STR) DNA profiling
  • Uses a set of primers (9 used at ECACC) recognizing micro-satellites using PCR and automated DNA sequencing techniques
  • Primers are species specific and are used only for human cell lines
  • Produces a color-coded banding pattern, that translates into a digital code that can easily be stored on a database and compared to other stored profiles
Click here for Figure 5. PCR DNA Profiling
back to top
back to Cell Culture