Cell Culture
Techniques 2
Fundamental Techniques in
Cell Culture
. . . a Laboratory Handbook
fundamental_tech_1_cropped
1.0   The ECACC and its Collections
2.0   Design and Equipment for the Cell Culture Laboratory
    2.1   Laboratory Design
    2.2   Microbiological Safety Cabinets
    2.3   Centrifuges
    2.4   Incubators
    2.5    Work Surfaces and Flooring
    2.6   Plasticware and Consumables
    2.7   Care and Maintenance of Laboratory Areas
3.0   Safety Aspects of Cell Culture
4.0   Sourcing of Cell Lines
5.0   Main Types of Cell Culture
6.0   The Cell Environment
7.0   Cryopreservation and Storage of Cell Lines
8.0   Good Cell Banking Practices
9.0   Quality Control Considerations
10.0   Authentication of Cell Lines
11.0   Alternative Cell Culture Systems
12.0   Cell Culture Protocols
Glossary of Terms
Who to Contact
2.0   Design and Equipment for the Cell Culture Laboratory
2.1    Laboratory Design
Perhaps one of the most under-rated aspects of tissue culture is the need to design the facility to ensure that good quality material is produced in a safe and efficient manner. Most tissue culture is undertaken in laboratories that have been adapted for the purpose and in conditions that are not ideal. However, as long as a few basic guidelines are adopted this should not compromise the work.
There are several aspects to the design of good tissue culture facilities. Ideally work should be conducted in a single use facility which, if at all possible, should be separated into an area reserved for handling newly received material (quarantine area) and an area for material which is known to be free of contaminants (main tissue culture facility). If this is not possible work should be separated by time with all manipulations on clean material being completed prior to manipulations involving the ‘quarantine material’. Different incubators should also be designated. In addition, the work surfaces should be thoroughly cleaned between activities. All new material should be handled as ‘quarantine material’ until it has been shown to be free of contaminants such as bacteria, fungi and particularly mycoplasma. Conducting tissue culture in a shared facility requires considerable planning and it is essential that a good technique is used throughout to minimize the risk of contamination occurring.
For most cell lines the laboratory should be designated to at least Category 2 based on the Advisory Committee on Dangerous Pathogens (ACDP) guidelines (ACDP, 1985) *. However, the precise category required is dependent upon the cell line and the nature of the work proposed. The guidelines make recommendations regarding the laboratory environment including lighting, heating, the type of work surfaces and flooring and provision of hand washing facilities. In addition it is recommended that laboratories should be run at air pressures that are negative to corridors to contain any risks within the laboratory.
*Advisory Committee on Dangerous Pathogens (1985) Categorization of Biological Agents According to Hazard and Categories of Containment, 4th edition, HSE books, Sudbury, UK.
Back to Top
2.2    Microbiological Safety Cabinets
A microbiological safety cabinet is probably the most important piece of equipment since, when operated correctly, it will provide a clean working environment for the product, whilst protecting the operator from aerosols. In these cabinets operator and/or product protection is provided through the use of HEPA (high efficiency particulate air) filters. The level of containment provided varies according to the class of cabinet used. Cabinets may be ducted to atmosphere or re-circulated through a second HEPA filter before passing to atmosphere.
Click here for Figure 1. Diagram of microbiological safety cabinet airflow patterns
Environmental monitoring with Tryptose Soya Broth agar settle plates inside the cabinet for a minimum of four hours should be a good indicator of how clean a cabinet is. There should be no growth of bacteria or fungi on such plates.
In most cases a class II cabinet is adequate for animal cell culture. However each study must be assessed for its hazard risk and it is possible that additional factors, such as a known virus infection or an uncertain provenance, may require a higher level of containment.
Back to Top
2.3   Centrifuges
Centrifuges are used routinely in tissue culture as part of the subculture routine for most cell lines and for the preparation of cells for cryopreservation. By their very nature centrifuges produce aerosols and thus it is necessary to minimize this risk. This can be achieved by purchasing models that have sealed buckets. Ideally the centrifuge should have a clear lid so that the condition of the load can be observed without opening the lid. This will reduce the risk of the operator being exposed to hazardous material if a centrifuge tube has broken during centrifugation. Care should always be taken not to over-fill the tubes and to balance them carefully. These simple steps will reduce the risk of aerosols being generated. The centrifuge should be situated where it can be easily accessed for cleaning and maintenance. Centrifuges should be checked frequently for signs of corrosion.
Back to Top
2.4    Incubators
Cell cultures require a strictly controlled environment in which to grow. Specialist incubators are used routinely to provide the correct growth conditions, such as temperature, degree of humidity and CO2 levels in a controlled and stable manner. Generally they can be set to run at temperatures in the range 28ºC (for insect cell lines) to 37ºC (for mammalian cell lines) and set to provide CO2 at the required level (e.g. 5-10%). Some incubators also have the facility to control the O2 levels. Copper-coated incubators are also now available. These are reported to reduce the risk of microbial contamination within the incubator due to the microbial inhibitory activity of copper. The inclusion of water bath treatment fluid (Prod. No. S5525) in the incubator water trays will also reduce the risk of bacterial and fungal growth in the water trays. However, there is no substitute for regular cleaning. (Note “Sigma Clean” Prod. No. S5525 is harmful by inhalation, contact with skin or if swallowed and is also a severe irritant.)
Back to Top
2.5    Work Surfaces and Flooring
In order to maintain a clean working environment the laboratory surfaces including benchtops, walls and flooring should be smooth and easy to clean. They should also be waterproof and resistant to a variety of chemicals (such as acids, alkalis, solvents and disinfectants). In areas used for the storage of materials in liquid nitrogen, the floors should be resistant to cracking if any liquid nitrogen is spilt. Refer to Section 7.3 for safety considerations on the use of liquid nitrogen. In addition, the floors and walls should be continuous with a covered skirting area to make cleaning easier and reduce the potential for dust to accumulate. Windows should be sealed. Work surfaces should be positioned at a comfortable working height.
Back to Top
2.6    Plasticware and Consumables
Almost every type of cell culture vessel, together with support consumables such as tubes and pipettes, are commercially available as single use, sterile packed, plasticware. Suppliers include Sigma-Aldrich, Nunc, Greiner, Bibby Sterilin and Corning. The use of such plasticware is more cost effective than recycling glassware, enables a higher level of quality assurance and removes the need for validation of cleaning and sterilization procedures. Plastic tissue culture flasks are usually treated to provide a hydrophilic surface to facilitate attachment of anchorage dependent cells.
Back to Top
2.7    Care and Maintenance of Laboratory Areas
In order to maintain a clean and safe working environment tidiness and cleanliness are key. Obviously all spills should be dealt with immediately. Routine cleaning should also be undertaken involving the cleaning of all work surfaces both inside and outside of the microbiological safety cabinet, the floors and all other pieces of equipment e.g. centrifuges. Humidified incubators are a particular area for concern due to the potential for fungal and bacterial growth in the water trays. This will create a contamination risk that can only be avoided by regular cleaning of the incubator. All major pieces of equipment should be regularly maintained and serviced by qualified engineers.
For Exmaple:
  • Microbiological safety cabinets should be checked six monthly to ensure that they are safe to use in terms of product and user protection. These tests confirm that the airflow is correct and that the HEPA filters are functioning properly.
  • The temperature of an incubator should be regularly checked with a NAMAS (National Accreditation of Measurement and Sampling, UK), or equivalent calibrated thermometer and the temperature adjusted as necessary.
  • Incubator CO2 and O2 levels should also be regularly checked to ensure the levels are being correctly maintained.
back to top
back to Cell Culture