Sigma-Aldrich
Life Science > Cell Culture > Learning Center > Media Cross Reference
In Stock and Ready to Ship
Media Cross Reference
Life Technologies/Gibco Tissue Culture Media Cross Reference Chart for Sigma-Aldrich® Tissue culture products. Find direct replacements for Gibco cell culture products.
Request a quote on all your cell culture liquid media, serum and supplements.
To place an immediate order or view package sizes and prices click on the Sigma® product number.
Stock up now on cell culture media and reagents.
We currently have large amounts of these media available for delivery today. You can depend on these media and reagents to meet your quality specifications and to help you to continue your research without delays from back orders or quality problems.
For more technical information visit our Cell Culture Home Page.
Life Technologies/Gibco to Sigma Product Cross Reference
Liquid products may be directly compared in the table. Any differences in products are noted in the right–hand column.
Life Technologies/Gibco
Product No.
Sigma
Product No.
Product Description Product Differences
10010023 P3813 Phosphate Buffered Saline, pH 7.4 Sigma is powder, Life Technologies/Gibco is liquid
10010031 P3813 Phosphate Buffered Saline, pH 7.4 Sigma is powder, Life Technologies/Gibco is liquid
10010049 P3813 Phosphate Buffered Saline, pH 7.4 Sigma is powder, Life Technologies/Gibco is liquid
10082139 F4135 Fetal Bovine Serum, Heat Inactivated Fetal Bovine Serum, Heat Inactivated
10082147 F4135 Fetal Bovine Serum, Heat Inactivated Exact match. No differences.
10082170 F4135 Fetal Bovine Serum, Heat Inactivated Exact match. No differences.
10099133 12003C Fetal Bovine Serum, Australian Origin Exact match. No differences.
10099141 12003C Fetal Bovine Serum, Australian Origin Exact match. No differences.
10099158 12003C Fetal Bovine Serum, Australian Origin Exact match. No differences.
10100139 F4135 Fetal Bovine Serum, Heat Inactivated Sigma is US origin, Life Technologies/Gibco is Australia origin
10100147 F4135 Fetal Bovine Serum, Heat Inactivated Sigma is US origin, Life Technologies/Gibco is Australia origin
10100154 F4135 Fetal Bovine Serum, Heat Inactivated Sigma is US origin, Life Technologies/Gibco is Australia origin
10131027 G8168 G418 disulfate solution Exact match. No differences.
10131035 G8168 G418 disulfate solution Exact match. No differences.
10177012 C3416 Carbenicillin disodium salt Exact match. No differences.
10213015 C7039 Cefotaxime sodium salt Exact match. No differences.
10313021 D6546 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
10370021 M5650 Minimum Essential Medium Eagle Exact match. No differences.
10370039 M5650 Minimum Essential Medium Eagle Exact match. No differences.
10372019 M8537 MCDB 131 Medium Sigma is powder, Life Technologies/Gibco is liquid
10378016 G1146 L-Glutamine-Penicillin-Streptomycin solution Exact match. No differences.
10437010 F0926 Fetal Bovine Serum, USDA approved Exact match. No differences.
10437028 F0926 Fetal Bovine Serum, USDA approved Exact match. No differences.
10437036 F0926 Fetal Bovine Serum, USDA approved Exact match. No differences.
10438018 12306C Fetal Bovine Serum, Heat Inactivated Exact match. No differences.
10438026 12306C Fetal Bovine Serum, Heat Inactivated Exact match. No differences.
10438034 12306C Fetal Bovine Serum, Heat Inactivated Exact match. No differences.
10564011 D6171 Dulbecco's Modified Eagle's Medium - high glucose Sigma is without L-glutamine, Life Technologies/Gibco contains dipeptide Glutamax
10565018 D8437 Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham Sigma contains L-glutamine, Life Technologies/Gibco contains dipeptide Glutamax
10566016 D0819 Dulbecco's Modified Eagle's Medium AQmedia™ Exact Match, No differences.
10569010 D6429 Dulbecco's Modified Eagle's Medium - high glucose Sigma contains L-glutamine, Life Technologies/Gibco contains dipeptide Glutamax
10575090 P9327 Potassium chloride solution Exact match. No differences.
10576015 L8902 Phytohemagglutinin PHA-M from Phaseolus vulgaris (red kidney bean) Exact match. No differences.
10639011 S0192 Stemline™ Hematopoietic Stem Cell Expansion Medium Recommended alternative.
10687010 H0654 Hygromycin B solution Exact Match, No difference.
10743011 C1490 EX-CELL® CD CHO-3 Media Recommended alternative.
10743029 C1490 EX-CELL® CD CHO-3 Media Recommended alternative.
10744019 S0196 Stemline® Keratinocyte II Medium Recommended alternative.
10907012 G1777 Glial Cell Line-derived Neurotrophic Factor human Exact match. No differences.
10908010 B3795 Brain-derived Neurotrophic Factor human Exact match. No differences.
11018041 A9418 Albumin from bovine serum Exact match. No differences.
11039021 D6434 Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham Sigma is without L-glutamine, Life Technologies/Gibco contains L-glutamine
11039047 D6434 Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham Sigma is without L-glutamine, Life Technologies/Gibco contains L-glutamine
11043023 M4530 Medium 199 Sigma contains phenol red, Life Technologies/Gibco is without phenol red
11067030 H0137 HT Media Supplement (50x) Hybri-Max. Sigma is 50X, lyophilized, Life Technologies/Gibco is 100X, liquid
11090073 M2279 Minimum Essential Medium Eagle Exact match. No differences.
11090081 M2279 Minimum Essential Medium Eagle Exact match. No differences.
11090099 M2279 Minimum Essential Medium Eagle Exact match. No differences.
11095072 M4655 Minimum Essential Medium Eagle Exact match. No differences.
11095080 M4655 Minimum Essential Medium Eagle Exact match. No differences.
11095098 M4655 Minimum Essential Medium Eagle Exact match. No differences.
11101011 C6164 Cell Freezing Medium-DMSO (1W) Recommended alternative.
11107018 T1283 holo-Transferrin bovine Exact match. No differences.
11108016 T1428 apo-Transferrin Exact match. No differences.
11120052 M6895 MEM Vitamin Solution (100x) Exact match. No differences.
11130051 M5550 MEM Amino Acids Solution (50W) Exact match. No differences.
11140050 M7145 MEM Non-essential Amino Acid Solution (100x) Exact match. No differences.
11140076 M7145 MEM Non-essential Amino Acid Solution (100x) Exact match. No differences.
11150059 M4530 Medium 199 Exact match. No differences.
11220035 W1625 Waymouth MB 752/1 Medium Sigma is powder, Life Technologies is liquid
11279023 14660C EX-CELL® Sp2/0 Serum-Free Medium Recommended lternative.
11300027 G9771 Grace's Insect Medium Exact match. No differences.
11320033 D8062 Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham Exact match. No differences.
11330032 D8437 Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham Exact match. No differences.
11330057 D8437 Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham Exact match. No differences.
11344041 H4034 HEPES Exact match. No differences.
11360070 S8636 Sodium pyruvate solution Exact match. No differences.
11380037 M8028 Minimum Essential Medium Eagle Exact match. No differences.
11405081 I7760 IPL-41 Insect Medium Exact match. No differences.
11415064 L1518 L-15 Medium (Leibovitz) Exact match. No differences.
11430030 M0275 Minimum Essential Medium Eagle Exact match. No differences.
11530037 C0422 CMRL-1066 Sigma is powder, Life Technologies is liquid
11550043 N6908 Nutrient Mixture F-10 Ham Exact match. No differences.
11575032 M4780 Minimum Essential Medium Eagle Exact match. No differences.
11593019 A0797 Ampicillin sodium salt Exact match. No differences.
11605094 G8142 Grace's Insect Medium Exact match. No differences.
11605102 G8142 Grace's Insect Medium Exact match. No differences.
11667037 G8142 Grace's Insect Medium Sigma is 1X, Life Technologies is 2X
11681020 M3678 MDCK Protein-free Medium Recommended alternative.
11700077 M0769 Minimum Essential Medium Eagle Exact match. No differences.
11710035 G5154 Glasgow Minimum Essential Medium Sigma is without L-glutamine, Life Technologies contains L-glutamine
11720034 S0146 Schneider's Insect Medium Exact match. No differences.
11765047 N6658 Nutrient Mixture F-12 Ham Exact match. No differences.
11765054 N6658 Nutrient Mixture F-12 Ham Exact match. No differences.
11765062 N6658 Nutrient Mixture F-12 Ham Exact match. No differences.
11765070 N6658 Nutrient Mixture F-12 Ham Exact match. No differences.
11800042 L9010 Lactalbumin Enzymatic Hydrolysate Exact match. No differences.
11805017 A9415 Actinomycin D Exact match. No differences.
11811023 A1720 G418 disulfate salt Non-branded form of Geneticin
11811031 A1720 G418 disulfate salt Non-branded form of Geneticin
11811098 A1720 G418 disulfate salt Non-branded form of Geneticin
11814019 M3536 Mycophenolic acid from Penicillium brevi-compactum Exact match. No differences.
11815024 K1637 Kanamycin monosulfate from Streptomyces kanamyceticus Exact match. No differences.
11815032 K1637 Kanamycin monosulfate from Streptomyces kanamyceticus Exact match. No differences.
11825015 M0650 Medium 199 Exact match. No differences.
11835030 R8758 RPMI 1640 Medium Sigma contains phenol red, Life Technologies is without phenol red
11835055 R8758 RPMI 1640 Medium Sigma contains phenol red, Life Technologies is without phenol red
11860038 S9137 Streptomycin sulfate salt Exact match. No differences.
11875085 R8758 RPMI 1640 Medium Exact match. No differences.
11875093 R8758 RPMI 1640 Medium Exact match. No differences.
11875101 R8758 RPMI 1640 Medium Exact match. No differences.
11875119 R8758 RPMI 1640 Medium Exact match. No differences.
11875127 R8758 RPMI 1640 Medium Exact match. No differences.
11875135 R8758 RPMI 1640 Medium Exact match. No differences.
11885076 D6046 Dulbecco's Modified Eagle's Medium - low glucose Exact match. No differences.
11885084 D6046 Dulbecco's Modified Eagle's Medium - low glucose Exact match. No differences.
11885092 D6046 Dulbecco's Modified Eagle's Medium - low glucose Exact match. No differences.
11900024 M0644 Minimum Essential Medium Eagle Exact match. No differences.
11900073 M0644 Minimum Essential Medium Eagle Exact match. No differences.
11905031 L0288 Lipid Mixture 1, Chemically Defined Exact match. No differences.
11960044 D5671 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
11960051 D5671 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
11960069 D5671 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
11960077 D5671 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
11965084 D5796 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
11965092 D5796 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
11965118 D5796 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
11965126 D5796 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
11995040 D6429 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
11995065 D6429 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
11995073 D6429 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
11995081 D6429 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
12000022 M0894 Minimum Essential Medium Eagle Exact match. No differences.
12000063 M0894 Minimum Essential Medium Eagle Exact match. No differences.
12040077 H4409 Hybridoma Medium, Animal Component-free Recommended alternative.
12045076 14610C EX-CELL® 610-HSF Serum-Free Medium Recommended alternative.
12045084 14610C EX-CELL® 610-HSF Serum-Free Medium Recommended alternative.
12052098 C5467 EX-CELL® ACF CHO Medium Recommended alternative.
12052114 C5467 EX-CELL® ACF CHO Medium Recommended alternative.
12100046 D5648 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
12100061 D5648 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
12200036 I2510 Iscove's Modified Dulbecco's Medium Exact match. No differences.
12200069 I2510 Iscove's Modified Dulbecco's Medium Exact match. No differences.
12318010 F1051 Fetal Bovine Serum, Canadian Origin Exact match. No differences.
12318028 F1051 Fetal Bovine Serum, Canadian Origin Exact match. No differences.
12318036 F1051 Fetal Bovine Serum, Canadian Origin Exact match. No differences.
12319018 12306C Fetal Bovine Serum, Heat Inactivated Sigma is US origin, Life Technologies is Canadian origin
12319026 12306C Fetal Bovine Serum, Heat Inactivated Sigma is US origin, Life Technologies is Canadian origin
12319034 12306C Fetal Bovine Serum, Heat Inactivated Sigma is US origin, Life Technologies is Canadian origin
12338018 14571C EX-CELL® 293 Medium Recommended alternative.
12330031 M8403 McCoy's 5A Medium Sigma is without L-glutamine, Life Technologies contains L-glutamine
12340030 M7528 Medium 199 Sigma is without L-glutamine, Life Technologies contains L-glutamine
12360038 M7278 Minimum Essential Medium Eagle Exact match. No differences.
12390035 N2147 Nutrient Mixture F-10 Ham Exact match. No differences.
12400024 D8900 Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham Exact match. No differences.
12430047 D6171 Dulbecco's Modified Eagle's Medium - high glucose Sigma is without L-glutamine, Life Technologies contains L-glutamine.
12430054 D6171 Dulbecco's Modified Eagle's Medium - high glucose Sigma is without L-glutamine, Life Technologies contains L-glutamine.
12430062 D6171 Dulbecco's Modified Eagle's Medium - high glucose Sigma is without L-glutamine, Life Technologies contains L-glutamine.
12440046 I3390 Iscove's Modified Dulbecco's Medium Sigma is without L-glutamine, Life Technologies contains L-glutamine
12440053 I3390 Iscove's Modified Dulbecco's Medium Sigma is without L-glutamine, Life Technologies contains L-glutamine
12440061 I3390 Iscove's Modified Dulbecco's Medium Sigma is without L-glutamine, Life Technologies contains L-glutamine
12500062 D0547 Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham Exact match. No differences.
12500096 D0547 Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham Exact match. No differences.
12551032 W4128 Williams' Medium E Exact match. No differences.
12561049 M4526 Minimum Essential Medium Eagle Sigma is without L-glutamine, Life Technologies contains L-glutamine
12561056 M4526 Minimum Essential Medium Eagle Sigma is without L-glutamine, Life Technologies contains L-glutamine
12571048 M8042 Minimum Essential Medium Eagle Sigma is without L-glutamine, Life Technologies contains L-glutamine
12571063 M8042 Minimum Essential Medium Eagle Sigma is without L-glutamine, Life Technologies contains L-glutamine
12571071 M8042 Minimum Essential Medium Eagle Sigma is without L-glutamine, Life Technologies contains L-glutamine
12800017 D7777 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
12800082 D7777 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
13007018 I0516 Insulin solution from bovine pancreas Sigma is liquid, Life Technologies is powder
13008016 T5391 apo-Transferrin Exact match. No differences.
13028014 P1476 Pituitary Extract bovine Recommended alternative.
13241013 F5542 Fibroblast Growth Factor-Acidic human Exact match. No differences.
13244033 P3201 Platelet-Derived Growth Factor-BB human Exact match. No differences.
13245063 I3769 Insulin-like Growth Factor-I human Exact match. No differences.
13247051 E9644 Epidermal Growth Factor human Exact match. No differences.
13256029 F0291 Fibroblast Growth Factor-Basic human Exact match. No differences.
13257019 N6009 Nerve Growth Factor-2.5S from murine submaxillary gland Exact match. No differences.
13290010 N0513 Nerve Growth Factor-7S from murine submaxillary gland Exact match. No differences.
14025076 H8264 Hanks' Balanced Salt Solutions Exact match. No differences.
14025092 H8264 Hanks' Balanced Salt Solutions Exact match. No differences.
14040117 D8662 Dulbecco's Phosphate Buffered Saline Exact match. No differences.
14040133 D8662 Dulbecco's Phosphate Buffered Saline Exact match. No differences.
14040141 D8662 Dulbecco's Phosphate Buffered Saline Exact match. No differences.
14040182 D8662 Dulbecco's Phosphate Buffered Saline Exact match. No differences.
14040216 D8662 Dulbecco's Phosphate Buffered Saline Exact match. No differences.
14065056 H1641 Hanks' Balanced Salt Solutions Sigma contains phenol red, Life Technologies is without phenol red
14080055 D1283 Dulbecco's Phosphate Buffered Saline Exact match. No differences.
14170112 H9394 Hanks' Balanced Salt Solutions Exact match. No differences.
14170120 H9394 Hanks' Balanced Salt Solutions Exact match. No differences.
14170161 H9394 Hanks' Balanced Salt Solutions Exact match. No differences.
14175079 H6648 Hanks' Balanced Salt Solutions Exact match. No differences.
14175095 H6648 Hanks' Balanced Salt Solutions Exact match. No differences.
14175103 H6648 Hanks' Balanced Salt Solutions Exact match. No differences.
14185052 H4641 Hanks' Balanced Salt Solutions Sigma contains phenol red, Life Technologies is without phenol red
14190136 D8537 Dulbecco's Phosphate Buffered Saline Exact match. No differences.
14190144 D8537 Dulbecco's Phosphate Buffered Saline Exact match. No differences.
14190235 D8537 Dulbecco's Phosphate Buffered Saline Exact match. No differences.
14190250 D8537 Dulbecco's Phosphate Buffered Saline Exact match. No differences.
14200075 D1408 Dulbecco's Phosphate Buffered Saline Exact match. No differences.
15023021 G7021 D-(+)-Glucose anhydrous Exact match. No differences.
15040066 E8008 Ethylenediaminetetraacetic acid solution Exact match. No differences.
15050057 T4424 Trypsin solution from porcine pancreas Exact match. No differences.
15050065 T4424 Trypsin solution from porcine pancreas Exact match. No differences.
15070063 P4458 Penicillin - Streptomycin Solution (50W) Stabilized Exact match. No differences.
15090046 T4549 Trypsin solution from porcine pancreas Exact match. No differences.
15140122 P4333 Penicillin - Streptomycin Solution (100x) Stabilized Exact match. No differences.
15140148 P4333 Penicillin - Streptomycin Solution (100x) Stabilized Exact match. No differences.
15160054 K0129 Kanamycin solution from Streptomyces kanamyceticus Exact match. No differences.
15210040 D1925 Colcemid. solution Exact match. No differences.
15212012 D1925 Colcemid. solution Sigma in HBSS, Life Technologies in PBS
15230147 W3500 Water, Tissue Culture Grade Exact match. No differences.
15230162 W3500 Water, Tissue Culture Grade Exact match. No differences.
15230196 W3500 Water, Tissue Culture Grade Exact match. No differences.
15230204 W3500 Water, Tissue Culture Grade Exact match. No differences.
15240062 A5955 Antibiotic Antimycotic Solution, stabilized (100x) Exact match. No differences.
15240096 A5955 Antibiotic Antimycotic Solution, stabilized (100x) Exact match. No differences.
15250061 T8154 Trypan Blue solution Exact match. No differences.
15260037 A8412 Albumin solution from bovine serum Exact match. No differences.
15290018 A2942 Amphotericin B preparation Exact match. No differences.
15295017 A9528 Amphotericin B solubilized Recommended alternative.
15305014 T3924 Trypsin-EDTA solution Sigma is liquid in HBSS, Life Technologies is lyophilized from 0.9% NaCl
15400054 T4174 Trypsin solution from porcine pancreas Exact match. No differences.
15630080 H0887 HEPES solution Exact match. No differences.
15630106 H0887 HEPES solution Exact match. No differences.
15640055 P4083 Penicillin - Streptomycin - Neomycin solution stabilized (100x) Exact match. No differences.
15710064 G1272 Gentamicin solution Exact match. No differences.
15710072 G1272 Gentamicin solution Exact match. No differences.
15750060 G1397 Gentamicin solution Exact match. No differences.
15750078 G1397 Gentamicin solution Exact match. No differences.
16000036 F2442 Fetal Bovine Serum Exact match. No differences.
16000044 F2442 Fetal Bovine Serum Exact match. No differences.
16000069 F2442 Fetal Bovine Serum Exact match. No differences.
16010142 N4637 Newborn Calf Serum Sigma is US origin, Life Technologies is New Zealand origin
16010159 N4637 Newborn Calf Serum Sigma is US origin, Life Technologies is New Zealand origin
16010167 N4637 Newborn Calf Serum Sigma is US origin, Life Technologies is New Zealand origin
16030074 12133C Calf Serum Sigma is US origin, Life Technologies is New Zealand origin
16050114 H1270 Horse Serum Sigma is US origin, Life Technologies is New Zealand origin
16050122 H1270 Horse Serum Sigma is US origin, Life Technologies is New Zealand origin
16050130 H1270 Horse Serum Sigma is US origin, Life Technologies is New Zealand origin
16070096 S2263 Sheep Serum Sigma is US origin, Life Technologies is New Zealand origin
16110082 C5405 Chicken Serum Sigma is US origin, Life Technologies is New Zealand origin
16120099 R4505 Rabbit serum Sigma is US origin, Life Technologies is New Zealand origin
16120107 R4505 Rabbit serum Sigma is US origin, Life Technologies is New Zealand origin
16140063 F4135 Fetal Bovine Serum, Heat Inactivated Recommended higher quality alternative.
16140071 F4135 Fetal Bovine Serum, Heat Inactivated Recommended higher quality alternative.
16140089 F4135 Fetal Bovine Serum, Heat Inactivated Recommended higher quality alternative.
16170060 12133C Calf Serum Sigma is US origin, Life Technologies is New Zealand origin
16170078 12133C Calf Serum Sigma is US origin, Life Technologies is New Zealand origin
16170086 12133C Calf Serum Sigma is US origin, Life Technologies is New Zealand origin
16210064 G6767 Goat Serum Donor Herd Sigma is US origin, Life Technologies is New Zealand origin
16210072 G6767 Goat Serum Donor Herd Sigma is US origin, Life Technologies is New Zealand origin
16600082 M8403 McCoy's 5A Medium Sigma is without L-glutamine, Life Technologies contains L-glutamine
16600108 M8403 McCoy's 5A Medium Sigma is without L-glutamine, Life Technologies contains L-glutamine
17005042 S0196 Stemline® Keratinocyte Medium Recommended product, consult Certificate of Analysis for exact specifications
17018029 C9891 Collagenase from Clostridium histolyticum Recommended product, consult Certificate of Analysis for exact specifications
17075029 T6522 Trypsin inhibitor from Glycine max (soybean) Recommended alternative.
17100017 C6885 Collagenase from Clostridium histolyticum Recommended product, consult Certificate of Analysis for exact specifications
17101015 C9263 Collagenase from Clostridium histolyticum Recommended product, consult Certificate of Analysis for exact specifications
17104019 C5138 Collagenase from Clostridium histolyticum Recommended product, consult Certificate of Analysis for exact specifications
17105041 P3417 Dispase from Bacillus polymyxa Recommended product, consult Certificate of Analysis for exact specifications
18180059 Y1000 Yeast extract Sigma is powder, Life Technologies is liquid
18200048 Y4375 Yeast Extract Ultrafiltrate (50W) Exact match. No differences.
20012027 P3288 Phosphate buffer pH 7.2 at 25° C Sigma is powder, Life Technologies is liquid
20012043 P3288 Phosphate buffer pH 7.2 at 25° C Sigma is powder, Life Technologies is liquid
20012050 P3288 Phosphate buffer pH 7.2 at 25° C Sigma is powder, Life Technologies is liquid
21010046 B1522 Basal Medium Eagle Sigma is without L-glutamine, Life Technologies contains L-glutamine
21013024 D0422 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
21051024 G8540 L-Glutamine Exact match. No differences.
21051040 G8540 L-Glutamine Exact match. No differences.
21063029 D1145 Dulbecco's Modified Eagle's Medium - high glucose Sigma is without L-glutamine, Life Technologies contains L-glutamine
21083027 L1518 L-15 Medium (Leibovitz) Sigma contains phenol red, Life Technologies is without phenol red
21127022 N3520 Nutrient Mixture F-12 Ham Sigma is powder, Life Technologies is liquid
21300025 D5773 Dulbecco's Phosphate Buffered Saline Sigma's does not contain CaCl2, Life Technologies supplies a bottle of CaCl2.
21300058 D5773 Dulbecco's Phosphate Buffered Saline Exact match. No differences.
21340039 N1140 NCTC 109 Medium Exact match. No differences.
21600010 D5652 Dulbecco's Phosphate Buffered Saline Exact match. No differences.
21600044 D5652 Dulbecco's Phosphate Buffered Saline Exact match. No differences.
21600069 D5652 Dulbecco's Phosphate Buffered Saline Exact match. No differences.
21700018 N6760 Nutrient Mixture F-12 Ham Exact match. No differences.
21700075 N6760 Nutrient Mixture F-12 Ham Exact match. No differences.
21810031 N6386 Neomycin trisulfate salt hydrate Exact match. No differences.
21850029 P4932 Polymyxin B sulfate salt Exact match. No differences.
21870076 R0883 RPMI 1640 Medium Exact match. No differences.
21870084 R0883 RPMI 1640 Medium Exact match. No differences.
21870092 R0883 RPMI 1640 Medium Exact match. No differences.
21985023 M7522 2-Mercaptoethanol Sigma is pure reagent, Life Technologies is 55mM solution
22400071 R5886 RPMI-1640 Medium Sigma is without L-glutamine, Life Technologies contains L-glutamine
22400089 R5886 RPMI-1640 Medium Sigma is without L-glutamine, Life Technologies contains L-glutamine
22400097 R5886 RPMI-1640 Medium Sigma is without L-glutamine, Life Technologies contains L-glutamine
22400105 R5886 RPMI-1640 Medium Sigma is without L-glutamine, Life Technologies contains L-glutamine
23017015 L2020 Laminin from Engelbreth-Holm-Swarm murine sarcoma (basement membrane) Exact match. No differences.
23400021 R4130 RPMI-1640 Medium Exact match. No differences.
23400062 R4130 RPMI-1640 Medium Exact match. No differences.
24020117 H9269 Hanks' Balanced Salt Solutions Exact match. No differences.
24020125 H9269 Hanks' Balanced Salt Solutions Exact match. No differences.
24040032 P5556 Pluronic F-68 solution Exact match. No differences.
25030081 G7513 L-Glutamine solution Exact match. No differences.
25030149 G7513 L-Glutamine solution Exact match. No differences.
25080094 S8761 Sodium bicarbonate solution Exact match. No differences.
25200056 T4049 Trypsin-EDTA solution Sigma contains 0.2g/L EDTA-4Na, Life Technologies contains 0.38g/L EDTA-4Na.
25200072 T4049 Trypsin-EDTA solution Sigma contains 0.2g/L EDTA-4Na, Life Technologies contains 0.38g/L EDTA-4Na.
25300054 T3924 Trypsin-EDTA solution Exact match. No differences.
25300062 T3924 Trypsin-EDTA solution Exact match. No differences.
26010066 N4762 Newborn Calf Serum Sigma is US origin, Life Technologies is New Zealand origin
26010074 N4762 Newborn Calf Serum Sigma is US origin, Life Technologies is New Zealand origin
26050070 H1138 Horse Serum Sigma is US origin, Life Technologies is New Zealand origin
26050088 H1138 Horse Serum Sigma is US origin, Life Technologies is New Zealand origin
26140079 F2442 Fetal Bovine Serum Recommended higher quality alternative.
26140087 F2442 Fetal Bovine Serum Recommended higher quality alternative.
26140095 F2442 Fetal Bovine Serum Recommended higher quality alternative.
26250084 P9783 Porcine Serum Sigma is US origin, Life Technologies is New Zealand origin
26400036 F0392 Fetal Bovine Serum Exact match. No differences.
26400044 F0392 Fetal Bovine Serum Exact match. No differences.
27016021 R6767 RPMI 1640 Medium Sigma is without L-glutamine, Life Technologies contains L-glutamine
27250018 T4799 Trypsin porcine pancreas Recommended alternative.
31033020 C5467 CHO Medium Recommended alternative.
31053028 D1145 Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
31062011 H0262 HAT Media Supplement (50W) Hybri-Max. Sigma is 50X, Life Technologies is 100X
31100019 M5017 Medium 199 Exact match. No differences.
31100035 M5017 Medium 199 Exact match. No differences.
31765035 N6658 Nutrient Mixture F-12 Ham Sigma contains L-glutamine, Life Technologies contains dipeptide Glutamax
31800022 R6504 RPMI 1640 Medium Exact match. No differences.
31800089 R6504 RPMI 1640 Medium Exact match. No differences.
31800105 R6504 RPMI 1640 Medium Exact match. No differences.
31980030 I3390 Iscove's Modified Dulbecco's Medium Sigma is without L-glutamine, Life Technologies contains dipeptide Glutamax
31985062 M3817 MegaCell™ RPMI-1640 Medium Recommended alternative.
31985070 M3817 MegaCell™ RPMI-1640 Medium Recommended alternative.
31985088 M3817 MegaCell™ RPMI-1640 Medium Recommended alternative.
32561037 M4526 Minimum Essential Medium Eagle Sigma is without L-glutamine, Life Technologies contains dipeptide Glutamax
32571036 M8042 Minimum Essential Medium Eagle Sigma is without L-glutamine, Life Technologies contains dipeptide Glutamax
33016015 F2006 Fibronectin from human plasma Exact match. No differences.
35050061 G8541 Ala-Gln Exact match. No differences.
41061029 M8042 Minimum Essential Medium Eagle Sigma contains phenol red, without L-glutamine, Life Technologies is without phenol red, contains L-glutamine
41090036 M0446 Minimum Essential Medium AQmedia™ Exact match. No differences.
41300039 L4386 L-15 Medium (Leibovitz) Exact match. No differences.
41300070 L4386 L-15 Medium (Leibovitz) Exact match. No differences.
41400045 I3146 ITS Liquid Media Supplement (100x) Exact match. No differences.
41500018 M0643 Minimum Essential Medium Eagle Exact match. No differences.
41500034 M0643 Minimum Essential Medium Eagle Exact match. No differences.
41500083 M0643 Minimum Essential Medium Eagle Exact match. No differences.
42360032 M7278 Minimum Essential Medium Eagle Sigma is without L-glutamine, Life Technologies contains dipeptide Glutamax
51200038 M2279 Minimum Essential Medium Eagle Sigma contains phenol red, Life Technologies is without phenol red
51300044 S5791 SPIT Medium Supplement (100x) Exact match. No differences.
51500056 S4920 SITE Liquid Media Supplement (100x) Exact match. No differences.
51985034 M3817 MegaCell™ RPMI-1640 Medium Recommended alternative.
61100061 M0268 Minimum Essential Medium Eagle Exact match. No differences.
61100087 M0268 Minimum Essential Medium Eagle Exact match. No differences.
61100103 M0268 Minimum Essential Medium Eagle Exact match. No differences.
61870036 R2405 RPMI 1640 AQmedia™ Exact match. No differences.
70011044 P3813 Phosphate Buffered Saline, pH 7.4 Sigma is powder, Life Technologies is liquid
70013032 P3288 Phosphate buffer pH 7.2 at 25° C Sigma is powder, Life Technologies is liquid
72400047 R7388 RPMI-1640 Medium Sigma contains L-glutamine, Life Technologies contains dipeptide Glutamax
Q10019 T7660 Tetracycline hydrochloride Exact match. No differences.
R25001 B8416 Bleomycin sulfate from Streptomyces verticillus Exact match. No differences.
R25005 B8416 Bleomycin sulfate from Streptomyces verticillus Exact match. No differences.