Sigma-Aldrich
Life Science > Cell Culture > Learning Center > Media Formulations > Click's Medium Formulation
Cell Culture
Click's Medium Formulation
Component C5572
[1x] g/L
Inorganic Salts
Calcium Chloride 0.1854
Magnesium Chloride • 6H2O 0.1649
Potassium Chloride 0.4
Potassium Phosphate Dibasic 0.06
Sodium Bicarbonate 1.35
Sodium Chloride 8
Sodium Phosphate Dibasic 0.0479
Amino Acids
Alanine 0.0356
Arginine • HCl 0.3175
Asparagine • H2O 0.06
L-Aspartic Acid 0.0532
Cystine • 2HCl 0.0782
Glutamic Acid 0.0588
Glycine 0.03
Histidine • HCl 0.1047
Isoleucine 0.13
Leucine 0.13
L-Lysine • HCl 0.1812
Methionine 0.0375
Phenylalanine 0.08
Proline 0.046
Serine 0.042
Threonine 0.12
Tryptophan 0.025
Tyrosine • 2Na 0.1189
Valine 0.115
Vitamins
D-Pantothenic Acid • ½Ca 0.002
Choline Chloride 0.002
Folic Acid 0.002
myo-Inositol 0.004
Niacinamide 0.002
Pyridoxal • HCl 0.002
Riboflavin 0.0002
Thiamine • HCl 0.002
Other
Glucose 1
Phenol Red • Na 0.01
Sodium Pyruvate 0.275
Add
L-Glutamine 0.584