Cell Culture
Dulbecco's Modified Eagle's Medium/Ham's Nutrient Mixture F-12 (DME/F12) Formulation
|
|
|
| Component |
D0547 g/L |
D2906 g/L |
D6421 [1x] g/L |
D6434 [1x] g/L |
D8062 [1x] g/L |
D8437 [1x] g/L |
D8900 g/L |
D9785 g/L |
| Inorganic Salts |
| Ammonium Molybdate • 4H2O |
0.00000618 |
— |
— |
— |
— |
— |
— |
— |
| Ammonium Metavandate |
0.00000058 |
— |
— |
— |
— |
— |
— |
— |
| Calcium Chloride |
0.1166 |
0.1166 |
0.1166 |
0.1166 |
0.1166 |
0.1166 |
0.1166 |
— |
| Cupric Sulfate • 5H2O |
0.0000013 |
0.0000013 |
0.0000013 |
0.0000013 |
0.0000013 |
0.0000013 |
0.0000013 |
0.0000013 |
| Ferric Nitrate • 9H2O |
0.00005 |
0.00005 |
0.00005 |
0.00005 |
0.00005 |
0.00005 |
0.00005 |
0.00005 |
| Ferrous Sulfate • 7H2O |
0.000417 |
0.000417 |
0.000417 |
0.000417 |
0.000417 |
0.000417 |
0.000417 |
0.000417 |
| Manganese Sulfate |
0.000000151 |
— |
— |
— |
— |
— |
— |
— |
| Magnesium Chloride • 6H2O |
0.0612 |
0.0612 |
0.0612 |
0.0612 |
0.0612 |
0.0612 |
0.0612 |
— |
| Magnesium Sulfate (anhydrous) |
0.04884 |
0.04884 |
0.04884 |
0.04884 |
0.04884 |
0.04884 |
0.04884 |
— |
| Nickel Chloride |
0.00000012 |
— |
— |
— |
— |
— |
— |
— |
| Potassium Chloride |
0.3118 |
0.3118 |
0.3118 |
0.3118 |
0.3118 |
0.3118 |
0.3118 |
0.3118 |
| Sodium Bicarbonate |
— |
— |
1.2 |
— |
— |
— |
— |
— |
| Sodium Chloride |
6.996 |
6.996 |
6.996 |
6.996 |
6.996 |
6.996 |
6.996 |
6.996 |
| Sodium Metasilicate • 9H2O |
0.0000142 |
— |
— |
— |
— |
— |
— |
— |
| Sodium Selenite |
0.00000519 |
— |
— |
— |
— |
— |
— |
— |
| Sodium Phosphate Dibasic (anhydrous) |
0.07102 |
0.07102 |
0.07102 |
0.07102 |
0.07102 |
0.07102 |
0.07102 |
0.07102 |
| Sodium Phosphate Monobasic (anhydrous) |
0.0543 |
0.0543 |
0.0543 |
0.0543 |
0.0543 |
0.0543 |
0.0543 |
0.0543 |
| Stannous Chloride • 2H2O |
0.00000011 |
— |
— |
— |
— |
— |
— |
— |
| Zinc Sulfate • 7H2O |
0.000432 |
0.000432 |
0.000432 |
0.000432 |
0.000432 |
0.000432 |
0.000432 |
0.000432 |
| Amino Acids |
| L-Alanine |
0.00445 |
0.00445 |
0.00445 |
0.00445 |
0.00445 |
0.00445 |
0.00445 |
0.00445 |
| L-Arginine • HCl |
0.1475 |
0.1475 |
0.1475 |
0.1475 |
0.1475 |
0.1475 |
0.1475 |
0.1475 |
| L-Asparagine • H2O |
0.0075 |
0.0075 |
0.0075 |
0.0075 |
0.0075 |
0.0075 |
0.0075 |
0.0075 |
| L-Aspartic Acid |
0.00665 |
0.00665 |
0.00665 |
0.00665 |
0.00665 |
0.00665 |
0.00665 |
0.00665 |
| L-Cystine • 2HCl |
0.03129 |
0.03129 |
0.03129 |
0.03129 |
0.03129 |
0.03129 |
0.03129 |
0.03129 |
| L-Cysteine • HCl • H2O |
0.01756 |
0.01756 |
0.01756 |
0.01756 |
0.01756 |
0.01756 |
0.01756 |
0.01756 |
| L-Glutamic Acid |
0.00735 |
0.00735 |
0.00735 |
0.00735 |
0.00735 |
0.00735 |
0.00735 |
0.00735 |
| L-Glutamine |
0.365 |
0.365 |
— |
— |
0.365 |
0.365 |
0.365 |
— |
| Glycine |
0.01875 |
0.01875 |
0.01875 |
0.01875 |
0.01875 |
0.01875 |
0.01875 |
0.01875 |
| L-Histidine • HCl • H2O |
0.03148 |
0.03148 |
0.03148 |
0.03148 |
0.03148 |
0.03148 |
0.03148 |
0.03148 |
| L-Isoleucine |
0.5447 |
0.5447 |
0.5447 |
0.05447 |
0.05447 |
0.05447 |
0.05447 |
0.05447 |
| L-Leucine |
0.05905 |
0.05905 |
0.05905 |
0.05905 |
0.05905 |
0.05905 |
0.05905 |
— |
| L-Lysine • HCl |
0.09125 |
0.09125 |
0.09125 |
0.09125 |
0.09125 |
0.09125 |
0.09125 |
— |
| L-Methionine |
0.01724 |
0.01724 |
0.01724 |
0.01724 |
0.01724 |
0.01724 |
0.01724 |
— |
| L-Phenylalanine |
0.03548 |
0.03548 |
0.03548 |
0.03548 |
0.03548 |
0.03548 |
0.03458 |
0.03548 |
| L-Proline |
0.01725 |
0.01725 |
0.01725 |
0.01725 |
0.01725 |
0.01725 |
0.01725 |
0.01725 |
| L-Serine |
0.02625 |
0.02625 |
0.02625 |
0.02625 |
0.02625 |
0.02625 |
0.02625 |
0.02625 |
| L-Threonine |
0.05345 |
0.05345 |
0.05345 |
0.05345 |
0.05345 |
0.05345 |
0.05345 |
0.05345 |
| L-Tryptophan |
0.00902 |
0.00902 |
0.00902 |
0.00902 |
0.00902 |
0.00902 |
0.00902 |
0.00902 |
| L-Tyrosine • 2Na • 2H2O |
0.05579 |
0.05579 |
0.05579 |
0.05579 |
0.05579 |
0.05579 |
0.05579 |
0.05579 |
| L-Valine |
0.05285 |
0.05285 |
0.05285 |
0.05285 |
0.05285 |
0.05285 |
0.05285 |
0.05285 |
| Vitamins |
| D-Biotin |
0.0000035 |
0.0000035 |
0.0000035 |
0.0000035 |
0.0000035 |
0.0000035 |
0.0000035 |
0.0000035 |
| Choline Chloride |
0.00898 |
0.00898 |
0.00898 |
0.00898 |
0.00898 |
0.00898 |
0.00898 |
0.00898 |
| Folic Acid |
0.00265 |
0.00265 |
0.00265 |
0.00266 |
0.00266 |
0.00266 |
0.00266 |
0.00266 |
| myo-Inositol |
0.0126 |
0.0126 |
0.0126 |
0.0126 |
0.0126 |
0.0126 |
0.0126 |
0.0126 |
| Niacinamide |
0.00202 |
0.00202 |
0.00202 |
0.00202 |
0.00202 |
0.00202 |
0.00202 |
0.00202 |
| D-Pantothenic Acid (hemicalcium) |
0.00224 |
0.00224 |
0.00224 |
0.00224 |
0.00224 |
0.00224 |
0.00224 |
0.00224 |
| Pyridoxal • HCl |
0.002 |
0.002 |
— |
0.002 |
0.002 |
0.002 |
0.002 |
0.002 |
| Pyridoxine • HCl |
0.002031 |
0.002031 |
0.002031 |
— |
— |
0.000031 |
0.000031 |
0.000031 |
| Riboflavin |
0.000219 |
0.000219 |
0.000219 |
0.000219 |
0.000219 |
0.000219 |
0.000219 |
0.000219 |
| Thiamine • HCl |
0.00217 |
0.00217 |
0.00217 |
0.00217 |
0.00217 |
0.00217 |
0.00217 |
0.00217 |
| Vitamin B12 |
0.00068 |
0.00068 |
0.00068 |
0.00068 |
0.00068 |
0.00068 |
0.00068 |
0.00068 |
| Other |
| D-Glucose |
3.15 |
3.15 |
3.15 |
3.15 |
3.15 |
3.15 |
3.15 |
3.15 |
| HEPES |
— |
3.5745 |
3.5745 |
3.5745 |
— |
3.5745 |
3.5745 |
3.5745 |
| Hypoxanthine |
0.0021 |
0.0021 |
0.0021 |
0.0021 |
0.0021 |
0.0021 |
0.0021 |
0.0021 |
| Linoleic Acid |
0.000042 |
0.000042 |
0.000042 |
0.000042 |
0.000042 |
0.000042 |
0.000042 |
0.000042 |
| Phenol Red • Na |
0.00863 |
— |
0.00863 |
— |
0.00863 |
0.00863 |
0.00863 |
— |
| Putrescine • HCl |
0.000081 |
0.000081 |
0.000081 |
0.000081 |
0.000081 |
0.000081 |
0.000081 |
0.000081 |
| Pyruvic Acid • Na |
0.11 |
0.055 |
0.055 |
0.055 |
0.055 |
0.055 |
0.055 |
0.055 |
| DL-Thioctic Acid |
0.000105 |
0.000105 |
0.000105 |
0.000105 |
0.000105 |
0.000105 |
0.000105 |
0.000105 |
| Thymidine |
0.000365 |
0.000365 |
0.000365 |
0.000365 |
0.000365 |
0.000365 |
0.000365 |
0.000365 |
| Add |
| Sodium Bicarbonate |
1.2 |
1.2 |
— |
— |
— |
— |
1.2 |
1.2 |
| Calcium Chloride • 2H2O |
— |
— |
— |
— |
— |
— |
— |
0.1545 |
| L-Glutamine |
— |
— |
0.365 |
0.365 |
— |
— |
— |
0.365 |
| L-Leucine |
— |
— |
— |
— |
— |
— |
— |
0.05905 |
| L-Lysine • HCl |
— |
— |
— |
— |
— |
— |
— |
0.09125 |
| Magnesium Chloride • 6H2O |
— |
— |
— |
— |
— |
— |
— |
0.0612 |
| Magnesium Sulfate |
— |
— |
— |
— |
— |
— |
— |
0.04884 |
| L-Methionine |
— |
— |
— |
— |
— |
— |
— |
0.0172 |
| back to top |
|