Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Culture > Learning Center > Media Formulations > Dulbecco's Modified Eagle's Medium (DME) Formulation
Cell Culture
Dulbecco's Modified Eagle's Medium (DME) Formulation
Component D0422
[1x] g/L
D1145
[1x] g/L
Inorganic Salts
Calcium Chloride 0.2 0.2
Ferric Nitrate • 9H2O 0.0001 0.0001
Magnesium Sulfate (anhydrous) 0.09767 0.09767
Potassium Chloride 0.4 0.4
Sodium Bicarbonate 3.7 3.7
Sodium Chloride 6.4 6.4
Sodium Phosphate Monobasic (anhydrous) 0.109 0.109
Amino Acids
L-Arginine • HCl 0.084 0.084
L-Cystine • 2HCl 0.0626
Glycine 0.03 0.03
L-Histidine • HCl • H2O 0.042 0.042
L-Isoleucine 0.105 0.105
L-Leucine 0.105 0.105
L-Lysine • HCl 1.46 0.146
L-Methionine 0.03
L-Phenylalanine 0.066 0.066
L-Serine 0.042 0.042
L-Threonine 0.095 0.095
L-Tryptophan 0.016 0.016
L-Tyrosine • 2Na •2H2O 0.10379 0.6351
L-Valine 0.094 0.094
Vitamins
Choline Chloride 0.004 0.004
Folic Acid 0.004 0.004
myo-Inositol 0.0072 0.0072
Niacinamide 0.004 0.004
D-Pantothenic Acid (hemicalcium) 0.004 0.004
Pyridoxal • HCl
Pyridoxine • HCl 0.004 0.004
Riboflavin 0.0004 0.0004
Thiamine • HCl 0.004 0.004
Other
D-Glucose 4.5 4.5
Phenol Red • Na 0.0159
Pyruvic Acid • Na 0.11
Add
L-Glutamine 0.584 0.584
back to top
Component D0819
[1x] g/L
D1152
[1x] g/L
D2429
[10x] g/L
D2902
g/L
D5030
g/L
D5523
g/L
Inorganic Salts
Calcium Chloride 0.2 0.2 2 0.2 0.2 0.2
Ferric Nitrate • 9H2O 0.0001 0.0001 0.001 0.0001 0.0001 0.0001
Magnesium Sulfate (anhydrous) 0.09767 0.09767 0.9767 0.09767 0.09767 0.09767
Potassium Chloride 0.4 0.4 4 0.4 0.4 0.4
Sodium Bicarbonate 3.7
Sodium Chloride 6.4 4.4 64 6.4 6.4 6.4
Sodium Phosphate Monobasic (anhydrous) 0.109 0.109 1.09 0.109 0.109 0.109
Amino Acids
L-Alanyl-L-Glutamine 0.869
L-Arginine • HCl 0.084 0.084 0.84 0.084 0.084 0.084
L-Cystine • 2HCl 0.0626 0.0626 0.626 0.0626 0.0626 0.0626
L-Glutamine 0.584 0.584 0.584
Glycine 0.03 0.03 0.3 0.03 0.03 0.03
L-Histidine • HCl • H2O 0.042 0.042 0.42 0.042 0.042 0.042
L-Isoleucine 0.105 0.105 1.05 0.105 0.105 0.105
L-Leucine 0.105 0.105 1.05 0.105 0.105 0.105
L-Lysine • HCl 0.146 0.146 1.46 0.146 0.146 0.146
L-Methionine 0.03 0.03 0.3 0.03 0.03 0.03
L-Phenylalanine 0.066 0.066 0.66 0.066 0.066 0.066
L-Serine 0.042 0.042 0.42 0.042 0.042 0.042
L-Threonine 0.095 0.095 0.95 0.095 0.095 0.095
L-Tryptophan 0.016 0.016 0.16 0.016 0.016 0.016
L-Tyrosine • 2Na • 2H2O 0.10379 0.10379 1.0379 0.10379 0.10379 0.10379
L-Valine 0.094 0.094 0.94 0.094 0.094 0.094
Vitamins
Choline Chloride 0.004 0.004 0.04 0.004 0.004 0.004
Folic Acid 0.004 0.004 0.004 0.004 0.004
myo-Inositol 0.072 0.0072 0.072 0.0072 0.0072 0.0072
Niacinamide 0.004 0.004 0.04 0.004 0.004 0.004
D-Pantothenic Acid (hemicalcium) 0.004 0.004 0.04 0.004 0.004 0.004
Pyridoxal • HCl 0.004 0.004 0.004 0.004
Pyridoxine • HCl 0.004 0.04
Riboflavin 0.0004 0.0004 0.004 0.0004 0.0004 0.0004
Thiamine • HCl 0.004 0.004 0.04 0.004 0.004 0.004
Other
D-Glucose 4.5 4.5 10 1 1
HEPES 5.958
Phenol Red • Na 0.0159 0.0159 0.159 0.0159
Pyruvic Acid • Na 1.1 0.11 0.11
Add
Glucose 1
L-Glutamine 0.584 at 1× 0.584
Sodium Bicarbonate 3.7 3.7 at 1× 3.7 3.7 3.7
back to top
Component D5546
[1x] g/L
D5648
g/L
D5671
[1x] g/L
D5796
[1x] g/L
D5921
[1x] g/L
Inorganic Salts
Calcium Chloride 0.2 0.2 0.2 0.2 0.2
Ferric Nitrate • 9H2O 0.0001 0.0001 0.0001 0.0001 0.0001
Magnesium Sulfate (anhydrous) 0.09767 0.09767 0.09767 0.09767 0.09767
Potassium Chloride 0.4 0.4 0.4 0.4 0.4
Sodium Bicarbonate 3.7 3.7 3.7 3.7
Sodium Chloride 6.4 6.4 6.4 6.4 6.4
Sodium Phosphate Monobasic (anhydrous) 0.109 0.109 0.109 0.109 0.109
Amino Acids
L-Arginine • HCl 0.084 0.084 0.084 0.084 0.084
L-Cystine • 2HCl 0.0626 0.0626 0.0626 0.0626 0.0626
L-Glutamine 0.584 0.584
Glycine 0.03 0.03 0.03 0.03 0.03
L-Histidine • HCl • H2O 0.042 0.042 0.042 0.042 0.042
L-Isoleucine 0.105 0.105 0.105 0.105 0.105
L-Leucine 0.105 0.105 0.105 0.105 0.105
L-Lysine • HCl 0.146 0.146 0.146 0.146 0.146
L-Methionine 0.03 0.03 0.03 0.03 0.03
L-Phenylalanine 0.066 0.066 0.066 0.066 0.066
L-Serine 0.042 0.042 0.042 0.042 0.042
L-Threonine 0.095 0.095 0.095 0.095 0.095
L-Tryptophan 0.016 0.016 0.016 0.016 0.016
L-Tyrosine • 2Na • 2H2O 0.10379 0.10379 0.10379 0.10379 0.10379
L-Valine 0.094 0.094 0.094 0.094 0.094
Vitamins
Choline Chloride 0.004 0.004 0.004 0.004 0.004
Folic Acid 0.004 0.004 0.004 0.004 0.004
myo-Inositol 0.0072 0.0072 0.0072 0.0072 0.0072
Niacinamide 0.004 0.004 0.004 0.004 0.004
D-Pantothenic Acid (hemicalcium) 0.004 0.004 0.004 0.004 0.004
Pyridoxal • HCl 0.004
Pyridoxine • HCl 0.004 0.004 0.004 0.004
Riboflavin 0.0004 0.0004 0.0004 0.0004 0.0004
Thiamine • HCl 0.004 0.004 0.004 0.004 0.004
Other
D-Glucose 1 4.5 4.5 4.5 1
HEPES
Phenol Red • Na 0.0159 0.0159 0.0159 0.0159
Pyruvic Acid • Na 0.11
Add
Glucose
L-Glutamine 0.584 0.584 0.584
Sodium Bicarbonate 3.7
back to top
Component D6046
[1x] g/L
D6171
[1x] g/L
D6429
[1x] g/L
D6546
[1x] g/L
D7777
g/L
D9443
[1x] g/L
Inorganic Salts
Calcium Chloride 0.2 0.2 0.2 0.2 0.2 0.2
Ferric Nitrate • 9H2O 0.0001 0.0001 0.0001 0.0001 0.0001 0.0001
Magnesium Sulfate (anhydrous) 0.09767 0.09767 0.09767 0.09767 0.09767 0.09767
Potassium Chloride 0.4 0.4 0.4 0.4 0.4 0.4
Sodium Bicarbonate 3.7 3.7 3.7 3.7 3.7
Sodium Chloride 6.4 4.4 6.4 6.4 6.4 6.4
Sodium Phosphate Monobasic (anhydrous) 0.109 0.109 0.109 0.109 0.109 0.109
Amino Acids
L-Arginine • HCl 0.084 0.084 0.084 0.084 0.084
L-Cystine • 2HCl 0.0626 0.0626 0.0626 0.0626 0.0626 0.0626
L-Glutamine 0.584 0.584 0.584 0.584
Glycine 0.03 0.03 0.03 0.03 0.03 0.03
L-Histidine • HCl • H2O 0.042 0.042 0.042 0.042 0.042 0.042
L-Isoleucine 0.105 0.105 0.105 0.105 0.105 0.105
L-Leucine 0.105 0.105 0.105 0.105 0.105
L-Lysine • HCl 0.146 0.146 0.146 0.146 0.146
L-Methionine 0.03 0.03 0.03 0.03 0.03 0.03
L-Phenylalanine 0.066 0.066 0.066 0.066 0.066 0.066
L-Serine 0.042 0.042 0.042 0.042 0.042 0.042
L-Threonine 0.095 0.095 0.095 0.095 0.095 0.095
L-Tryptophan 0.016 0.016 0.016 0.016 0.016 0.016
L-Tyrosine • 2Na • 2H2O 0.10379 0.10379 0.10379 0.10379 0.10379 0.10379
L-Valine 0.094 0.094 0.094 0.094 0.094 0.094
Vitamins
Choline Chloride 0.004 0.004 0.004 0.004 0.004 0.004
Folic Acid 0.004 0.004 0.004 0.004 0.004 0.004
myo-Inositol 0.0072 0.0072 0.0072 0.0072 0.0072 0.0072
Niacinamide 0.004 0.004 0.004 0.004 0.004 0.004
D-Pantothenic Acid (hemicalcium) 0.004 0.004 0.004 0.004 0.004 0.004
Pyridoxal • HCl 0.004
Pyridoxine • HCl 0.004 0.004 0.004 0.004 0.004
Riboflavin 0.0004 0.0004 0.0004 0.0004 0.0004 0.0004
Thiamine • HCl 0.004 0.004 0.004 0.004 0.004 0.004
Other
D-Glucose 1 4.5 4.5 4.5 4.5 1
HEPES 5.958
Phenol Red • Na 0.0159 0.0159 0.0159 0.0159 0.0159
Pyruvic Acid • Na 0.11 0.11 0.11 0.11
Add
Glucose
L-Glutamine 0.584 0.584
Sodium Bicarbonate 3.7
back to top