Cell Culture
Nutrient Mixture F-10 Ham Formulation
|
| Component |
N2147 [1x] g/L |
N6013 g/L |
| Inorganic Salts |
| Calcium Chloride |
0.0333 |
0.0333 |
| Cupric Sulfate • 5H2O |
0.0000025 |
0.0000025 |
| Ferrous Sulfate • 7H2O |
0.000834 |
0.000834 |
| Magnesium Sulfate (anhydrous) |
0.07464 |
0.07464 |
| Potassium Chloride |
0.285 |
0.285 |
| Potassium Phosphate Monobasic (anhydrous) |
0.083 |
0.083 |
| Sodium Bicarbonate |
— |
1.2 |
| Sodium Chloride |
7.4 |
7.4 |
| Sodium Phosphate Dibasic (anhydrous) |
0.1537 |
0.1537 |
| Zinc Sulfate • 7H2O |
0.0000288 |
0.0000288 |
| Amino Acids |
| L-Alanine |
0.009 |
0.009 |
| L-Arginine • HCl |
0.211 |
0.211 |
| L-Asparagine • H2O |
0.01501 |
0.01501 |
| L-Aspartic Acid |
0.0133 |
0.0133 |
| L-Cysteine • HCl • H2O |
0.035 |
0.035 |
| L-Glutamic Acid |
0.0147 |
0.0147 |
| Glycine |
0.00751 |
0.00751 |
| L-Histidine • 3HCl • H2O |
0.021 |
0.021 |
| L-Isoleucine |
0.0026 |
0.0026 |
| L-Leucine |
0.0131 |
0.0131 |
| L-Lysine • HCl |
0.0293 |
0.0293 |
| L-Methionine |
0.00448 |
0.00448 |
| L-Phenylalanine |
0.00496 |
0.00496 |
| L-Proline |
0.0115 |
0.0115 |
| L-Serine |
0.0105 |
0.0105 |
| L-Threonine |
0.00357 |
0.00357 |
| L-Tryptophan |
0.0006 |
0.0006 |
| L-Tyrosine • 2Na • 2H2O |
0.00261 |
0.00261 |
| L-Valine |
0.0035 |
0.0035 |
| Vitamins |
| D-Biotin |
0.000024 |
0.000024 |
| Choline Chloride |
0.000698 |
0.000698 |
| Folic Acid |
0.00132 |
0.00132 |
| myo-Inositol |
0.000541 |
0.000541 |
| Niacinamide |
0.000615 |
0.000615 |
| D-Pantothenic Acid (hemicalcium) |
0.000715 |
0.000715 |
| Pyridoxine • HCl |
0.000206 |
0.000206 |
| Riboflavin |
0.000376 |
0.000376 |
| Thiamine • HCl |
0.001 |
0.001 |
| Vitamin B12 |
0.00136 |
0.00136 |
| Other |
| D-Glucose |
1.1 |
1.1 |
| HEPES |
4.77 |
— |
| Hypoxanthine |
0.00408 |
0.00408 |
| Phenol Red (sodium) |
0.0013 |
0.0013 |
| Pyruvic Acid (sodium) |
0.11 |
0.11 |
| Thioctic Acid |
0.00021 |
0.00021 |
| Thymidine |
0.00073 |
0.00073 |
| Add |
| L-Glutamine |
0.146 |
0.146 |
|
| Component |
N6635 g/L |
N6908 [1x] g/L |
| Inorganic Salts |
| Calcium Chloride |
0.0333 |
0.0333 |
| Cupric Sulfate • 5H2O |
0.0000025 |
0.000025 |
| Ferrous Sulfate • 7H2O |
0.000834 |
0.000834 |
| Magnesium Sulfate (anhydrous) |
0.07464 |
0.07464 |
| Potassium Chloride |
0.285 |
0.285 |
| Potassium Phosphate Monobasic (anhydrous) |
0.083 |
0.083 |
| Sodium Bicarbonate |
— |
1.2 |
| Sodium Chloride |
7.4 |
7.4 |
| Sodium Phosphate Dibasic (anhydrous) |
0.1537 |
0.1537 |
| Zinc Sulfate • 7H2O |
0.0000288 |
0.0000288 |
| Amino Acids |
| L-Alanine |
0.009 |
0.009 |
| L-Arginine • HCl |
0.211 |
0.211 |
| L-Asparagine • H2O |
0.01501 |
0.01501 |
| L-Aspartic Acid |
0.0133 |
0.0133 |
| L-Cysteine • HCl • H2O |
0.035 |
0.035 |
| L-Glutamic Acid |
0.0147 |
0.0147 |
| L-Glutamine |
0.146 |
0.146 |
| Glycine |
0.00751 |
0.00751 |
| L-Histidine • HCl • H2O |
0.021 |
0.021 |
| L-Isoleucine |
0.0026 |
0.0026 |
| L-Leucine |
0.0131 |
0.0131 |
| L-Lysine • HCl |
0.0293 |
0.0293 |
| L-Methionine |
0.00448 |
0.00488 |
| L-Phenylalanine |
0.00496 |
0.00496 |
| L-Proline |
0.0115 |
0.0115 |
| L-Serine |
0.0105 |
0.0105 |
| L-Threonine |
0.00357 |
0.00357 |
| L-Tryptophan |
0.0006 |
0.0006 |
| L-Tyrosine • 2Na • 2H2O |
0.00261 |
0.00261 |
| L-Valine |
0.0035 |
0.0035 |
| Vitamins |
| D-Biotin |
0.000024 |
0.000024 |
| Choline Chloride |
0.000698 |
0.000698 |
| Folic Acid |
0.00132 |
0.00132 |
| myo-Inositol |
0.000541 |
0.000541 |
| Niacinamide |
0.000615 |
0.000615 |
| D-Pantothenic Acid (hemicalcium) |
0.000715 |
0.000715 |
| Pyridoxine • HCl |
0.000206 |
0.000206 |
| Riboflavin |
0.000376 |
0.000376 |
| Thiamine • HCl |
0.001 |
0.001 |
| Vitamin B12 |
0.00136 |
0.00136 |
| Other |
| D-Glucose |
1.1 |
1.1 |
| Hypoxanthine |
0.00408 |
0.00408 |
| Phenol Red • Na |
0.0013 |
0.0013 |
| Pyruvic Acid • Na |
0.11 |
0.11 |
| Thioctic Acid |
0.00021 |
0.00021 |
| Thymidine |
0.00073 |
0.00073 |
| Add |
| Sodium Bicarbonate |
1.2 |
— |
| back to top |