| Component |
F6636 g/L |
| Inorganic Salts |
| Calcium Chloride |
0.1245 |
| Cupric Sulfate • 5H2O |
0.0000025 |
| Ferrrous Sulfate • 7H2O |
0.000834 |
| Magnesium Chloride • 6H2O |
0.046659 |
| Magnesium Sulfate (anhydrous) |
0.02528 |
| Potassium Chloride |
0.305 |
| Potassium Phosphate Monobasic |
0.06124 |
| Sodium Chloride |
7.517 |
| Sodium Phosphate Dibasic (anhydrous) |
0.1324 |
| Zinc Sulfate • 7H2O |
0.000863 |
| Amino Acids |
| L-Alanine |
0.018 |
| L-Arginine • HCl |
0.422 |
| L-Asparagine • H2O |
0.03 |
| L-Aspartic Acid |
0.026 |
| L-Cysteine • HCl • H2O |
0.07026 |
| L-Glutamic Acid |
0.03 |
| L-Glutamine |
0.292 |
| Glycine |
0.016 |
| L-Histidine • HCl • H2O |
0.042 |
| L-Isoleucine |
0.0078 |
| L-Leucine |
0.0262 |
| L-Lysine • HCl |
0.073 |
| L-Methionine |
0.009 |
| L-Phenylalanine |
0.01 |
| L-Proline |
0.07 |
| L-Serine |
0.021 |
| L-Threonine |
0.0238 |
| L-Tryptophan |
0.004 |
| L-Tyrosine • 2Na • 2H2O |
0.01586 |
| L-Valine |
0.0234 |
| Vitamins |
| L-Ascorbic Acid |
0.15 |
| D-Biotin |
0.0000073 |
| Choline Chloride |
0.01396 |
| Folic Acid |
0.00132 |
| myo-Inositol |
0.01802 |
| Niacinamide |
0.00004 |
| L-Pantothenic Acid (hemicalcium) |
0.000238 |
| Pyridoxine • HCl |
0.00006 |
| Riboflavin |
0.00004 |
| Thiamine • HCl |
0.000337 |
| Vitamin B12 |
0.00136 |
| Other |
| D-Glucose |
1.802 |
| Hypoxanthine |
0.00404 |
| Linoleic Acid |
0.00009 |
| Phenol Red • Na |
0.00125 |
| Putrescine • 2HCl |
0.000161 |
| Pyruvic Acid • Na |
0.22 |
| Thioctic Acid |
0.000206 |
| Thymidine |
0.0007 |
| Add |
| Sodium Bicarbonate |
2.676 |
| back to top |