|
Nutrient Mixture F-12 Ham
| Component |
N4888 [1x] g/L |
N6658 [1x] g/L |
N6760 g/L |
| Inorganic Salts |
| Calcium Chloride |
0.0333 |
0.0333 |
0.0333 |
| Cupric Sulfate • 5H2O |
0.0000025 |
0.0000025 |
0.0000025 |
| Ferrous Sulfate • 7H2O |
0.000834 |
0.000834 |
0.000834 |
| Magnesium Chloride |
0.0576 |
0.0576 |
0.0576 |
| Potassium Chloride |
0.224 |
0.224 |
0.224 |
| Sodium Bicarbonate |
1.176 |
1.176 |
— |
| Sodium Chloride |
7.599 |
7.599 |
7.599 |
| Sodium Phosphate Dibasic (anhydrous) |
0.14204 |
0.14204 |
0.14204 |
| Zinc Sulfate • 7H2O |
0.000863 |
0.000863 |
0.000863 |
| Amino Acids |
| L-Alanine |
0.009 |
0.009 |
0.009 |
| L-Arginine • HCl |
0.211 |
0.211 |
0.211 |
| L-Asparagine • H2O |
0.01501 |
0.01501 |
0.01501 |
| L-Aspartic Acid |
0.0133 |
0.0133 |
0.0133 |
| L-Cysteine • HCl • H2O |
0.035 |
0.035 |
0.035 |
| L-Glutamic Acid |
0.0147 |
0.0147 |
0.0147 |
| L-Glutamine |
— |
0.146 |
0.146 |
| Glycine |
0.00751 |
0.00751 |
0.00751 |
| L-Histidine • 3HCl • H2O |
0.02096 |
0.02096 |
0.02096 |
| L-Isoleucine |
0.00394 |
0.00394 |
0.00394 |
| L-Leucine |
0.0131 |
0.0131 |
0.0131 |
| L-Lysine • HCl |
0.0365 |
0.0365 |
0.0365 |
| L-Methionine |
0.00448 |
0.00448 |
0.00448 |
| L-Phenylalanine |
0.00496 |
0.00496 |
0.00496 |
| L-Proline |
0.0345 |
0.0345 |
0.345 |
| L-Serine |
0.0105 |
0.0105 |
0.0105 |
| L-Threonine |
0.0119 |
0.0119 |
0.0119 |
| L-Tryptophan |
0.00204 |
0.00204 |
0.00204 |
| L-Tyrosine • 2Na • 2H2O |
0.00778 |
0.00778 |
0.00778 |
| L-Valine |
0.0117 |
0.0117 |
0.0117 |
| Vitamins |
| D-Biotin |
0.0000073 |
0.0000073 |
0.000073 |
| Choline Chloride |
0.01396 |
0.01396 |
0.01396 |
| Folic Acid |
0.00132 |
0.00132 |
0.00132 |
| myo-Inositol |
0.018 |
0.018 |
0.018 |
| Niacinamide |
0.000037 |
0.000037 |
0.000037 |
| D-Pantothenic Acid (hemicalcium) |
0.00048 |
0.00048 |
0.00048 |
| Pyridoxine • HCl |
0.000062 |
0.000062 |
0.000062 |
| Riboflavin |
0.000038 |
0.000038 |
0.000038 |
| Thiamine • HCl |
0.00034 |
0.00034 |
0.00034 |
| Vitamin B12 |
0.00136 |
0.00136 |
0.00136 |
| Other |
| D-Glucose |
1.802 |
1.802 |
1.802 |
| Hypoxanthine |
0.00408 |
0.00408 |
0.00408 |
| Linoleic Acid |
0.000084 |
0.000084 |
0.000084 |
| Phenol Red • Na |
0.0013 |
0.0013 |
0.0013 |
| Putrescine • HCl |
0.000161 |
0.000161 |
0.000161 |
| Pyruvic Acid • Na |
0.11 |
0.11 |
0.11 |
| Thioctic Acid |
0.00021 |
0.00021 |
0.00021 |
| Thymidine |
0.00073 |
0.00073 |
0.00073 |
| Add |
| L-Glutamine |
0.146 |
— |
— |
| Sodium Bicarbonate |
— |
— |
1.176 |
| back to top |
Nutrient Mixture F-12 Ham HEPES Modification
| Component |
N8641 [1x] g/L |
| Inorganic Salts |
| Calcium Chloride |
0.0333 |
| Cupric Sulfate • 5H2O |
0.0000025 |
| Ferrous Sulfate • 7H2O |
0.000834 |
| Magnesium Chloride |
0.0576 |
| Potassium Chloride |
0.224 |
| Sodium Bicarbonate |
1.176 |
| Sodium Chloride |
7.1 |
| Sodium Phosphate Dibasic (anhydrous) |
0.14204 |
| Zinc Sulfate • 7H2O |
0.000863 |
| Amino Acids |
| L-Alanine |
0.009 |
| L-Arginine • HCl |
0.211 |
| L-Asparagine • H2O |
0.01501 |
| L-Aspartic Acid |
0.0133 |
| L-Cysteine • HCl • H2O |
0.035 |
| L-Glutamic Acid |
0.0147 |
| Glycine |
0.00751 |
| L-Histidine • 3HCl • H2O |
0.02096 |
| L-Isoleucine |
0.00394 |
| L-Leucine |
0.0131 |
| L-Lysine • HCl |
0.0365 |
| L-Methionine |
0.00448 |
| L-Phenylalanine |
0.00496 |
| L-Proline |
0.0345 |
| L-Serine |
0.0105 |
| L-Threonine |
0.0119 |
| L-Tryptophan |
0.00204 |
| L-Tyrosine • 2Na • 2H2O |
0.00778 |
| L-Valine |
0.0117 |
| Vitamins |
| D-Biotin |
0.0000073 |
| Choline Chloride |
0.01396 |
| Folic Acid |
0.00132 |
| myo-Inositol |
0.018 |
| Niacinamide |
0.000037 |
| D-Pantothenic Acid (hemicalcium) |
0.000238 |
| Pyridoxine • HCl |
0.000062 |
| Riboflavin |
0.000038 |
| Thiamine • HCl |
0.00034 |
| Vitamin B12 |
0.00136 |
| Other |
| D-Glucose |
1.802 |
| HEPES |
5.958 |
| Hypoxanthine |
0.00408 |
| Linoleic Acid |
0.000084 |
| Phenol Red • Na |
0.0013 |
| Putrescine • HCl |
1.000161 |
| Pyruvic Acid • Na |
0.11 |
| Thioctic Acid |
0.00021 |
| Thymidine |
0.00073 |
| Add |
| L-Glutamine |
0.146 |
| back to top |
Nutrient Mixture F-12 Ham Kaighn's Modification
| Component |
N3520 [1x] g/L |
| Inorganic Salts |
| Calcium Chloride |
0.10207 |
| Cupric Sulfate • 5H2O |
0.0000025 |
| Ferrous Sulfate • 7H2O |
0.000834 |
| Magnesium Chloride |
0.04961 |
| Magnesium Sulfate |
0.19264 |
| Potassium Chloride |
0.28329 |
| Potassium Phosphate Monobasic |
0.058523 |
| Sodium Phosphate Dibasic |
0.11502 |
| Sodium Chloride |
7.5972 |
| Zinc Sulfate • 7H2O |
0.00014375 |
| Amino Acids |
| L-Alanine |
0.017818 |
| L-Arginine |
0.4214 |
| L-Asparagine • H2O |
0.03002 |
| L-Aspartic Acid |
0.02662 |
| L-Cysteine • HCl • H2O |
0.07024 |
| L-Glutamic Acid |
0.02942 |
| L-Glutamine |
0.2922 |
| Glycine |
0.015014 |
| L-Histidine • HCl • H2O |
0.04192 |
| L-Isoleucine |
0.007872 |
| L-Leucine |
0.02624 |
| L-Lysine • HCl |
0.07304 |
| L-Methionine |
0.008952 |
| L-Phenylalanine |
0.009912 |
| L-Proline |
0.06906 |
| L-Serine |
0.02102 |
| L-Threonine |
0.02382 |
| L-Tryptophan |
0.004084 |
| L-Tyrosine |
0.010872 |
| L-Valine |
0.02342 |
| Vitamins |
| D-Biotin |
0.00007329 |
| Choline Chloride |
0.01396 |
| Folic Acid |
0.0013242 |
| Hypoxanthine |
0.004083 |
| myo-Inositol |
0.01802 |
| Niacinamide |
0.00003663 |
| D-Pantothenic Acid (hemicalcium) |
0.000477 |
| Putrescine • 2HCl |
0.0003222 |
| Pyridoxine • HCl |
0.00006168 |
| Riboflavin |
0.00003764 |
| Thiamine • HCl |
0.0003373 |
| Thymidine |
0.0007266 |
| Vitamin B12 |
0.0013554 |
| Other |
| D+-Glucose |
1.26 |
| Phenol Red • Na |
0.00331806 |
| Pyruvic Acid • Na |
0.22 |
| DL-6,8-Thioctic Acid |
0.0002063 |
| Add |
| Sodium Bicarbonate |
2.5 |
| back to top |
|