Sigma-Aldrich
Life Science > Cell Culture > Learning Center > Media Formulations > Iscove's Modified Dulbecco's Media (IMDM) Formulation
Cell Culture
Iscove's Modified Dulbecco's Media (IMDM) Formulation
Component I3390
[1x] g/L
I7633
g/L
Inorganic Salts
Calcium Chloride 0.1653 0.1653
Magnesium Sulfate (anhydrous) 0.09767 0.9767
Potassium Chloride 0.33 0.33
Potassium Nitrate 0.000076 0.000076
Sodium Bicarbonate 3.024 —
Sodium Chloride 4.505 4.505
Sodium Phosphate Monobasic (anhydrous) 0.109 0.109
Sodium Selenite 0.000017 0.000017
Amino Acids
L-Alanine 0.025 0.025
L-Arginine • HCl 0.084 0.084
L-Asparagine • H2O 0.0284 0.0284
L-Aspartic Acid 0.03 0.03
L-Cystine • 2HCl 0.09124 0.09124
L-Glutamic Acid 0.075 0.075
L-Glutamine — 0.584
Glycine 0.03 0.03
L-Histidine • HCl • H2O 0.042 0.042
L-Isoleucine 0.105 0.105
L-Leucine 0.105 0.105
L-Lysine • HCl 0.146 0.146
L-Methionine 0.03 0.03
L-Phenylalanine 0.066 0.066
L-Proline 0.04 0.04
L-Serine 0.042 0.042
L-Threonine 0.095 0.095
L-Tryptophan 0.016 0.016
L-Tyrosine • 2Na • 2H2O 0.10379 0.10379
L-Valine 0.094 0.094
Vitamins
D-Biotin 0.000013 0.000013
Choline Chloride 0.004 0.004
Folic Acid 0.004 0.004
myo-Inositol 0.0072 0.0072
Niacinamide 0.004 0.004
D-Pantothenic Acid (hemicalcium) 0.004 0.004
Pyridoxal • HCl 0.004 0.004
Riboflavin 0.0004 0.0004
Thiamine • HCl 0.004 0.004
Vitamin B12 0.000013 0.000013
Other
D-Glucose 4.5 4.5
HEPES 5.958 5.958
Phenol Red • Na 0.016 0.016
Pyruvic Acid • Na 0.11 0.11
Add
Sodium Bicarbonate — 3.024
L-Glutamine 0.584 —