| Component |
M4530 [1x] g/L |
M5017 g/L |
M7528 [1x] g/L |
M7653 [1x] g/L |
M9163 [10x] g/L |
| Inorganic Salts |
| Calcium Chloride |
0.2 |
0.2 |
0.2 |
0.1396 |
1.396 |
| Ferric Nitrate • 9H2O |
0.00072 |
0.00072 |
0.00072 |
0.00072 |
0.0072 |
| Magnesium Sulfate (anhydrous) |
0.09767 |
0.09767 |
0.09767 |
0.09767 |
0.9767 |
| Potassium Chloride |
0.4 |
0.4 |
0.4 |
0.4 |
4 |
| Potassium Phosphate Monobasic |
— |
— |
— |
0.06 |
0.6 |
| Sodium Acetate (anhydrous) |
0.05 |
0.05 |
0.05 |
0.05 |
0.5 |
| Sodium Bicarbonate |
2.2 |
— |
2.2 |
0.35 |
— |
| Sodium Chloride |
6.8 |
6.8 |
6 |
8 |
80 |
| Sodium Phosphate Dibasic (anhydrous) |
— |
— |
— |
0.04788 |
0.4788 |
| Sodium Phosphate Monobasic (anhydrous) |
0.122 |
0.122 |
0.122 |
— |
— |
| Amino Acids |
| L-Alanine |
0.025 |
0.025 |
0.025 |
0.025 |
0.25 |
| L-Arginine • HCl |
0.07 |
0.07 |
0.07 |
0.07 |
0.7 |
| L-Aspartic Acid |
0.03 |
0.03 |
0.03 |
0.03 |
0.3 |
| L-Cysteine • HCl • H2O |
0.00011 |
0.00011 |
0.00011 |
0.00011 |
0.0011 |
| L-Cystine • 2HCl |
0.026 |
0.026 |
0.026 |
0.026 |
0.26 |
| L-Glutamic Acid |
0.0668 |
0.0668 |
0.0668 |
0.0668 |
0.668 |
| L-Glutamine |
0.1 |
0.1 |
— |
— |
— |
| Glycine |
0.05 |
0.05 |
0.05 |
0.05 |
0.5 |
| L-Histidine • HCl • H2O |
0.02188 |
0.02188 |
0.02188 |
0.02188 |
0.2188 |
| Hydroxy-L-Proline |
0.01 |
0.01 |
0.01 |
0.01 |
0.1 |
| L-Isoleucine |
0.02 |
0.02 |
0.02 |
0.02 |
0.2 |
| L-Leucine |
0.06 |
0.06 |
0.06 |
0.06 |
0.6 |
| L-Lysine • HCl |
0.07 |
0.07 |
0.07 |
0.07 |
0.7 |
| L-Methionine |
0.015 |
0.015 |
0.015 |
0.015 |
0.15 |
| L-Phenylalanine |
0.025 |
0.025 |
0.025 |
0.025 |
0.25 |
| L-Proline |
0.04 |
0.04 |
0.04 |
0.04 |
0.4 |
| L-Serine |
0.025 |
0.025 |
0.025 |
0.025 |
0.25 |
| L-Threonine |
0.03 |
0.03 |
0.03 |
0.03 |
0.3 |
| L-Tryptophan |
0.01 |
0.01 |
0.01 |
0.01 |
0.1 |
| L-Tyrosine • 2Na • 2H2O |
0.05766 |
0.05766 |
0.05766 |
0.05766 |
0.5766 |
| L-Valine |
0.025 |
0.025 |
0.025 |
0.025 |
0.25 |
| Vitamins |
| Ascorbic Acid • Na |
0.0000566 |
0.0000566 |
0.0000566 |
0.0000566 |
0.000566 |
| D-Biotin |
0.00001 |
0.00001 |
0.00001 |
0.00001 |
0.0001 |
| Calciferol |
0.0001 |
0.0001 |
0.0001 |
0.0001 |
0.001 |
| Choline Chloride |
0.0005 |
0.0005 |
0.0005 |
0.0005 |
0.005 |
| Folic Acid |
0.00001 |
0.00001 |
0.00001 |
0.00001 |
0.0001 |
| Menadione (sodium bisulfite) |
0.000016 |
0.000016 |
0.000016 |
0.000016 |
0.00016 |
| myo-Inositol |
0.00005 |
0.00005 |
0.00005 |
0.00005 |
0.0005 |
| Niacinamide |
0.000025 |
0.000025 |
0.000025 |
0.000025 |
0.00025 |
| Nicotinic Acid |
0.000025 |
0.000025 |
0.000025 |
0.000025 |
0.00025 |
| p-Aminobenzoic Acid |
0.00005 |
0.00005 |
0.00005 |
0.00005 |
0.0005 |
| D-Pantothenic Acid (hemicalcium) |
0.00001 |
0.00001 |
0.00001 |
0.00001 |
0.0001 |
| Pyridoxal • HCl |
0.000025 |
0.000025 |
0.000025 |
0.000025 |
0.00025 |
| Pyridoxine • HCl |
0.000025 |
0.000025 |
0.000025 |
0.000025 |
0.00025 |
| Retinol Acetate |
0.00014 |
0.00014 |
0.00014 |
0.00014 |
0.0014 |
| Riboflavin |
0.00001 |
0.00001 |
0.00001 |
0.00001 |
0.0001 |
| DL-α-Tocopherol Phosphate • Na |
0.00001 |
0.00001 |
0.00001 |
0.00001 |
0.0001 |
| Thiamine • HCl |
0.00001 |
0.00001 |
0.00001 |
0.00001 |
0.0001 |
| Other |
| Adenine Sulfate |
0.01 |
0.01 |
0.01 |
0.01 |
0.1 |
| Adenosine Triphosphate • 2Na |
0.001 |
0.001 |
0.001 |
0.001 |
0.01 |
| Adenosine Monophosphate • Na |
0.0002385 |
0.0002385 |
0.0002385 |
0.0002385 |
0.002385 |
| Cholesterol |
0.0002 |
0.0002 |
0.0002 |
0.0002 |
0.002 |
| Deoxyribose |
0.0005 |
0.0005 |
0.0005 |
0.0005 |
0.005 |
| Glucose |
1 |
1 |
1 |
1 |
10 |
| Glutathione (reduced) |
0.00005 |
0.00005 |
0.00005 |
0.00005 |
0.0005 |
| Guanine • HCl |
0.0003 |
0.0003 |
0.0003 |
0.0003 |
0.003 |
| HEPES |
— |
— |
5.958 |
— |
— |
| Hypoxanthine |
0.0003 |
0.0003 |
0.0003 |
0.0003 |
0.003 |
| Phenol Red • Na |
0.0213 |
0.0213 |
0.0213 |
0.0213 |
0.0213 |
| Polyoxyethylenesorbitan Monooleate (TWEEN 80) |
0.02 |
0.02 |
0.02 |
0.02 |
0.2 |
| Ribose |
0.0005 |
0.0005 |
0.0005 |
0.0005 |
0.005 |
| Thymine |
0.0003 |
0.0003 |
0.0003 |
0.0003 |
0.003 |
| Uracil |
0.0003 |
0.0003 |
0.0003 |
0.0003 |
— |
| Xanthine • Na |
0.000344 |
0.000344 |
0.000344 |
0.000344 |
— |
| Add |
| L-Glutamine |
— |
— |
0.1 |
1 |
0.1 at 1× |
| Sodium Bicarbonate |
— |
2.2 |
— |
— |
0.35 at 1× |
| back to top |