|
Minimum Essential Medium Eagle (MEM)
| Component |
M0268 g/L |
M0275 [10x] g/L |
M0446 [1x] g/L |
M0643 g/L |
M1018 g/L |
| Inorganic Salts |
| Calcium Chloride |
0.2 |
2 |
0.2 |
0.2 |
0.1396 |
| Magnesium Sulfate (anhydrous) |
0.09767 |
0.9767 |
0.09767 |
0.09767 |
0.09767 |
| Potassium Chloride |
0.4 |
4 |
0.4 |
0.4 |
0.4 |
| Potassium Phosphate Monobasic (anhydrous) |
— |
— |
— |
— |
0.06 |
| Sodium Bicarbonate |
— |
— |
2.2 |
— |
— |
| Sodium Chloride |
6.8 |
68 |
6.8 |
6.8 |
8 |
| Sodium Phosphate Dibasic (anhydrous) |
— |
— |
— |
— |
0.04788 |
| Sodium Phosphate Monobasic (anhydrous) |
0.122 |
1.22 |
0.122 |
0.122 |
— |
| Amino Acids |
| L-Alanine |
— |
— |
— |
0.0089 |
0.0089 |
| L-Alanyl-L-Glutamine |
— |
— |
0.4344 |
— |
— |
| L-Arginine • HCl |
0.126 |
1.26 |
0.126 |
0.126 |
0.126 |
| L-Asparagine • H2O |
— |
— |
— |
0.015 |
0.015 |
| L-Aspartic Acid |
— |
— |
— |
0.0133 |
0.0133 |
| L-Cystine • 2HCl |
0.0313 |
0.313 |
0.0313 |
0.0313 |
0.0313 |
| L-Glutamic Acid |
— |
— |
— |
0.0147 |
0.0147 |
| L-Glutamine |
0.292 |
— |
0.292 |
0.292 |
0.292 |
| Glycine |
— |
— |
— |
0.0075 |
0.0075 |
| L-Histidine • HCl • H2O |
0.042 |
0.42 |
0.042 |
0.042 |
0.042 |
| L-Isoleucine |
0.052 |
0.52 |
0.052 |
0.052 |
0.052 |
| L-Leucine |
0.052 |
0.52 |
0.052 |
0.052 |
0.052 |
| L-Lysine • HCl |
0.0725 |
0.725 |
0.0725 |
0.0725 |
0.0725 |
| L-Methionine |
0.015 |
0.15 |
0.015 |
0.015 |
0.015 |
| L-Phenylalanine |
0.032 |
0.32 |
0.032 |
0.032 |
0.032 |
| L-Proline |
— |
— |
— |
0.0115 |
0.0115 |
| L-Serine |
— |
— |
— |
0.0105 |
0.0105 |
| L-Threonine |
0.048 |
0.48 |
0.048 |
0.048 |
0.048 |
| L-Tryptophan |
0.01 |
0.1 |
0.01 |
0.01 |
0.01 |
| L-Tyrosine • 2Na • 2H2O |
0.0519 |
0.519 |
0.0519 |
0.0519 |
0.0519 |
| L-Valine |
0.046 |
0.46 |
0.046 |
0.046 |
0.046 |
| Vitamins |
| Choline Chloride |
0.001 |
0.01 |
0.001 |
0.001 |
0.001 |
| Folic Acid |
0.001 |
0.01 |
0.001 |
0.001 |
0.001 |
| myo-Inositol |
0.002 |
0.02 |
0.002 |
0.002 |
0.002 |
| Niacinamide |
0.001 |
0.01 |
0.001 |
0.001 |
0.001 |
| D-Pantothenic Acid (hemicalcium) |
0.001 |
0.01 |
0.001 |
0.001 |
0.001 |
| Pyridoxal • HCl |
0.001 |
0.01 |
0.001 |
0.001 |
0.001 |
| Riboflavin |
0.0001 |
0.001 |
0.0001 |
0.0001 |
0.0001 |
| Thiamine • HCl |
0.001 |
0.01 |
0.001 |
0.001 |
0.001 |
| Other |
| Glucose |
1 |
10 |
1 |
1 |
1 |
| Phenol Red • Na |
0.011 |
0.11 |
0.011 |
0.011 |
0.011 |
| Add |
| L-Glutamine |
— |
0.292 at 1× |
— |
— |
— |
| Sodium Bicarbonate |
2.2 |
2.2 at 1× |
— |
2.2 |
0.35 |
| back to top |
Minimum Essential Medium Eagle (MEM) (cont.)
| Component |
M2279 g/L |
M2414 [1x] g/L |
M4642 [1x] g/L |
M4655 g/L |
| Inorganic Salts |
| Calcium Chloride |
0.2 |
0.2 |
0.2 |
0.2 |
| Magnesium Sulfate (anhydrous) |
0.09767 |
0.09767 |
0.09767 |
0.09767 |
| Potassium Chloride |
0.4 |
0.4 |
0.4 |
0.4 |
| Potassium Phosphate Monobasic (anhydrous) |
— |
— |
0.06 |
— |
| Sodium Bicarbonate |
— |
2.2 |
— |
2.2 |
| Sodium Chloride |
6.8 |
6.8 |
8 |
6.8 |
| Sodium Phosphate Dibasic (anhydrous) |
— |
— |
0.04788 |
— |
| Sodium Phosphate Monobasic (anhydrous) |
0.122 |
0.122 |
— |
0.122 |
| Amino Acids |
| L-Alanine |
— |
— |
— |
— |
| L-Arginine • HCl |
0.126 |
0.126 |
0.126 |
0.126 |
| L-Asparagine • H2O |
— |
— |
— |
— |
| L-Aspartic Acid |
— |
— |
— |
— |
| L-Cystine • 2HCl |
0.0313 |
0.0313 |
0.0313 |
0.0313 |
| L-Glutamic Acid |
— |
— |
— |
— |
| L-Glutamine |
— |
0.292 |
0.292 |
0.292 |
| Glycine |
— |
— |
— |
— |
| L-Histidine • HCl • H2O |
0.042 |
0.042 |
0.042 |
0.042 |
| L-Isoleucine |
0.052 |
0.052 |
0.052 |
0.052 |
| L-Leucine |
0.052 |
0.052 |
0.052 |
0.052 |
| L-Lysine • HCl |
0.0725 |
0.0725 |
0.0725 |
0.0725 |
| L-Methionine |
0.015 |
0.015 |
0.015 |
0.015 |
| L-Phenylalanine |
0.032 |
0.032 |
0.032 |
0.032 |
| L-Proline |
— |
— |
— |
— |
| L-Serine |
— |
— |
— |
— |
| L-Threonine |
0.048 |
0.048 |
0.048 |
0.048 |
| L-Tryptophan |
0.01 |
0.01 |
0.01 |
0.01 |
| L-Tyrosine • 2Na • 2H2O |
0.0519 |
0.0519 |
0.0519 |
0.0519 |
| L-Valine |
0.046 |
0.046 |
0.046 |
0.046 |
| Vitamins |
| Choline Chloride |
0.001 |
0.001 |
0.001 |
0.001 |
| Folic Acid |
0.001 |
0.001 |
0.001 |
0.001 |
| myo-Inositol |
0.002 |
0.002 |
0.002 |
0.002 |
| Niacinamide |
0.001 |
0.001 |
0.001 |
0.001 |
| D-Pantothenic Acid (hemicalcium) |
0.001 |
0.001 |
0.001 |
0.001 |
| Pyridoxal • HCl |
0.001 |
0.001 |
0.001 |
0.001 |
| Riboflavin |
0.0001 |
0.0001 |
0.0001 |
0.0001 |
| Thiamine • HCl |
0.001 |
0.001 |
0.001 |
0.001 |
| Other |
| Glucose |
1 |
1 |
1 |
1 |
| Phenol Red • Na |
0.011 |
0.011 |
0.011 |
0.011 |
| Add |
| L-Glutamine |
0.292 |
— |
— |
— |
| Sodium Bicarbonate |
2.2 |
— |
0.35 |
— |
| back to top |
Minimum Essential Medium Eagle (MEM) (cont.)
| Component |
M4780 [1x] g/L |
M5650 [1x] g/L |
M5775 [1x] g/L |
M9288 [10x] g/L |
| Inorganic Salts |
| Calcium Chloride |
0.1396 |
0.2 |
0.1396 |
1.396 |
| Magnesium Sulfate (anhydrous) |
0.09767 |
0.09767 |
0.09767 |
0.9767 |
| Potassium Chloride |
0.4 |
0.4 |
0.4 |
4 |
| Potassium Phosphate Monobasic (anhydrous) |
0.06 |
— |
0.06 |
0.6 |
| Sodium Bicarbonate |
0.35 |
2.2 |
0.35 |
— |
| Sodium Chloride |
8 |
6.8 |
8 |
80 |
| Sodium Phosphate Dibasic (anhydrous) |
0.04788 |
— |
0.04788 |
0.4788 |
| Sodium Phosphate Monobasic (anhydrous) |
— |
0.122 |
— |
— |
| Amino Acids |
| L-Alanine |
— |
0.0089 |
— |
— |
| L-Arginine • HCl |
0.126 |
0.126 |
0.126 |
1.26 |
| L-Asparagine • H2O |
— |
0.015 |
— |
— |
| L-Aspartic Acid |
— |
0.0133 |
— |
— |
| L-Cystine • 2HCl |
0.0313 |
0.0313 |
0.0313 |
0.313 |
| L-Glutamic Acid |
— |
0.0147 |
— |
— |
| L-Glutamine |
0.292 |
— |
— |
— |
| Glycine |
— |
0.0075 |
— |
— |
| L-Histidine • HCl • H2O |
0.042 |
0.042 |
0.042 |
0.42 |
| L-Isoleucine |
0.052 |
0.052 |
0.052 |
0.52 |
| L-Leucine |
0.052 |
0.052 |
0.052 |
0.52 |
| L-Lysine • HCl |
0.0725 |
0.0725 |
0.0725 |
0.725 |
| L-Methionine |
0.015 |
0.015 |
0.015 |
0.15 |
| L-Phenylalanine |
0.032 |
0.032 |
0.032 |
0.32 |
| L-Proline |
— |
0.0115 |
— |
— |
| L-Serine |
— |
0.0105 |
— |
— |
| L-Threonine |
0.048 |
0.048 |
0.048 |
0.48 |
| L-Tryptophan |
0.01 |
0.01 |
0.01 |
0.1 |
| L-Tyrosine • 2Na • 2H2O |
0.0519 |
0.0519 |
0.0519 |
0.519 |
| L-Valine |
0.046 |
0.046 |
0.046 |
0.46 |
| Vitamins |
| Choline Chloride |
0.001 |
0.001 |
0.001 |
0.01 |
| Folic Acid |
0.001 |
0.001 |
0.001 |
0.01 |
| myo-Inositol |
0.002 |
0.002 |
0.002 |
0.02 |
| Niacinamide |
0.001 |
0.001 |
0.001 |
0.01 |
| D-Pantothenic Acid (hemicalcium) |
0.001 |
0.001 |
0.001 |
0.01 |
| Pyridoxal • HCl |
0.001 |
0.001 |
0.001 |
0.01 |
| Riboflavin |
0.0001 |
0.0001 |
0.0001 |
0.001 |
| Thiamine • HCl |
0.001 |
0.001 |
0.001 |
0.01 |
| Other |
| Glucose |
1 |
1 |
1 |
10 |
| Phenol Red • Na |
0.011 |
0.011 |
0.011 |
0.11 |
| Add |
| L-Glutamine |
— |
0.292 |
0.292 |
0.292 at 1× |
| Sodium Bicarbonate |
— |
— |
— |
0.35 at 1× |
| back to top |
Minimum Essential Medium Eagle (MEM) Alpha Modification
| Component |
M0644 g/L |
M0894 g/L |
M4526 [1x] g/L |
M8042 [1x] g/L |
| Inorganic Salts |
| Calcium Chloride |
0.2 |
0.2 |
0.2 |
0.2 |
| Magnesium Sulfate (anhydrous) |
0.09767 |
0.09767 |
0.09767 |
0.09767 |
| Potassium Chloride |
0.4 |
0.4 |
0.4 |
0.4 |
| Sodium Bicarbonate |
— |
— |
2.2 |
2.2 |
| Sodium Chloride |
6.8 |
6.8 |
6.8 |
6.8 |
| Sodium Phosphate Monobasic (anhydrous) |
0.122 |
0.122 |
0.122 |
0.122 |
| Amino Acids |
| L-Alanine |
0.025 |
0.025 |
0.025 |
0.025 |
| L-Arginine • HCl |
0.126 |
0.126 |
0.126 |
0.126 |
| L-Asparagine • H2O |
0.05 |
0.05 |
0.05 |
0.05 |
| L-Aspartic Acid |
0.03 |
0.03 |
0.03 |
0.03 |
| L-Cysteine • HCl • H2O |
0.1 |
0.1 |
0.1 |
0.1 |
| L-Cystine • 2HCl |
0.0313 |
0.0313 |
0.0313 |
0.0313 |
| L-Glutamic Acid |
0.075 |
0.075 |
0.075 |
0.075 |
| L-Glutamine |
0.292 |
0.292 |
— |
— |
| Glycine |
0.05 |
0.05 |
0.05 |
0.05 |
| L-Histidine • HCl • H2O |
0.042 |
0.042 |
0.042 |
0.042 |
| L-Isoleucine |
0.052 |
0.052 |
0.052 |
0.052 |
| L-Leucine |
0.052 |
0.052 |
0.052 |
0.052 |
| L-Lysine • HCl |
0.0725 |
0.0725 |
0.0725 |
0.0725 |
| L-Methionine |
0.015 |
0.015 |
0.015 |
0.015 |
| L-Phenylalanine |
0.032 |
0.032 |
0.032 |
0.032 |
| L-Proline |
0.04 |
0.04 |
0.04 |
0.04 |
| L-Serine |
0.025 |
0.025 |
0.025 |
0.025 |
| L-Threonine |
0.048 |
0.048 |
0.048 |
0.048 |
| L-Tryptophan |
0.01 |
0.01 |
0.01 |
0.01 |
| L-Tyrosine • 2Na • 2H2O |
0.0519 |
0.0519 |
0.0519 |
0.0519 |
| L-Valine |
0.046 |
0.046 |
0.046 |
0.046 |
| Vitamins |
| L-Ascorbic Acid • Na |
0.05 |
0.05 |
0.05 |
0.05 |
| Biotin |
0.0001 |
0.0001 |
0.0001 |
0.0001 |
| Choline Chloride |
0.001 |
0.001 |
0.001 |
0.001 |
| Folic Acid |
0.001 |
0.001 |
0.001 |
0.001 |
| myo-Inositol |
0.002 |
0.002 |
0.002 |
0.002 |
| Lipoic Acid |
— |
— |
0.0002 |
— |
| Niacinamide |
0.001 |
0.001 |
0.001 |
0.001 |
| D-Pantothenic Acid (hemicalcium) |
0.001 |
0.001 |
0.001 |
0.001 |
| Pyridoxal • HCl |
0.001 |
0.001 |
0.001 |
0.001 |
| Riboflavin |
0.0001 |
0.0001 |
0.0001 |
0.0001 |
| Thiamine • HCl |
0.001 |
0.001 |
0.001 |
0.001 |
| Vitamin B12 |
0.00136 |
0.00136 |
0.00136 |
0.00136 |
| Other |
| Adenosine |
0.01 |
— |
— |
0.01 |
| Cytidine |
0.01 |
— |
— |
0.01 |
| 2'-Deoxyadenosine |
0.01 |
— |
— |
0.01 |
| 2'-Deoxycytidine • HCl |
0.011 |
— |
— |
0.011 |
| 2'-Deoxyguanosine |
0.01 |
— |
— |
0.01 |
| Glucose |
1 |
1 |
1 |
1 |
| Guanosine |
0.01 |
— |
— |
0.01 |
| Phenol Red • Na |
0.011 |
0.011 |
0.011 |
0.011 |
| Pyruvic Acid • Na |
0.11 |
0.11 |
0.11 |
0.11 |
| Thioctic Acid |
0.0002 |
0.0002 |
— |
0.0002 |
| Thymidine |
0.01 |
— |
— |
0.01 |
| Uridine |
0.01 |
— |
— |
0.01 |
| Add |
| L-Glutamine |
— |
— |
0.292 |
0.292 |
| Sodium Bicarbonate |
2.2 |
2.2 |
— |
— |
| back to top |
Minimum Essential Medium Eagle (MEM) HEPES Modification
| Component |
M2645 g/L |
M7278 [1x] g/L |
| Inorganic Salts |
| Calcium Chloride |
0.2 |
0.2 |
| Magnesium Sulfate (anhydrous) |
0.09767 |
0.09767 |
| Potassium Chloride |
0.4 |
0.4 |
| Sodium Bicarbonate |
— |
2.2 |
| Sodium Chloride |
5.5 |
5.5 |
| Sodium Phosphate Monobasic (anhydrous) |
0.122 |
0.122 |
| Amino Acids |
| L-Arginine • HCl |
0.126 |
0.126 |
| L-Cystine • 2HCl |
0.0313 |
0.0313 |
| L-Glutamine |
0.292 |
— |
| L-Histidine • HCl • H2O |
0.042 |
0.042 |
| L-Isoleucine |
0.052 |
0.052 |
| L-Leucine |
0.052 |
0.052 |
| L-Lysine • HCl |
0.0725 |
0.0725 |
| L-Methionine |
0.015 |
0.015 |
| L-Phenylalanine |
0.032 |
0.032 |
| L-Threonine |
0.048 |
0.048 |
| L-Tryptophan |
0.01 |
0.01 |
| L-Tyrosine • 2Na • 2H2O |
0.0519 |
0.0519 |
| L-Valine |
0.046 |
0.046 |
| Vitamins |
| Choline Chloride |
0.001 |
0.001 |
| Folic Acid |
0.001 |
0.001 |
| myo-Inositol |
0.002 |
0.002 |
| Niacinamide |
0.001 |
0.001 |
| D-Pantothenic Acid (hemicalcium) |
0.001 |
0.001 |
| Pyridoxal • HCl |
0.001 |
0.001 |
| Riboflavin |
0.0001 |
0.0001 |
| Thiamine • HCl |
0.001 |
0.001 |
| Other |
| Glucose |
1 |
1 |
| HEPES |
5.958 |
5.958 |
| Phenol Red • Na |
0.011 |
0.011 |
| Add |
| L-Glutamine |
— |
0.292 |
| Sodium Bicarbonate |
2.2 |
— |
| back to top |
Minimum Essential Medium Eagle (MEM) Joklik Modification for Suspension Cultures
| Component |
M0518 g/L |
M8028 [1x] g/L |
| Inorganic Salts |
| Magnesium Chloride • 6H2O |
0.2 |
0.2 |
| Potassium Chloride |
0.4 |
0.4 |
| Sodium Bicarbonate |
— |
2 |
| Sodium Chloride |
6.5 |
6.5 |
| Sodium Phosphate Monobasic (anhydrous) |
1.154 |
1.154 |
| Amino Acids |
| L-Arginine • HCl |
0.126 |
1.26 |
| L-Cystine • 2HCl |
0.0324 |
0.0324 |
| L-Glutamine |
0.292 |
— |
| L-Histidine • HCl • H2O |
0.042 |
0.042 |
| L-Isoleucine |
0.052 |
0.052 |
| L-Leucine |
0.052 |
0.052 |
| L-Lysine • HCl |
0.0725 |
0.0725 |
| L-Methionine |
0.015 |
0.015 |
| L-Phenylalanine |
0.032 |
0.032 |
| L-Threonine |
0.048 |
0.048 |
| L-Tryptophan |
0.01 |
0.01 |
| L-Tyrosine • 2Na • 2H2O |
0.05452 |
0.05452 |
| L-Valine |
0.046 |
0.046 |
| Vitamins |
| Choline Chloride |
0.001 |
0.001 |
| Folic Acid |
0.001 |
0.001 |
| myo-Inositol |
0.002 |
0.002 |
| Niacinamide |
0.001 |
0.001 |
| D-Pantothenic Acid (hemicalcium) |
0.001 |
0.001 |
| Pyridoxal • HCl |
0.001 |
0.001 |
| Riboflavin |
0.0001 |
0.0001 |
| Thiamine • HCl |
0.001 |
0.001 |
| Other |
| Glucose |
2 |
2 |
| Phenol Red • Na |
0.011 |
0.011 |
| Add |
| L-Glutamine |
— |
0.292 |
| Sodium Bicarbonate |
2 |
— |
| back to top |
Minimum Essential Medium Eagle (MEM) Modified
| Component |
M3024 g/L |
| Inorganic Salts |
| Calcium Chloride |
0.2 |
| Magnesium Sulfate (anhydrous) |
0.09767 |
| Potassium Chloride |
0.4 |
| Sodium Chloride |
6.8 |
| Sodium Phosphate Monobasic (anhydrous) |
0.122 |
| Amino Acids |
| L-Alanine |
0.0089 |
| L-Arginine • HCl |
0.126 |
| L-Asparagine • H2O |
0.015 |
| L-Aspartic Acid |
0.0133 |
| L-Cystine • 2HCl |
0.0313 |
| L-Glutamic Acid |
0.0147 |
| Glycine |
0.0075 |
| L-Histidine • HCl • H2O |
0.042 |
| L-Isoleucine |
0.052 |
| L-Leucine |
0.052 |
| L-Lysine • HCl |
0.0725 |
| L-Methionine |
0.015 |
| L-Phenylalanine |
0.032 |
| L-Proline |
0.0115 |
| L-Serine |
0.0105 |
| L-Threonine |
0.048 |
| L-Tryptophan |
0.01 |
| L-Tyrosine • 2Na • 2H2O |
0.0519 |
| L-Valine |
0.046 |
| Vitamins |
| Choline Chloride |
0.001 |
| Folic Acid |
0.001 |
| myo-Inositol |
0.002 |
| Niacinamide |
0.001 |
| D-Pantothenic Acid (hemicalcium) |
0.001 |
| Pyridoxal • HCl |
0.001 |
| Riboflavin |
0.0001 |
| Thiamine • HCl |
0.001 |
| Other |
| Glucose |
1 |
| Add |
| L-Glutamine |
0.292 |
| Sodium Bicarbonate |
2.2 |
| back to top |
Minimum Essential Medium Eagle (MEM) Auto-Mod™ Modified for Autoclaving
| Component |
M0769 g/L |
| Inorganic Salts |
| Calcium Chloride |
0.2 |
| Magnesium Sulfate (anhydrous) |
0.09767 |
| Potassium Chloride |
0.4 |
| Sodium Chloride |
6.8 |
| Sodium Phosphate Monobasic (anhydrous) |
0.122 |
| Sodium Succinate • 6H2O |
0.1 |
| Succinic Acid (free acid) |
0.075 |
| Amino Acids |
| L-Arginine • HCl |
0.126 |
| L-Cystine • 2HCl |
0.0313 |
| L-Histidine • HCl • H2O |
0.042 |
| L-Isoleucine |
0.052 |
| L-Leucine |
0.052 |
| L-Lysine • HCl |
0.0725 |
| L-Methionine |
0.015 |
| L-Phenylalanine |
0.032 |
| L-Threonine |
0.048 |
| L-Tryptophan |
0.01 |
| L-Tyrosine (free base) |
0.036 |
| L-Valine |
0.046 |
| Vitamins |
| Choline Bitartrate |
0.0018 |
| Folic Acid |
0.001 |
| myo-Inositol |
0.002 |
| Niacinamide |
0.001 |
| D-Pantothenic Acid (hemicalcium) |
0.001 |
| Pyridoxal • HCl |
0.001 |
| Riboflavin |
0.0001 |
| Thiamine • HCl |
0.001 |
| Other |
| Glucose |
1 |
| Phenol Red • Na |
0.0064 |
| Add |
| L-Glutamine |
0.292 |
| Sodium Bicarbonate |
2.2 |
| back to top |
Minimum Essential Medium Eagle (S-MEM) Spinner Modification
| Component |
M8167 [1x] g/L |
| Inorganic Salts |
| Magnesium Sulfate (anhydrous) |
0.09767 |
| Potassium Chloride |
0.4 |
| Sodium Chloride |
6.8 |
| Sodium Phosphate Monobasic (anhydrous) |
0.122 |
| Amino Acids |
| L-Arginine • HCl |
0.126 |
| L-Cystine • 2HCl |
0.0313 |
| L-Histidine • HCl • H2O |
0.042 |
| L-Isoleucine |
0.052 |
| L-Leucine |
0.052 |
| L-Lysine • HCl |
0.0725 |
| L-Methionine |
0.015 |
| L-Phenylalanine |
0.032 |
| L-Threonine |
0.048 |
| L-Tryptophan |
0.01 |
| L-Tyrosine • 2Na • 2H2O |
0.0519 |
| L-Valine |
0.046 |
| Vitamins |
| Choline Chloride |
0.001 |
| Folic Acid |
0.001 |
| myo-Inositol |
0.002 |
| Niacinamide |
0.001 |
| D-Pantothenic Acid (hemicalcium) |
0.001 |
| Pyridoxal • HCl |
0.001 |
| Riboflavin |
0.0001 |
| Thiamine • HCl |
0.001 |
| Other |
| Glucose |
1 |
| Phenol Red • Na |
0.011 |
| Sodium Bicarbonate |
2.2 |
| Add |
| L-Glutamine |
0.292 |
| back to top |
|