|
RPMI-1640 Medium
| Component |
R0883 [1x] g/L |
R1145 [10x] g/L |
R1383 g/L |
R1780 [1x] g/L |
R2405 [1x] g/L |
R6504 g/L |
R8758 [1x] g/L |
| Inorganic Salts |
| Calcium Nitrate • 4H2O |
0.1 |
1 |
0.1 |
0.1 |
0.1 |
0.1 |
0.1 |
| Magnesium Sulfate (anhydrous) |
0.04884 |
0.4884 |
0.04884 |
0.04884 |
0.04884 |
0.04884 |
0.04884 |
| Potassium Chloride |
0.4 |
4 |
0.4 |
0.4 |
0.4 |
0.4 |
0.4 |
| Sodium Bicarbonate |
2 |
— |
— |
2 |
2 |
— |
2 |
| Sodium Chloride |
6 |
60 |
6 |
6 |
6 |
6 |
6 |
| Sodium Phosphate Dibasic (anhydrous) |
0.8 |
8 |
0.8 |
0.8 |
0.8 |
0.8 |
0.8 |
| Amino Acids |
| L-Alanyl-L-Glutamine |
— |
— |
— |
— |
0.4344 |
— |
— |
| L-Arginine |
0.2 |
2 |
0.2 |
— |
0.2 |
0.2 |
0.2 |
| L-Asparagine (anhydrous) |
0.05 |
0.5 |
0.05 |
0.05 |
0.05 |
0.05 |
0.05 |
| L-Aspartic Acid |
0.02 |
0.2 |
0.02 |
0.02 |
0.02 |
0.02 |
0.02 |
| L-Cystine • 2HCl |
0.0652 |
0.652 |
0.0652 |
0.0652 |
0.0652 |
0.0652 |
0.0652 |
| L-Glutamic Acid |
0.02 |
0.2 |
0.02 |
0.02 |
0.02 |
0.02 |
0.02 |
| L-Glutamine |
— |
— |
0.3 |
0.3 |
— |
0.3 |
0.3 |
| Glycine |
0.01 |
0.1 |
0.01 |
0.01 |
0.01 |
0.01 |
0.01 |
| L-Histidine |
0.015 |
0.15 |
0.015 |
0.015 |
0.015 |
0.015 |
0.015 |
| Hydroxy-L-Proline |
0.02 |
0.2 |
0.02 |
0.02 |
0.02 |
0.02 |
0.02 |
| L-Isoleucine |
0.05 |
0.5 |
0.05 |
0.05 |
0.05 |
0.05 |
0.05 |
| L-Leucine |
0.05 |
0.5 |
0.05 |
— |
0.05 |
0.05 |
0.05 |
| L-Lysine • HCl |
0.04 |
0.4 |
0.04 |
— |
0.04 |
0.04 |
0.04 |
| L-Methionine |
0.015 |
0.15 |
0.015 |
0.015 |
0.015 |
0.015 |
0.015 |
| L-Phenylalanine |
0.015 |
0.15 |
0.015 |
0.015 |
0.015 |
0.015 |
0.015 |
| L-Proline |
0.02 |
0.2 |
0.02 |
0.02 |
0.02 |
0.02 |
0.02 |
| L-Serine |
0.03 |
0.3 |
0.03 |
0.03 |
0.03 |
0.03 |
0.03 |
| L-Threonine |
0.02 |
0.2 |
0.02 |
0.02 |
0.02 |
0.02 |
0.02 |
| L-Tryptophan |
0.005 |
0.05 |
0.005 |
0.005 |
0.005 |
0.005 |
0.005 |
| L-Tyrosine • 2Na • 2H2O |
0.02883 |
0.2883 |
0.02883 |
0.02883 |
0.02883 |
0.02883 |
0.02883 |
| L-Valine |
0.02 |
0.2 |
0.02 |
0.02 |
0.02 |
0.02 |
0.02 |
| Vitamins |
| D-Biotin |
0.0002 |
0.002 |
0.0002 |
0.002 |
0.002 |
0.0002 |
0.0002 |
| Choline Chloride |
0.003 |
0.03 |
0.003 |
0.003 |
0.003 |
0.003 |
0.003 |
| Folic Acid |
0.001 |
— |
0.001 |
0.001 |
0.001 |
0.001 |
0.001 |
| myo-Inositol |
0.035 |
0.35 |
0.035 |
0.035 |
0.035 |
0.035 |
0.035 |
| Niacinamide |
0.001 |
0.01 |
0.001 |
0.001 |
0.001 |
0.001 |
0.001 |
| p-Aminobenzoic Acid |
0.001 |
0.01 |
0.001 |
0.001 |
0.001 |
0.001 |
0.001 |
| D-Pantothenic Acid (hemicalcium) |
0.00025 |
0.0025 |
0.00025 |
0.00025 |
0.00025 |
0.00025 |
0.00025 |
| Pyridoxine • HCl |
0.001 |
0.01 |
0.001 |
0.001 |
0.001 |
0.001 |
0.001 |
| Riboflavin |
0.0002 |
0.002 |
0.0002 |
0.0002 |
0.0002 |
0.0002 |
0.0002 |
| Thiamine • HCl |
0.001 |
0.01 |
0.001 |
0.001 |
0.001 |
0.001 |
0.001 |
| Vitamin B12 |
0.000005 |
0.00005 |
0.000005 |
0.000005 |
0.000005 |
0.000005 |
0.000005 |
| Other |
| D-Glucose |
2 |
20 |
— |
2 |
2 |
2 |
2 |
| Glutathione (reduced) |
0.001 |
0.01 |
0.001 |
0.001 |
0.001 |
0.001 |
0.001 |
| Phenol Red • Na |
0.0053 |
0.053 |
0.0053 |
— |
0.0053 |
0.0053 |
0.0053 |
| Add |
| L-Glutamine |
0.3 |
0.3 at 1× |
— |
— |
— |
— |
— |
| Sodium Bicarbonate |
— |
2.0 at 1× |
2 |
— |
— |
2 |
— |
| back to top |
RPMI-1640 Medium Dutch Modification
| Component |
R7638 [1x] g/L |
| Inorganic Salts |
| Calcium Nitrate • 4H2O |
0.1 |
| Magnesium Sulfate (anhydrous) |
0.04884 |
| Potassium Chloride |
0.4 |
| Sodium Bicarbonate |
1 |
| Sodium Chloride |
6.4 |
| Sodium Phosphate Dibasic (anhydrous) |
0.8 |
| Amino Acids |
| L-Arginine |
0.2 |
| L-Asparagine (anhydrous) |
0.05 |
| L-Aspartic Acid |
0.02 |
| L-Cystine • 2HCl |
0.0652 |
| L-Glutamic Acid |
0.02 |
| Glycine |
0.01 |
| L-Histidine |
0.015 |
| Hydroxy-L-Proline |
0.02 |
| L-Isoleucine |
0.05 |
| L-Leucine |
0.05 |
| L-Lysine • HCl |
0.04 |
| L-Methionine |
0.015 |
| L-Phenylalanine |
0.015 |
| L-Proline |
0.02 |
| L-Serine |
0.03 |
| L-Threonine |
0.02 |
| L-Tryptophan |
0.005 |
| L-Tyrosine • 2Na • 2H2O |
0.02883 |
| L-Valine |
0.02 |
| Vitamins |
| D-Biotin |
0.0002 |
| Choline Chloride |
0.003 |
| Folic Acid |
0.001 |
| myo-Inositol |
0.035 |
| Niacinamide |
0.001 |
| p-Aminobenzoic Acid |
0.001 |
| D-Pantothenic Acid (hemicalcium) |
0.00025 |
| Pyridoxine • HCl |
0.001 |
| Riboflavin |
0.0002 |
| Thiamine • HCl |
0.001 |
| Vitamin B12 |
0.000005 |
| Other |
| D-Glucose |
2 |
| Glutathione (reduced) |
0.001 |
| HEPES |
4.77 |
| Phenol Red • Na |
0.0053 |
| Add |
| L-Glutamine |
0.3 |
| back to top |
RPMI-1640 Medium HEPES Modification
| Component |
R4130 g/L |
R5886 [1x] g/L |
R8005 g/L |
| Inorganic Salts |
| Calcium Nitrate • 4H2O |
0.1 |
0.1 |
0.1 |
| Magnesium Sulfate (anhydrous) |
0.04884 |
0.04884 |
0.04884 |
| Potassium Chloride |
0.4 |
0.4 |
0.4 |
| Sodium Bicarbonate |
— |
2 |
— |
| Sodium Chloride |
6 |
6 |
5.9 |
| Sodium Phosphate Dibasic (anhydrous) |
0.8 |
0.8 |
0.8 |
| Amino Acids |
| L-Arginine |
0.2 |
0.2 |
0.2 |
| L-Asparagine (anhydrous) |
0.05 |
0.05 |
0.05 |
| L-Aspartic Acid |
0.02 |
0.02 |
0.02 |
| L-Cystine • 2HCl |
0.0652 |
0.0652 |
0.0652 |
| L-Glutamic Acid |
0.02 |
0.02 |
0.02 |
| L-Glutamine |
0.3 |
— |
0.3 |
| Glycine |
0.01 |
0.01 |
0.01 |
| L-Histidine |
0.015 |
0.015 |
0.015 |
| Hydroxy-L-Proline |
0.02 |
0.02 |
0.02 |
| L-Isoleucine |
0.05 |
0.05 |
0.05 |
| L-Leucine |
0.05 |
0.05 |
0.05 |
| L-Lysine • HCl |
0.04 |
0.04 |
0.04 |
| L-Methionine |
0.015 |
0.015 |
0.015 |
| L-Phenylalanine |
0.015 |
0.015 |
0.015 |
| L-Proline |
0.02 |
0.02 |
0.02 |
| L-Serine |
0.03 |
0.03 |
0.03 |
| L-Threonine |
0.02 |
0.02 |
0.02 |
| L-Tryptophan |
0.005 |
0.005 |
0.005 |
| L-Tyrosine • 2Na • 2H2O |
0.02883 |
0.02883 |
0.02883 |
| L-Valine |
0.02 |
0.02 |
0.02 |
| Vitamins |
| D-Biotin |
0.0002 |
0.0002 |
0.0002 |
| Choline Chloride |
0.003 |
0.003 |
0.003 |
| Folic Acid |
0.001 |
0.001 |
0.001 |
| myo-Inositol |
0.035 |
0.035 |
0.035 |
| Niacinamide |
0.001 |
0.001 |
0.001 |
| p-Aminobenzoic Acid |
0.001 |
0.001 |
0.001 |
| D-Pantothenic Acid (hemicalcium) |
0.00025 |
0.00025 |
0.00025 |
| Pyridoxine • HCl |
0.001 |
0.001 |
0.001 |
| Riboflavin |
0.0002 |
0.0002 |
0.0002 |
| Thiamine • HCl |
0.001 |
0.001 |
0.001 |
| Vitamin B12 |
0.000005 |
0.000005 |
0.000005 |
| Other |
| D-Glucose |
2 |
2 |
4.5 |
| Glutathione (reduced) |
0.001 |
0.001 |
0.001 |
| HEPES |
5.96 |
5.96 |
3.5745 |
| Phenol Red • Na |
0.0053 |
0.0053 |
0.0053 |
| Add |
| L-Glutamine |
— |
0.3 |
— |
| Sodium Bicarbonate |
2 |
— |
2 |
| back to top |
RPMI-1640 Medium Modified
| Component |
R7388 [1x] g/L |
R7509 [1x] g/L |
R7513 [1x] g/L |
R8755 g/L |
| Inorganic Salts |
| Calcium Nitrate • 4H2O |
0.1 |
0.1 |
0.1 |
0.1 |
| Magnesium Sulfate (anhydrous) |
0.04884 |
0.04884 |
0.04884 |
0.04884 |
| Potassium Chloride |
0.4 |
0.4 |
0.4 |
0.4 |
| Sodium Bicarbonate |
— |
2 |
2 |
— |
| Sodium Chloride |
6 |
6 |
6 |
6 |
| Sodium Phosphate Dibasic (anhydrous) |
0.8 |
0.8 |
0.8 |
0.8 |
| Amino Acids |
| L-Arginine |
0.2 |
0.2 |
0.2 |
0.2 |
| L-Asparagine (anhydrous) |
0.05 |
0.05 |
0.05 |
0.05 |
| L-Aspartic Acid |
0.02 |
0.02 |
0.02 |
0.02 |
| L-Cystine • 2HCl |
0.0652 |
0.0652 |
— |
0.0652 |
| L-Glutamic Acid |
0.02 |
0.02 |
0.02 |
0.02 |
| L-Glutamine |
0.3 |
— |
— |
0.3 |
| Glycine |
0.01 |
0.01 |
0.01 |
0.01 |
| L-Histidine |
0.015 |
0.015 |
0.015 |
0.015 |
| Hydroxy-L-Proline |
0.02 |
0.02 |
0.02 |
0.02 |
| L-Isoleucine |
0.05 |
0.05 |
0.05 |
0.05 |
| L-Leucine |
0.05 |
0.05 |
0.05 |
0.05 |
| L-Lysine • HCl |
0.04 |
0.04 |
0.04 |
0.04 |
| L-Methionine |
0.015 |
0.015 |
— |
0.015 |
| L-Phenylalanine |
0.015 |
0.015 |
0.015 |
0.015 |
| L-Proline |
0.02 |
0.02 |
0.02 |
0.02 |
| L-Serine |
0.03 |
0.03 |
0.03 |
0.03 |
| L-Threonine |
0.02 |
0.02 |
0.02 |
0.02 |
| L-Tryptophan |
0.005 |
0.005 |
0.005 |
0.005 |
| L-Tyrosine • 2Na • 2H2O |
0.02883 |
0.02883 |
0.02883 |
0.02883 |
| L-Valine |
0.02 |
0.02 |
0.02 |
0.02 |
| Vitamins |
| D-Biotin |
0.0002 |
0.0002 |
0.0002 |
0.0002 |
| Choline Chloride |
0.003 |
0.003 |
0.003 |
0.003 |
| Folic Acid |
0.001 |
0.001 |
0.001 |
0.001 |
| myo-Inositol |
0.035 |
0.035 |
0.035 |
0.035 |
| Niacinamide |
0.001 |
0.001 |
0.001 |
0.001 |
| p-Aminobenzoic Acid |
0.001 |
0.001 |
0.001 |
0.001 |
| D-Pantothenic Acid (hemicalcium) |
0.00025 |
0.00025 |
0.00025 |
0.00025 |
| Pyridoxine • HCl |
0.001 |
0.001 |
0.001 |
0.001 |
| Riboflavin |
0.0002 |
0.0002 |
0.0002 |
0.0002 |
| Thiamine • HCl |
0.001 |
0.001 |
0.001 |
0.001 |
| Vitamin B12 |
0.000005 |
0.000005 |
0.000005 |
0.000005 |
| Other |
| D-Glucose |
2 |
2 |
2 |
2 |
| Glutathione (reduced) |
0.001 |
0.001 |
0.001 |
0.001 |
| HEPES |
4.77 |
— |
— |
— |
| Phenol Red • Na |
0.0053 |
— |
0.0053 |
— |
| Add |
| L-Cystine • 2HCl |
— |
— |
0.0652 |
— |
| L-Glutamine |
— |
0.3 |
0.3 |
— |
| L-Methionine |
— |
— |
0.015 |
— |
| Sodium Bicarbonate |
2 |
— |
— |
2 |
| back to top |
RPMI-1640 Medium Auto-Mod™ Modified for Autoclaving
| Component |
R7755 g/L |
| Inorganic Salts |
| Calcium Nitrate • 4H2O |
0.1 |
| Magnesium Sulfate (anhydrous) |
0.04884 |
| Potassium Chloride |
0.4 |
| Sodium Chloride |
6 |
| Sodium Phosphate Dibasic (anhydrous) |
0.8 |
| Succinic Acid • 6H2O • Na |
0.1 |
| Succinic Acid (free acid) |
0.075 |
| Amino Acids |
| L-Arginine |
0.2 |
| L-Asparagine (anhydrous) |
0.05 |
| L-Aspartic Acid |
0.02 |
| L-Cystine • 2HCl |
0.0652 |
| L-Glutamic Acid |
0.02 |
| Glycine |
0.01 |
| L-Histidine |
0.015 |
| Hydroxy-L-Proline |
0.02 |
| L-Isoleucine |
0.05 |
| L-Leucine |
0.05 |
| L-Lysine • HCl |
0.04 |
| L-Methionine |
0.015 |
| L-Phenylalanine |
0.015 |
| L-Proline |
0.02 |
| L-Serine |
0.03 |
| L-Threonine |
0.02 |
| L-Tryptophan |
0.005 |
| L-Tyrosine |
0.02 |
| L-Valine |
0.02 |
| Vitamins |
| D-Biotin |
0.0002 |
| Choline Bitartrate |
0.00544 |
| Folic Acid |
0.001 |
| myo-Inositol |
0.035 |
| Niacinamide |
0.001 |
| p-Aminobenzoic Acid |
0.001 |
| D-Pantothenic Acid (hemicalcium) |
0.00025 |
| Pyridoxine • HCl |
0.001 |
| Riboflavin |
0.0002 |
| Thiamine • HCl |
0.001 |
| Vitamin B12 |
0.000005 |
| Other |
| D-Glucose |
2 |
| Glutathione (reduced) |
0.001 |
| Phenol Red • Na |
0.00318 |
| Add |
| L-Glutamine |
0.3 |
| Sodium Bicarbonate |
2 |
| back to top |
|