Cell Culture
Waymouth MB 752/1 Medium Formulation
|
|
|
| Component |
W1625 g/L |
| Inorganic Salts |
| Calcium Chloride |
0.0906 |
| Magnesium Chloride |
0.11239 |
| Magnesium Sulfate (anhydrous) |
0.09767 |
| Potassium Chloride |
0.15 |
| Potassium Phosphate Monobasic (anhydrous) |
0.08 |
| Sodium Chloride |
6 |
| Sodium Phosphate Dibasic (anhydrous) |
0.3 |
| Amino Acids |
| L-Arginine • HCl |
0.075 |
| L-Aspartic Acid |
0.06 |
| L-Cysteine • HCl • H2O |
0.10029 |
| L-Cystine • 2HCl |
0.01955 |
| L-Glutamic Acid |
0.15 |
| L-Glutamine |
0.35 |
| Glycine |
0.05 |
| L-Histidine • HCl • H2O |
0.1641 |
| L-Isoleucine |
0.025 |
| L-Leucine |
0.05 |
| L-Lysine • HCl |
0.24 |
| L-Methionine |
0.05 |
| L-Phenylalanine |
0.05 |
| L-Proline |
0.05 |
| L-Threonine |
0.075 |
| L-Tryptophan |
0.04 |
| L-Tyrosine • 2Na • 2H2O |
0.0577 |
| L-Valine |
0.065 |
| Vitamins |
| Ascorbic Acid • Na |
0.0198 |
| D-Biotin |
0.00002 |
| Choline Bitartrate |
0.45352 |
| Folic Acid |
0.0004 |
| myo-Inositol |
0.001 |
| Niacinamide |
0.001 |
| D-Pantothenic Acid (hemicalcium) |
0.001 |
| Pyridoxine • HCl |
0.001 |
| Riboflavin |
0.001 |
| Thiamine • HCl |
0.01 |
| Vitamin B12 |
0.0002 |
| Other |
| D-Glucose |
5 |
| Glutathione (reduced) |
0.015 |
| Hypoxanthine |
0.025 |
| Phenol Red • Na |
0.011 |
| Add |
| Sodium Bicarbonate |
2.24 |
| back to top |
|