| Component |
W1878 [1x] g/L |
W4125 g/L |
W4128 [1x] g/L |
| Inorganic Salts |
| Calcium Chloride |
0.2 |
0.2 |
0.2 |
| Cupric Sulfate • 5H2O |
0.0000001 |
0.0000001 |
0.0000001 |
| Ferric Nitrate • 9H2O |
0.0000001 |
0.0000001 |
0.0000001 |
| Magnesium Chloride • 4H2O |
0.0000001 |
0.0000001 |
0.0000001 |
| Magnesium Sulfate (anhydrous) |
0.0977 |
0.0977 |
0.0977 |
| Potassium Chloride |
0.4 |
0.4 |
0.4 |
| Sodium Bicarbonate |
2.2 |
— |
2.2 |
| Sodium Chloride |
6.8 |
6.8 |
6.8 |
| Sodium Phosphate Monobasic (anhydrous) |
0.122 |
0.122 |
0.122 |
| Zinc Sulfate • 7H2O |
0.0000002 |
0.0000002 |
0.0000002 |
| Amino Acids |
| L-Alanine |
0.09 |
0.09 |
0.09 |
| L-Arginine (free base) |
0.05 |
0.05 |
0.05 |
| L-Asparagine • H2O |
0.02 |
0.02 |
0.02 |
| L-Aspartic Acid |
0.03 |
0.03 |
0.03 |
| L-Cysteine (free acid) |
0.04 |
0.04 |
0.04 |
| L-Cystine |
0.02 |
0.02 |
0.02 |
| L-Glutamic Acid |
0.0445 |
0.0445 |
0.0445 |
| L-Glutamine |
— |
0.292 |
— |
| Glycine |
0.05 |
0.05 |
0.05 |
| L-Histidine (free base) |
0.015 |
0.015 |
0.015 |
| L-Isoleucine |
0.05 |
0.05 |
0.05 |
| L-Leucine |
0.075 |
0.075 |
0.075 |
| L-Lysine • HCl |
0.08746 |
0.08746 |
0.08746 |
| L-Methionine |
0.015 |
0.015 |
0.015 |
| L-Phenylalanine |
0.025 |
0.025 |
0.025 |
| L-Proline |
0.03 |
0.03 |
0.03 |
| L-Serine |
0.01 |
0.01 |
0.01 |
| L-Threonine |
0.04 |
0.04 |
0.04 |
| L-Tryptophan |
0.01 |
0.01 |
0.01 |
| L-Tyrosine • 2Na • 2H2O |
0.05045 |
0.05045 |
0.05045 |
| L-Valine |
0.05 |
0.05 |
0.05 |
| Vitamins |
| Ascorbic Acid • Na |
0.00227 |
0.00227 |
0.00227 |
| D-Biotin |
0.0005 |
0.0005 |
0.0005 |
| Calciferol |
0.0001 |
0.0001 |
0.0001 |
| Choline Chloride |
0.0015 |
0.0015 |
0.0015 |
| Folic Acid |
0.001 |
0.001 |
0.001 |
| myo-Inositol |
0.002 |
0.002 |
0.002 |
| Menadione (sodium bisulfite) |
0.00001 |
0.00001 |
0.00001 |
| Niacinamide |
0.001 |
0.001 |
0.001 |
| D-Pantothenic Acid (hemicalcium) |
0.001 |
0.001 |
0.001 |
| Pyridoxal • HCl |
0.001 |
0.001 |
0.001 |
| Retinol Acetate |
0.0001 |
0.0001 |
0.0001 |
| Riboflavin |
0.0001 |
0.0001 |
0.0001 |
| Thiamine • HCl |
0.001 |
0.001 |
0.001 |
| DL-α-Tocopherol Phosphate • Na |
0.00001 |
0.00001 |
0.00001 |
| Vitamin B12 |
0.002 |
0.0002 |
0.0002 |
| Other |
| D-Glucose |
2 |
2 |
2 |
| Glutathione (reduced) |
0.00005 |
0.00005 |
0.00005 |
| Methyl Linoleate |
0.00003 |
0.00003 |
0.00003 |
| Phenol Red • Na |
— |
0.0107 |
0.0107 |
| Pyruvic Acid • Na |
0.025 |
0.025 |
0.025 |
| Add |
| L-Glutamine |
0.292 |
— |
0.292 |
| Sodium Bicarbonate |
— |
2.2 |
— |
| back to top |