Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Culture > Learning Center > Mediatech to Sigma® Cell Culture Cross-Reference Chart
In Stock and Ready to Ship
Mediatech to Sigma® Cell Culture Cross-Reference Chart
Mediatech, Inc. cell culture cross-reference chart for Sigma cell culture products. Find direct replacements for Mediatech's cell culture products.
Stock up now on cell culture media and reagents
We currently have large amounts of these products available for delivery today. You can depend on Sigma's media and reagents to meet your quality specifications and to help you to continue your research without delays from back orders or quality problems.
Request a quote on all your cell culture liquid media, serum and supplements.
To place an immediate order or view package sizes and prices, click on the Sigma product numbers in the below cross-reference table.
For more technical information visit our Cell Culture Home Page.
Mediatech to Sigma Product Cross-Reference
Liquid products may be directly compared in the table. Any differences in products are noted in the right–hand column.
Mediatech Catalog No.
Sigma Catalog No.
Product Description Product Differences
10-010-CM
M4655
Minimum Essential Medium Eagle Exact match. No differences.
10-010-CV
M4655
Minimum Essential Medium Eagle Exact match. No differences.
10-013-CM
D6429
Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
10-013-CV
D6429
Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
10-014-CM
D6046
Dulbecco's Modified Eagle's Medium - low glucose Exact match. No differences.
10-014-CV
D6046
Dulbecco's Modified Eagle's Medium - low glucose Exact match. No differences.
10-016-CM
I3390
Iscove's Modified Dulbecco's Medium Sigma is without L-glutamine, Mediatech contains L-glutamine
10-016-CV
I3390
Iscove's Modified Dulbecco's Medium Sigma is without L-glutamine, Mediatech contains L-glutamine
10-017-CM
D5796
Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
10-017-CV
D5796
Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
10-022-CV
M8042
Minimum Essential Medium Eagle Sigma is without L-glutamine, Mediatech contains L-glutamine
10-040-CM
R8758
RPMI 1640 Medium Exact match. No differences.
10-040-CV
R8758
RPMI 1640 Medium Exact match. No differences.
10-041-CM
R5886
RPMI 1640 Medium Sigma is without L-glutamine, Mediatech contains L-glutamine
10-041-CV
R5886
RPMI 1640 Medium Sigma is without L-glutamine, Mediatech contains L-glutamine
10-043-CV
R1383
RPMI 1640 Medium Sigma is powder, Mediatech is liquid
10-045-CV
L1518
L-15 Medium (Leibovitz) Exact match. No differences.
10-050-CV
M9309
McCoy's 5A Medium, Modified Exact match. No differences.
10-060-CV
M4530
Medium 199 Exact match. No differences.
10-070-CV
N6908
Nutrient Mixture F-10 Ham Exact match. No differences.
10-080-CM
N6658
Nutrient Mixture F-12 Ham Exact match. No differences.
10-080-CV
N6658
Nutrient Mixture F-12 Ham Exact match. No differences.
10-090-CV
D8062
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham Sigma contains 1.2 g/L sodium bicarbonate, Mediatech contains 2.4 g/L
10-092-CM
D8437
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham Sigma contains 1.2 g/L sodium bicarbonate, Mediatech contains 2.4 g/L
10-092-CV
D8437
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham Sigma contains 1.2 g/L sodium bicarbonate, Mediatech contains 2.4 g/L
13-100-CV
T3285
TNM-FH Insect Medium Exact match. No differences.
13-200-CV
G8142
Grace's Insect Medium Exact match. No differences.
13-410-CV
I5408
EX-CELL TiterHigh Medium Recommended alternative
13-415-CV
14405C
EX-CELL 405 Serum-Free Medium Recommended alternative.
13-901-CI
P5556
Pluronic F-68 solution Exact match. No differences.
15-010-CM
M2279
Minimum Essential Medium Eagle Exact match. No differences.
15-010-CV
M2279
Minimum Essential Medium Eagle Exact match. No differences.
15-012-CV
M4526
Minimum Essential Medium Eagle Exact match. No differences.
15-013-CM
D6546
Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
15-013-CV
D6546
Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
15-016-CV
I3390
Iscove's Modified Dulbecco's Medium Exact match. No differences.
15-017-CM
D5671
Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
15-017-CV
D5671
Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
15-040-CM
R0883
RPMI 1640 Medium Exact match. No differences.
15-040-CV
R0883
RPMI 1640 Medium Exact match. No differences.
15-041-CV
R5886
RPMI 1640 Medium Exact match. No differences.
15-100-CV
M8537
MCDB 131 Medium Sigma is powder, Mediatech is liquid
17-104-CI
R7513
RPMI 1640 Medium Exact match. No differences.
17-105-CV
R7509
RPMI 1640 Medium Exact match. No differences.
17-204-CI
D0422
Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
17-205-CV
D1145
Dulbecco's Modified Eagle's Medium - high glucose Sigma is without sodium pyruvate, Mediatech is with sodium pyruvate
20-021-CV
H4641
Hanks' Balanced Salt Solutions Exact match. No differences.
20-023-CV
H1641
Hanks' Balanced Salt Solutions Sigma is with phenol red, Mediatech is without phenol red
20-030-CV
D1283
Dulbecco's Phosphate Buffered Saline Exact match. No differences.
20-031-CV
D1408
Dulbecco's Phosphate Buffered Saline Exact match. No differences.
21-020-CM
H9269
Hanks' Balanced Salt Solutions Exact match. No differences.
21-020-CV
H9269
Hanks' Balanced Salt Solutions Exact match. No differences.
21-021-CM
H9394
Hanks' Balanced Salt Solutions Exact match. No differences.
21-021-CV
H9394
Hanks' Balanced Salt Solutions Exact match. No differences.
21-022-CM
H6648
Hanks' Balanced Salt Solutions Exact match. No differences.
21-022-CV
H6648
Hanks' Balanced Salt Solutions Exact match. No differences.
21-023-CM
H8264
Hanks' Balanced Salt Solutions Exact match. No differences.
21-023-CV
H8264
Hanks' Balanced Salt Solutions Exact match. No differences.
21-030-CM
D8662
Dulbecco's Phosphate Buffered Saline Exact match. No differences.
21-030-CV
D8662
Dulbecco's Phosphate Buffered Saline Exact match. No differences.
21-031-CM
D8537
Dulbecco's Phosphate Buffered Saline Exact match. No differences.
21-031-CV
D8537 Dulbecco's Phosphate Buffered Saline Exact match. No differences.
21-040-CM
P3813
Phosphate Buffered Saline, pH 7.4 Sigma is powder, Mediatech is liquid
21-040-CV
P3813
Phosphate Buffered Saline, pH 7.4 Sigma is powder, Mediatech is liquid
25-000-CI
S8636
Sodium pyruvate solution Exact match. No differences.
25-005-CI
G7513
L-Glutamine solution Exact match. No differences.
25-005-CV
G7513
L-Glutamine solution Exact match. No differences.
25-015-CI
G8541
Ala-Gln Exact match. No differences.
25-020-CI
M6895
MEM Vitamin Solution (100x) Exact match. No differences.
25-025-CI
M7145
MEM Non-essential Amino Acid Solution (100x) Exact match. No differences.
25-030-CI
M5550
MEM Amino Acids Solution (50W) Exact match. No differences.
25-035-CI
S8761
Sodium bicarbonate solution Exact match. No differences.
25-037-CI
G8769
D-Glucose Solution Exact match. No differences.
25-046-CI
H0262
HAT Media Supplement (50W) Hybri-Max. Exact match. No differences.
25-047-CI
H0137
HT Media Supplement (50x) Hybri-Max. Exact match. No differences.
25-050-CI
T4424
Trypsin solution from porcine pancreas Exact match. No differences.
25-051-CI
T3924
Trypsin-EDTA solution Exact match. No differences.
25-052-CI
T3924
Trypsin-EDTA solution Sigma is with sodium bicarbonate, Mediatech is without sodium bicarbonate
25-052-CV
T3924
Trypsin-EDTA solution Sigma is with sodium bicarbonate, Mediatech is without sodium bicarbonate
25-053-CI
T4049
Trypsin-EDTA solution Sigma is with sodium bicarbonate, Mediatech is without sodium bicarbonate
25-054-CI
T4674
Trypsin solution from porcine pancreas, 10x Sigma is with phenol red, Mediatech is without phenol red
25-055-CM
W3500
Water, Tissue Culture Grade Exact match. No differences.
25-055-CV
W3500
Water, Tissue Culture Grade Exact match. No differences.
25-056-CI
C5789
Cell Dissociation Solution, Non-enzymatic Recommended alternative.
25-060-CI
H0887
HEPES solution Exact match. No differences.
25-072-CI
H8889
Histopaque 1077 HybriMax Exact match. No differences.
25-072-CV
H8889
Histopaque 1077 HybriMax Exact match. No differences.
25-800-CR
I3146
ITS Liquid Media Supplement (100x) Exact match. No differences.
25-900-CI
T8154
Trypan Blue solution Exact match. No differences.
30-001-CI
P4458
Penicillin - Streptomycin Solution (50x) Stabilized Exact match. No differences.
30-002-CI
P4333
Penicillin - Streptomycin Solution (100x) Stabilized Exact match. No differences.
30-003-CF
A2942
Amphotericin B preparation Exact match. No differences.
30-004-CI
A5955
Antibiotic Antimycotic Solution, stabilized (100x) Exact match. No differences.
30-005-CR
G1397
Gentamicin solution Exact match. No differences.
30-006-CF
K0129
Kanamycin solution from Streptomyces kanamyceticus Sigms is 10 mg/ml, Mediatech is 5 mg/ml
30-009-CI
G1146
L-Glutamine-Penicillin-Streptomycin solution Exact match. No differences.
30-234-CI
G8168
G418 disulfate solution Exact match. No differences.
30-234-CR
G8168
G418 disulfate solution Exact match. No differences.
35-010-CV
F0926
Fetal Bovine Serum, USDA approved Exact match. No differences.
35-011-CV
12306C
Fetal Bovine Serum, Heat Inactivated Exact match. No differences.
35-015-CV
F2442
Fetal Bovine Serum Exact match. No differences.
35-016-CV
F4135
Fetal Bovine Serum, Heat Inactivated Exact match. No differences.
35-022-CV
12133C
Calf Serum Recommended alternative.
35-023-CM
C8056
Calf Serum Exact match. No differences.
35-030-CV
H1270
Horse Serum Exact match. No differences.
35-040-CV
P9783
Porcine Serum Exact match. No differences.
35-060-CI
H4522
Human Serum Exact match. No differences.
40-200-CV
C5467
CHO Medium Exact match. No differences.
40-500-CM
14660C
EX-CELL Sp2/0 Serum-Free Medium Recommended alternative.
40-510-CM
14660C
EX-CELL Sp2/0 Serum-Free Medium Recommended alternative.
40-600-CV
14571C
EX-CELL 293 Serum-Free Medium Recommended alternative.
40-700-CV
M3553
EX-CELL MDBK-GM Medium Recommended alternative.
50-003-PB
D7777
Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
50-003-PC
D7777
Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
50-004-PB
G6148
Glasgow Minimum Essential Medium Exact match. No differences.
50-005-PB
B9638
Basal Medium Eagle, Earle's salts Exact match. No differences.
50-005-PC
B9638
Basal Medium Eagle, Earle's salts Exact match. No differences.
50-010-PB
M0268
Minimum Essential Medium Eagle Exact match. No differences.
50-010-PC
M0268
Minimum Essential Medium Eagle Exact match. No differences.
50-012-PB
M0894
Minimum Essential Medium Eagle Exact match. No differences.
50-012-PC
M0894
Minimum Essential Medium Eagle Exact match. No differences.
50-013-PB
D5648
Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
50-013-PC
D5648
Dulbecco's Modified Eagle's Medium - high glucose Exact match. No differences.
50-014-PB
D5523
Dulbecco's Modified Eagle's Medium - low glucose Exact match. No differences.
50-014-PC
D5523
Dulbecco's Modified Eagle's Medium - low glucose Exact match. No differences.
50-016-PB
I7633
Iscove's Modified Dulbecco's Medium Exact match. No differences.
50-016-PC
I7633
Iscove's Modified Dulbecco's Medium Exact match. No differences.
50-019-PB
M4642
Minimum Essential Medium Eagle Exact match. No differences.
50-019-PC
M4642
Minimum Essential Medium Eagle Exact match. No differences.
50-020-PB
R6504
RPMI 1640 Medium Exact match. No differences.
50-020-PC
R6504
RPMI 1640 Medium Exact match. No differences.
50-025-PB
M4892
McCoy's 5A Medium Exact match. No differences.
50-025-PC
M4892
McCoy's 5A Medium Exact match. No differences.
50-030-PB
N6635
Nutrient Mixture F-10 Ham Exact match. No differences.
50-030-PC
N6635
Nutrient Mixture F-10 Ham Exact match. No differences.
50-040-PB
N6760
Nutrient Mixture F-12 Ham Exact match. No differences.
50-040-PC
N6760
Nutrient Mixture F-12 Ham Exact match. No differences.
50-045-PB
L4386
L-15 Medium (Leibovitz) Exact match. No differences.
50-045-PC
L4386
L-15 Medium (Leibovitz) Exact match. No differences.
50-050-BC
M5017
Medium 199 Exact match. No differences.
50-050-PB
M5017
Medium 199 Exact match. No differences.
50-051-PB
M0393
Medium 199 Exact match. No differences.
50-051-PC
M0393
Medium 199 Exact match. No differences.
51-010-PB
M0769
Minimum Essential Medium Eagle Exact match. No differences.
51-010-PC
M0769
Minimum Essential Medium Eagle Exact match. No differences.
51-034-RR
H3375
HEPES Exact match. No differences.
55-021-PB
H2387
Hanks' Balanced Salts Exact match. No differences.
55-021-PC
H2387
Hanks' Balanced Salts Exact match. No differences.
55-022-PB
H4891
Hanks' Balanced Salts, Modified Exact match. No differences.
55-022-PC
H4891
Hanks' Balanced Salts, Modified Exact match. No differences.
55-031-PB
D5652
Dulbecco's Phosphate Buffered Saline Exact match. No differences.
55-031-PC
D5652
Dulbecco's Phosphate Buffered Saline Exact match. No differences.
61-030-RM
G8540
L-Glutamine Exact match. No differences.
61-030-RO
G8540
L-Glutamine Exact match. No differences.
61-030-RR
G8540
L-Glutamine Exact match. No differences.
61-034-RM
H4034
HEPES Exact match. No differences.
61-034-RO
H4034
HEPES Exact match. No differences.
61-034-RR
H4034
HEPES Exact match. No differences.
61-053-MU
P7794
Penicillin G potassium salt Exact match. No differences.
61-065-RO
S5761
Sodium Bicarbonate Exact match. No differences.
61-065-RR
S5761
Sodium Bicarbonate Exact match. No differences.
61-088-RM
S9137
Streptomycin sulfate salt Exact match. No differences.
61-098-RA
G1264
Gentamicin sulfate salt Exact match. No differences.
61-098-RF
G1264
Gentamicin sulfate salt Exact match. No differences.
61-161-RM
P1300
Pluronic F-68 Exact match. No differences.
61-176-RG
K1377
Kanamycin sulfate Exact match. No differences.
61-196-RM
F8016
Ficoll 400 Exact match. No differences.
61-196-RO
F8016
Ficoll 400 Exact match. No differences.
61-201-RM
P4963
PRIMATONE RL Exact match. No differences.
61-201-RO
P4963
PRIMATONE RL Exact match. No differences.
61-233-RM
T1503
Trizma base Exact match. No differences.
61-233-RO
T1503
Trizma base Exact match. No differences.
61-233-RR
T1503
Trizma base Exact match. No differences.
61-233-RT
T1503
Trizma base Exact match. No differences.
61-234-RF
A1720
G418 disulfate salt Exact match. No differences.
61-234-RG
A1720
G418 disulfate salt Exact match. No differences.
61-234-RK
A1720
G418 disulfate salt Exact match. No differences.
61-238-RH
A0166
Ampicillin sodium salt Exact match. No differences.
61-238-RM
A0166
Ampicillin sodium salt Exact match. No differences.
61-239-RI
C1863
Chloramphenicol, Biotechnology Performance Certified Exact match. No differences.
61-241-RG
N6386
Neomycin trisulfate salt hydrate Exact match. No differences.
61-241-RM
N6386
Neomycin trisulfate salt hydrate Exact match. No differences.
61-242-RG
T7760
Tetracycline hydrochloride Exact match. No differences.
61-277-RF
17850
Ciprofloxacin Exact match. No differences.
61-277-RG
17850
Ciprofloxacin Exact match. No differences.
61-385-RA
P8833
Puromycin dihydrochloride Exact match. No differences.
90-022-PB
R8755
RPMI 1640 Medium Sigma contains L-glutamine, Mediatech is without L-glutamine
90-022-PC
R8755
RPMI 1640 Medium Sigma contains L-glutamine, Mediatech is without L-glutamine
90-050-PB
M3769
Medium 199 Exact match. No differences.
90-050-PC
M3769
Medium 199 Exact match. No differences.
90-113-PB
D5030
Dulbecco's Modified Eagle's Medium - low glucose Exact match. No differences.
 back to top