| Cell Name |
Product # |
Species of Origin
|
Basal Media to Supply (Europe Only)
|
| Hep G2 |
85011430 |
Human |
M2279, Minimum Essential Medium Eagle |
| THP 1 |
88081201 |
Human |
|
| MRC–5 |
84101801 |
Human |
M2279, Minimum Essential Medium Eagle |
| HeLa |
93021013 |
Human |
M2279, Minimum Essential Medium Eagle |
| A549 |
86012804 |
Human |
D6546, Dulbecco's Modified Eagle's Medium – high glucose |
| CHO–K1 |
85051005 |
Hamster |
N4888, Nutrient Mixture F–12 Ham |
| Vero |
84113001 |
Monkey |
D6546, Dulbecco's Modified Eagle's Medium – high glucose |
| U937 |
85011440 |
Human |
|
| RAW 264.7 |
91062702 |
Mouse |
D6546, Dulbecco's Modified Eagle's Medium – high glucose |
| SH–SY5Y |
94030304 |
Human |
N4888:M2279 (1:1), Nutrient Mixture F–12 Ham:Minimum Essential Medium Eagle |
| MCF7 |
86012803 |
Human |
M2279, Minimum Essential Medium Eagle |
| K562 |
89121407 |
Human |
|
| NSO – Exclusive |
85110503 |
Mouse |
|
| C2C12 |
91031101 |
Mouse |
D6546, Dulbecco's Modified Eagle's Medium – high glucose |
| MDCK |
84121903 |
Dog |
M2279, Minimum Essential Medium Eagle |
| BHK 21 (clone 13) |
85011433 |
Hamster |
G5154, Glasgow Minimum Essential Medium |
| A431 |
85090402 |
Human |
M2279, Minimum Essential Medium Eagle |
| COS–1 |
88031701 |
Monkey (SV40 transformed) |
D6546, Dulbecco's Modified Eagle's Medium – high glucose |
| LNCap.FGC |
89110211 |
Human |
|
| U373 MG |
89081403 |
Human |
M2279, Minimum Essential Medium Eagle |
| MDA–MB–231 |
92020424 |
Human |
L5520, L–15 Medium (Leibovitz) |
| SP2/0–Ag14 |
85072401 |
Mouse |
|
| J774.2 – Exclusive |
85011428 |
Mouse |
D6546, Dulbecco's Modified Eagle's Medium – high glucose |
| Fao – Exclusive |
89042701 |
Rat |
VOUCHER |
| ISHIKAWA – Exclusive |
99040201 |
Human |
M2279, Minimum Essential Medium Eagle |
| OE33 – Exclusive |
96070808 |
Human |
|
| RPMI 8866 – Exclusive |
95041316 |
Human |
|
| Hep–2C (HeLa Derivative) – Exclusive |
85020207 |
Human |
|
| NB2–11 – Exclusive |
97041101 |
Rat |
|
| A2780cis – Exclusive |
93112517 |
Human |
|
| MDBK |
90050801 |
Bovine |
M2279, Minimum Essential Medium Eagle |
| L929 |
85011425 |
Mouse |
D6546, Dulbecco's Modified Eagle's Medium – high glucose |
| EL4 |
85023105 |
Mouse |
D6546, Dulbecco's Modified Eagle's Medium – high glucose |
| TF1 |
93022307 |
Human |
|