Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Culture > Serum Free Media > CHO Platform > CHO DHFR Medium
Cell Culture
CHO DHFR Medium
Get Maximum Results from Your CHO DHFR- System with New CHO DHFR- Medium, Animal Component-Free
(Product Code C 8862)
This latest addition to the growing line of animal component-free CHO media from Sigma-Aldrich has been designed to provide outstanding cell growth and recombinant protein productivity in Chinese hamster ovary (CHO) cells that are engineered to contain the dihydrofolate reductase (DHFR) amplification system. Contains recombinant human insulin, Pluronic® F-68, and plant-based hydrolysates
CHO DHFR- Medium provides the following benefits:
  • Animal Component-free — contains no transferrin or other products of animal origin
  • Designed for DHFR- CHO clones — no hypoxanthine or thymidine
  • Outperforms the competition in terms of cell growth and productivity
  • Supports proper glycosylation of recombinant proteins
  • Exceptional performance in other selection systems, too
Product Code Description Pack Size
C8862 CHO DHFR- Medium, Animal Component-free 1 L
6 x 1 L
Life Science Quarterly: Volume 4, Issue 3, Summer 2003