Cell Culture
TiterHigh Sf Insect Medium
|
|
|
TiterHigh Sf Insect Medium, Animal Component-Free (I 5408) is the latest addition to the fastest growing line of serum-free media in the industry.
With TiterHigh Sf Insect Medium You Get:
- Exceptional cell growth - viable cell densities of up to 2 x 107 cells/mL can be achieved in shaker cultures, with little or no adaptation required
- Excellent baculoviral expression - beats the competition in terms of per cell productivity with wild type and recombinant baculoviruses
- No serum or any animal-derived components - satisfies regulatory requirements and simplifies downstream processing
|
|
Sƒ21 cells were grown in TiterHigh Sf Insect Medium and the leading copetitor's medium in shaker flasks, at 27° C with a shaker speed of 130 rpm. As is shown, TiterHigh Sf supports much higher viable cell densities than the competition. |
|
|
Sƒ21 cells grown in TiterHigh Sf Insect Medium and the leading competitor's medium were infected with a recombinant baculovirus expressing b-galactosidase. Cells were infected at a cell concentration of 1x106 cells per mL, with a multiplicity of infection of 5 virus particles per cell. Enzyme levels were measured at days 2, 3, and 4 post-infection. As is demonstrated, TiterHigh Sf beats the competition in terms of per cell baculovirus productivity. |
| Product Code |
Description |
Package Size |
Price US $ |
| I 5408 |
TiterHigh Sf Insect Medium |
1 L 6 X 1 L |
53.45 240.55 |
| Product Code |
Description |
Package Size |
Price US $ |
| L 3287 |
ESCORT™ IV Transfection Reagent |
1 mL |
86.05 |
| E 9770 |
ESCORT™ Transfection Reagent |
.25 mL .5 mL |
44.35 78.95 |
| G 1397 |
Gentamicin Solution |
10 mL 100 mL |
41.00 340.35 |
back to top Cell Culture Home