Sigma-Aldrich
Life Science > Cell Culture > Specialty Media > MCDB Media
Cell Culture
MCDB Media
MCDB media were developed in the 1970s and 1980s, primarily by Ham and associates,   for the low-protein and serum free growth of specific cell types using hormones, growth factors, trace elements and/or low levels of dialyzed fetal bovine serum protein (FBSP).  Each MCDB medium was formulated (quantitatively and qualitatively) to provide a defined and optimally balanced nutritional environment that selectively promoted the growth of a specific cell line.
  • MCDB 105 and 110 are modifications of MCDB 104 medium, optimized for long-term survival and rapid clonal growth of human diploid fibroblast-like cells (WI-38, MRC-5, IMR-90) and low passaged human foreskin fibroblasts using FBSP, hormone, and growth factor supplements.
  • MCDB 151, 201 and 302 are modifications of Ham's nutrient mixture F-12, designed for the growth of human keratinocytes, clonal growth of chicken embryo fibroblasts (CEF) and Chinese hamster ovary (CHO) cells using low levels of FBSP, extensive trace elements or no serum protein.
View our MCDB Media selection.