Sigma-Aldrich
Life Science > Cell Culture > TrypZean Solution
Cell Culture
TrypZean Solution
Non-animal recombinant trypsin solution for dissociating attached cells
TrypZean Solution, 1X (Product Code T 3449) is the first animal component-free trypsin solution optimized for cell dissociation.
Percent Viability
TrypZean Solution, 1X provides the following features and benefits:
  • Animal component-free
    TrypZean eliminates the risk of viruses, BSE, or other potential adventitious agents.
  • TrypZean is recombinant trypsin
    Using the same enzyme (with the same kinetics) for cell detachment means minimal protocol changes for you.
  • Enzyme inhibition
    Soybean trypsin inhibitor and other inhibitors work the same with TrypZean as they do with native trypsins (on a weight-to-weight basis).
  • High purity
    TrypZean provides increased specificity and eliminates contaminating activities found in lower purity enzymes.
  • Convenience
    TrypZean Solution is formulated at the optimal concentration to dissociate adherent cells.
Kinetics of TrypZean
Trypsin vs. TrypZean
Product Name
Product # 
Package Size
TrypZean™ Solution, 1X
T 3449
100 ml
500 ml
back to top
Back to Cell Culture Home