Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Cell Design Studio Highlighted Projects
Click here to learn about some of our highlighted projects.
 Cardiomyocytes Beating
ZFNs were used to target the endogenous NKX2-5 allele and introduce a GFP lineage marker following a ribosomal skip sequence in iPSCs. GFP was not expressed following clonal selection, but was robustly expressed following directed differentiation into cardiomyocytes, in which NKX2-5 is expressed. The video shows beating cardiomyocytes expressing GFP under the control of the NKX2-5 locus.
 STAT1 and STAT3 Localizing to the Nucleus
ZFNs were used to target the endogenous STAT1 and STAT3 alleles in A549 cells to produce GFP-tagged STAT1 and RFP-tagged STAT3, respectively. Here, the modified cells have been stimulated with IFN-γ and IL-6 and imaged over time to monitor the nuclear localization of STAT1 (green) and STAT3 (red), shown on the right. The left panel shows a DIC image of the same field with DAPI nuclear stain in blue.
 Tubulin in Response to Paclitaxel
ZFNs were used to target the endogenous TUBA1B locus and produce an RFP-tagged TUBA1B allele in MCF10A cells. When indicated, the modified cells were treated with paclitaxel to inhibit microtubule dynamics, visualized by the RFP-α-tubulin. The left panel shows a DIC image of the same field.