| Sigma-Aldrich® BIS-TRIS Propane Physical Properties |
| Synonyms |
1,3-Bis[tris(hydroxymethyl)methylamino]propane |
 |
| Molecular Formula |
CH2[CH2NHC(CH2OH)3]2 |
Beilstein Registry |
1786109 |
Useful pH range |
6.3 - 9.5 |
| Molecular Weight |
282.33 |
MDL Number |
MFCD00004689 |
pKa (at 25) |
(1) 6.8, (2) 9.0 |
| CAS Number |
64431-96-5 |
EG/EC Number |
264-899-3 |
Melting Point |
164 - 165 °C(lit.) |
| Sigma-Aldrich BIS-TRIS Propane Specifications Comparison |
| |
B6755 |
B4679 |
B9410 |
| Grade |
≥99.0% (titration) |
BioReagent, cell culture tested, ≥99.0% (titration) |
BioXtra, ≥99.0% (titration) |
| Purity |
≥99.0% |
≥99.0% |
>99.0% |
| Reagent Testing |
| |
B6755 |
B4679 |
B9410 |
Enhanced Quality Profile Buffers |
X |
X |
X |
| 260NM |
|
|
≤0.1 |
| 280NM |
|
|
≤0.05 |
| 290NM |
≤0.05 |
≤0.05 |
|
| IR |
X |
X |
X |
| pH |
|
|
10.0 - 12.0 (20 °C, 1 M in H2O) |
| Karl Fischer |
≤0.2% |
≤0.2% |
≤0.2% |
| Insoluble Matter |
|
|
≤0.1% |
| Residue on Ignition |
|
|
≤0.1% |
| Life Science Testing |
| |
B6755 |
B4679 |
B9410 |
| Endotoxin |
|
≤1 eu/mg |
|
| SPC |
|
≤100 cfu/g |
|
| Cell test |
|
X |
|
| RNAse |
|
X |
|
| DNAse |
|
X |
|
| Nickase |
|
X |
|
| Protease |
|
X |
|
| Proton NMR Spectrum |
≤0.5% Tris |
≤0.5% Tris |
≤0.5% Tris |
| Trace Elemental Impurities |
| |
B6755 |
B4679 |
B9410 |
| Ammonia(NH4) |
|
|
≤0.05% |
| Sodium(Na) |
|
|
≤0.005% |
| Phosphorus(P) |
|
|
≤0.005% |
| Chloride(Cl) |
|
|
≤0.05% |
| Sulfate(SO4) |
|
|
≤0.05% |
| Aluminum(Al) |
|
|
≤0.0005% |
| Calcium(Ca) |
|
|
≤0.0005% |
| Copper(Cu) |
|
|
≤0.0005% |
| Iron(Fe) |
|
|
≤0.0005% |
| Potassium(K) |
|
|
≤0.005% |
| Lead(Pb) |
≤0.0005% |
≤0.0005% |
≤0.001% |
| Magnesium(Mg) |
|
|
≤0.0005% |
| Zinc(Zn) |
|
|
≤0.0005% |
| |
back to top back to Biological Buffers |
|