| Sigma-Aldrich® BIS-TRIS Physical Properties |
| Synonyms |
2,2-Bis(hydroxymethyl)-2,2',2"-nitrilotriethanol 2-Bis(2-hydroxyethyl)amino-2-(hydroxymethyl)-1,3-propanediol Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane |
 |
| Molecular Formula |
C8H19NO5 |
Beilstein Registry |
2205275 |
Useful pH range |
5.8 - 7.2 |
| Molecular Weight |
209.24 |
MDL Number |
MFCD00002853 |
pKa (at 25) |
6.5 |
| CAS Number |
6976-37-0 |
EG/EC Number |
230-237-7 |
pKa (at 37) |
6.36 |
| Sigma-Aldrich BIS-TRIS Specifications Comparison |
| |
B9754 |
B4429 |
B7535 |
14879 |
| Grade |
≥98.0% (titration) |
BioReagent, cell culture tested, suitable for insect cell culture, ≥98% (titration) |
BioXtra, ≥98.0% (titration) |
BioUltra, ≥99.0% (NT) |
| Purity |
≥98.0% |
≥98.0% |
>98.0% |
≥99.0% |
| Reagent Testing |
| |
B9754 |
B4429 |
B7535 |
14879 |
Enhanced Quality Profile Buffers |
X |
X |
X |
|
| 280NM |
|
|
|
≤0.15 |
| IR |
X |
X |
X |
|
| pH |
9.0 - 10.1 |
|
9.0 - 11.0 (20 °C, 1 M in H2O) |
9.5 - 11.0 (25 °C, 1 M in H2O) |
| Karl Fischer |
≤1% |
≤1% |
≤1% |
|
| LOD |
|
|
|
≤0.5% |
| Insoluble Matter |
|
|
<0.1% |
filter test |
| Residue on Ignition |
|
|
<0.1% |
≤0.3% |
| Life Science Testing |
| |
B9754 |
B4429 |
B7535 |
14879 |
| Endotoxin |
|
≤1 eu/mg |
|
|
| SPC |
|
≤100 cfu/g |
|
|
| Cell test |
|
X |
|
|
| Insect cell test |
|
X |
|
|
| RNAse |
|
X |
|
|
| DNAse |
|
X |
|
|
| Nickase |
|
X |
|
|
| Protease |
|
X |
|
|
| Trace Elemental Impurities |
| |
B9754 |
B4429 |
B7535 |
14879 |
| Ammonia(NH4) |
|
|
<0.05% |
|
| Sodium(Na) |
|
|
<0.005% |
≤0.005% |
| Phosphorus(P) |
|
|
<0.005% |
|
| Chloride(Cl) |
|
|
<0.05% |
≤0.005% |
| Sulfate(SO4) |
|
|
<0.05% |
≤0.005% |
| Aluminum(Al) |
|
|
<0.0005% |
≤0.0005% |
| Arsenic(As) |
|
|
|
≤0.00001% |
| Barium(Ba) |
|
|
|
≤0.0005% |
| Bismuth(Bi) |
|
|
|
≤0.0005% |
| Calcium(Ca) |
|
|
<0.0005% |
≤0.001% |
| Cadmium(Cd) |
|
|
|
≤0.0005% |
| Cobalt(Co) |
|
|
|
≤0.0005% |
| Chromium(Cr) |
|
|
|
≤0.0005% |
| Copper(Cu) |
|
|
<0.0005% |
≤0.0005% |
| Iron(Fe) |
|
|
<0.0005% |
≤0.0005% |
| Potassium(K) |
|
|
<0.005% |
≤0.005% |
| Lithium(Li) |
|
|
|
≤0.0005% |
| Magnesium(Mg) |
|
|
<0.0005% |
≤0.0005% |
| Manganese(Mn) |
|
|
|
≤0.0005% |
| Molybdenum(Mo) |
|
|
|
≤0.0005% |
| Nickel(Ni) |
|
|
|
≤0.0005% |
| Lead(Pb) |
≤0.0005% |
≤0.0005% |
<0.001% |
≤0.0005% |
| Strontium(Sr) |
|
|
|
≤0.0005% |
| Zinc(Zn) |
|
|
<0.0005% |
≤0.0005% |
| |
back to top back to Biological Buffers |
|